Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103940692-103941210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RD3L
Entrez Gene ID
647286
Gene Name
retinal degeneration 3-like
Gene Aliases
TDRD9-AS1, TDRD9AS1
UniGene
Hs.632325
Gene Chromosome Location
Chr.14: 103940426-103942308 on Build GRCh38
Transcripts
NM_001257268.1 NM_153046.2 XM_005267309.3 XM_006720019.3 XM_006720020.3 XM_011536397.2 XM_011536398.2 XM_011536400.2 XM_017020955.1

Gene Symbol
TDRD9
Entrez Gene ID
122402
Gene Name
tudor domain containing 9
Gene Aliases
C14orf75, HIG-1, NET54
UniGene
Hs.21454
Gene Chromosome Location
Chr.14: 103928343-104052667 on Build GRCh38
Transcripts
NM_001257268.1 NM_153046.2 XM_005267309.3 XM_006720019.3 XM_006720020.3 XM_011536397.2 XM_011536398.2 XM_011536400.2 XM_017020955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5478637 c.377T>G p.F126C Chr.14: 103941026-103941026 on Build GRCh38
3386425 c.412G>A p.D138N Chr.14: 103940991-103940991 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs552593437 Chr.14: 103940785-103940785 on Build GRCh38
rs901992296 Chr.14: 103940984-103940984 on Build GRCh38
rs1003230761 Chr.14: 103940966-103940966 on Build GRCh38
rs765247695 Chr.14: 103940875-103940875 on Build GRCh38
rs919167813 Chr.14: 103940885-103940885 on Build GRCh38
rs763336035 Chr.14: 103941045-103941045 on Build GRCh38
rs917046897 Chr.14: 103940802-103940802 on Build GRCh38
rs1032645745 Chr.14: 103941078-103941078 on Build GRCh38
rs987988087 Chr.14: 103940724-103940724 on Build GRCh38
rs1040456638 Chr.14: 103940969-103940969 on Build GRCh38
rs3830949 Chr.14: 103940797-103940800 on Build GRCh38
rs72712880 Chr.14: 103941047-103941047 on Build GRCh38
rs563086202 Chr.14: 103940790-103940790 on Build GRCh38
rs904838714 Chr.14: 103940941-103940941 on Build GRCh38
rs1023410962 Chr.14: 103940930-103940930 on Build GRCh38
rs1047677693 Chr.14: 103940914-103940914 on Build GRCh38
rs568438434 Chr.14: 103940887-103940887 on Build GRCh38
rs539919880 Chr.14: 103941123-103941123 on Build GRCh38
rs758168162 Chr.14: 103940902-103940902 on Build GRCh38
rs35337422 Chr.14: 103940906-103940906 on Build GRCh38
rs17101946 Chr.14: 103940776-103940776 on Build GRCh38
rs767359440 Chr.14: 103940925-103940925 on Build GRCh38
rs901445731 Chr.14: 103940876-103940876 on Build GRCh38
rs547372848 Chr.14: 103940917-103940917 on Build GRCh38
rs1005703241 Chr.14: 103940924-103940924 on Build GRCh38
rs118060694 Chr.14: 103941017-103941017 on Build GRCh38
rs796576988 Chr.14: 103940801-103940804 on Build GRCh38
rs188189727 Chr.14: 103940791-103940791 on Build GRCh38
rs747006758 Chr.14: 103940958-103940959 on Build GRCh38
rs759177625 Chr.14: 103940865-103940865 on Build GRCh38
rs930225846 Chr.14: 103941155-103941155 on Build GRCh38
rs1001558113 Chr.14: 103941053-103941053 on Build GRCh38
rs776279179 Chr.14: 103940846-103940846 on Build GRCh38
rs766715331 Chr.14: 103941056-103941056 on Build GRCh38
rs1034907914 Chr.14: 103941010-103941010 on Build GRCh38
rs773559421 Chr.14: 103941044-103941044 on Build GRCh38
rs556423772 Chr.14: 103941150-103941150 on Build GRCh38
rs904103103 Chr.14: 103941042-103941042 on Build GRCh38
rs947461922 Chr.14: 103940766-103940766 on Build GRCh38
rs987955171 Chr.14: 103941029-103941029 on Build GRCh38
rs942733285 Chr.14: 103940853-103940853 on Build GRCh38
rs34264357 Chr.14: 103940886-103940887 on Build GRCh38
rs374574508 Chr.14: 103940919-103940919 on Build GRCh38
rs942674356 Chr.14: 103941114-103941114 on Build GRCh38
rs968736613 Chr.14: 103940767-103940767 on Build GRCh38
rs922817821 Chr.14: 103941148-103941148 on Build GRCh38
rs913284083 Chr.14: 103940738-103940738 on Build GRCh38
rs1041257259 Chr.14: 103940859-103940859 on Build GRCh38
rs749356254 Chr.14: 103941069-103941069 on Build GRCh38
rs780319617 Chr.14: 103941004-103941004 on Build GRCh38
rs889148332 Chr.14: 103940922-103940922 on Build GRCh38
rs1009727065 Chr.14: 103940998-103940998 on Build GRCh38
rs991212085 Chr.14: 103941151-103941151 on Build GRCh38
rs1024073565 Chr.14: 103941156-103941156 on Build GRCh38
rs1021222104 Chr.14: 103940739-103940739 on Build GRCh38
rs956619154 Chr.14: 103941022-103941022 on Build GRCh38
rs956503977 Chr.14: 103940716-103940716 on Build GRCh38
rs761917610 Chr.14: 103940900-103940900 on Build GRCh38
rs568830436 Chr.14: 103940827-103940827 on Build GRCh38
rs142355459 Chr.14: 103940820-103940820 on Build GRCh38
rs756331635 Chr.14: 103940975-103940975 on Build GRCh38
rs750597417 Chr.14: 103940963-103940963 on Build GRCh38

View Full Product Details