Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 22826353-22825843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPA1
Entrez Gene ID
123606
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 1
Gene Aliases
FSP3, SPG6
UniGene
Hs.511797
Gene Chromosome Location
Chr.15: 22786225-22829789 on Build GRCh38
Transcripts
NM_001142275.1 NM_144599.4

View Full Product Details