Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65286264-65286766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP16
Entrez Gene ID
54956
Gene Name
poly(ADP-ribose) polymerase family member 16
Gene Aliases
ARTD15, C15orf30, pART15
UniGene
Hs.30634
Gene Chromosome Location
Chr.15: 65231691-65286886 on Build GRCh38
Transcripts
NM_001316943.1 NM_001316944.1 NM_017851.5 XM_006720589.2 XM_006720590.2 XM_006720592.2 XM_011521742.2 XM_017022386.1 XM_017022387.1 XM_017022388.1 XM_017022389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7155877 c.147C>T p.R49R Chr.15: 65286280-65286280 on Build GRCh38
6522185 c.68G>T p.R23L Chr.15: 65286359-65286359 on Build GRCh38
6263257 c.17G>A p.W6* Chr.15: 65286410-65286410 on Build GRCh38
4542525 c.31G>A p.E11K Chr.15: 65286396-65286396 on Build GRCh38
94929 c.11C>T p.S4L Chr.15: 65286416-65286416 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756176502 Chr.15: 65286376-65286376 on Build GRCh38
rs556715772 Chr.15: 65286533-65286533 on Build GRCh38
rs986973538 Chr.15: 65286427-65286427 on Build GRCh38
rs752988998 Chr.15: 65286370-65286370 on Build GRCh38
rs943306889 Chr.15: 65286462-65286462 on Build GRCh38
rs867686181 Chr.15: 65286369-65286369 on Build GRCh38
rs749285238 Chr.15: 65286310-65286310 on Build GRCh38
rs1045648231 Chr.15: 65286637-65286637 on Build GRCh38
rs974458945 Chr.15: 65286580-65286580 on Build GRCh38
rs544136455 Chr.15: 65286516-65286516 on Build GRCh38
rs558438594 Chr.15: 65286470-65286470 on Build GRCh38
rs748193657 Chr.15: 65286298-65286298 on Build GRCh38
rs766294840 Chr.15: 65286402-65286402 on Build GRCh38
rs911372827 Chr.15: 65286553-65286553 on Build GRCh38
rs1001337078 Chr.15: 65286673-65286673 on Build GRCh38
rs751888722 Chr.15: 65286346-65286346 on Build GRCh38
rs201444053 Chr.15: 65286454-65286454 on Build GRCh38
rs751385143 Chr.15: 65286463-65286464 on Build GRCh38
rs973584504 Chr.15: 65286380-65286380 on Build GRCh38
rs12906848 Chr.15: 65286733-65286733 on Build GRCh38
rs1026452056 Chr.15: 65286549-65286549 on Build GRCh38
rs745494229 Chr.15: 65286321-65286321 on Build GRCh38
rs1038824411 Chr.15: 65286449-65286449 on Build GRCh38
rs757220100 Chr.15: 65286392-65286392 on Build GRCh38
rs61736628 Chr.15: 65286378-65286378 on Build GRCh38
rs1020699821 Chr.15: 65286744-65286744 on Build GRCh38
rs1045079209 Chr.15: 65286717-65286718 on Build GRCh38
rs895844461 Chr.15: 65286711-65286711 on Build GRCh38
rs755181612 Chr.15: 65286287-65286287 on Build GRCh38
rs777686630 Chr.15: 65286307-65286307 on Build GRCh38
rs754841469 Chr.15: 65286522-65286522 on Build GRCh38
rs943095573 Chr.15: 65286437-65286437 on Build GRCh38
rs999104619 Chr.15: 65286334-65286334 on Build GRCh38
rs987107235 Chr.15: 65286446-65286446 on Build GRCh38
rs1012086739 Chr.15: 65286299-65286299 on Build GRCh38
rs191111106 Chr.15: 65286712-65286712 on Build GRCh38
rs970125178 Chr.15: 65286328-65286328 on Build GRCh38
rs369623814 Chr.15: 65286565-65286565 on Build GRCh38
rs142349271 Chr.15: 65286734-65286734 on Build GRCh38
rs552656918 Chr.15: 65286710-65286710 on Build GRCh38
rs773831553 Chr.15: 65286339-65286339 on Build GRCh38
rs1001556059 Chr.15: 65286515-65286515 on Build GRCh38
rs766723091 Chr.15: 65286345-65286345 on Build GRCh38
rs764118933 Chr.15: 65286474-65286474 on Build GRCh38
rs1010655127 Chr.15: 65286528-65286528 on Build GRCh38
rs946576160 Chr.15: 65286646-65286646 on Build GRCh38
rs770692681 Chr.15: 65286313-65286313 on Build GRCh38
rs949969863 Chr.15: 65286631-65286631 on Build GRCh38
rs1047860365 Chr.15: 65286616-65286616 on Build GRCh38
rs987072764 Chr.15: 65286559-65286559 on Build GRCh38
rs1024086164 Chr.15: 65286508-65286508 on Build GRCh38
rs934746291 Chr.15: 65286708-65286708 on Build GRCh38
rs903175314 Chr.15: 65286495-65286495 on Build GRCh38
rs767641011 Chr.15: 65286354-65286354 on Build GRCh38
rs376584427 Chr.15: 65286740-65286740 on Build GRCh38
rs61736626 Chr.15: 65286367-65286367 on Build GRCh38
rs977958648 Chr.15: 65286675-65286675 on Build GRCh38
rs1057155467 Chr.15: 65286507-65286507 on Build GRCh38
rs561284197 Chr.15: 65286531-65286531 on Build GRCh38
rs751771181 Chr.15: 65286283-65286283 on Build GRCh38
rs774991944 Chr.15: 65286412-65286412 on Build GRCh38
rs763591553 Chr.15: 65286344-65286344 on Build GRCh38
rs771967309 Chr.15: 65286411-65286411 on Build GRCh38
rs755166310 Chr.15: 65286352-65286352 on Build GRCh38
rs896076678 Chr.15: 65286336-65286336 on Build GRCh38
rs957371095 Chr.15: 65286526-65286526 on Build GRCh38
rs1010354591 Chr.15: 65286742-65286742 on Build GRCh38
rs751375140 Chr.15: 65286476-65286476 on Build GRCh38
rs767986748 Chr.15: 65286469-65286469 on Build GRCh38
rs776307743 Chr.15: 65286450-65286450 on Build GRCh38
rs749193373 Chr.15: 65286389-65286389 on Build GRCh38
rs915076878 Chr.15: 65286627-65286627 on Build GRCh38
rs964584289 Chr.15: 65286579-65286579 on Build GRCh38
rs756368685 Chr.15: 65286471-65286471 on Build GRCh38
rs911772940 Chr.15: 65286447-65286447 on Build GRCh38
rs112165616 Chr.15: 65286408-65286408 on Build GRCh38
rs893217362 Chr.15: 65286735-65286735 on Build GRCh38
rs763752978 Chr.15: 65286338-65286338 on Build GRCh38
rs759816873 Chr.15: 65286468-65286468 on Build GRCh38
rs368575488 Chr.15: 65286337-65286337 on Build GRCh38
rs540266112 Chr.15: 65286551-65286551 on Build GRCh38
rs575411904 Chr.15: 65286513-65286513 on Build GRCh38
rs758343932 Chr.15: 65286317-65286317 on Build GRCh38
rs774056642 Chr.15: 65286314-65286314 on Build GRCh38
rs1043573760 Chr.15: 65286490-65286490 on Build GRCh38
rs934613984 Chr.15: 65286505-65286505 on Build GRCh38
rs780328703 Chr.15: 65286282-65286282 on Build GRCh38
rs879170634 Chr.15: 65286486-65286486 on Build GRCh38
rs905692092 Chr.15: 65286664-65286664 on Build GRCh38
rs955805185 Chr.15: 65286537-65286537 on Build GRCh38
rs12906596 Chr.15: 65286730-65286730 on Build GRCh38
rs538009655 Chr.15: 65286407-65286407 on Build GRCh38
rs569016206 Chr.15: 65286745-65286745 on Build GRCh38
rs913492902 Chr.15: 65286423-65286423 on Build GRCh38
rs920358969 Chr.15: 65286594-65286594 on Build GRCh38
rs930289034 Chr.15: 65286614-65286614 on Build GRCh38
rs989064330 Chr.15: 65286403-65286403 on Build GRCh38
rs1035978342 Chr.15: 65286336-65286336 on Build GRCh38
rs768247444 Chr.15: 65286425-65286425 on Build GRCh38
rs907568763 Chr.15: 65286629-65286629 on Build GRCh38
rs141558905 Chr.15: 65286297-65286297 on Build GRCh38
rs1014010742 Chr.15: 65286498-65286498 on Build GRCh38
rs774997546 Chr.15: 65286326-65286326 on Build GRCh38
rs1033540207 Chr.15: 65286520-65286520 on Build GRCh38
rs1054505966 Chr.15: 65286729-65286729 on Build GRCh38
rs769656995 Chr.15: 65286305-65286305 on Build GRCh38
rs774713506 Chr.15: 65286464-65286464 on Build GRCh38
rs1057254 Chr.15: 65286695-65286695 on Build GRCh38
rs766806266 Chr.15: 65286457-65286457 on Build GRCh38
rs911426520 Chr.15: 65286430-65286430 on Build GRCh38
rs746707827 Chr.15: 65286418-65286418 on Build GRCh38

View Full Product Details