Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 687483-687977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR24
Entrez Gene ID
84219
Gene Name
WD repeat domain 24
Gene Aliases
C16orf21, JFP7
UniGene
Hs.459632
Gene Chromosome Location
Chr.16: 684622-690661 on Build GRCh38
Transcripts
NM_032259.3 XM_011522699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667905 c.1008C>G p.H336Q Chr.16: 687603-687603 on Build GRCh38
3512113 c.542C>T p.S181F Chr.16: 687679-687679 on Build GRCh38
6821633 c.910C>A p.R304R Chr.16: 687701-687701 on Build GRCh38
6821638 c.627C>T p.P209P Chr.16: 687594-687594 on Build GRCh38
3512115 c.522G>A p.R174R Chr.16: 687699-687699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753599279 Chr.16: 687624-687624 on Build GRCh38
rs966199618 Chr.16: 687927-687927 on Build GRCh38
rs574435776 Chr.16: 687861-687861 on Build GRCh38
rs769004225 Chr.16: 687684-687684 on Build GRCh38
rs780636695 Chr.16: 687666-687666 on Build GRCh38
rs1009393703 Chr.16: 687844-687844 on Build GRCh38
rs373732528 Chr.16: 687616-687616 on Build GRCh38
rs146642085 Chr.16: 687708-687708 on Build GRCh38
rs774603570 Chr.16: 687663-687674 on Build GRCh38
rs563147637 Chr.16: 687516-687516 on Build GRCh38
rs541763762 Chr.16: 687503-687503 on Build GRCh38
rs1019385371 Chr.16: 687908-687908 on Build GRCh38
rs778973720 Chr.16: 687903-687903 on Build GRCh38
rs1018804 Chr.16: 687793-687793 on Build GRCh38
rs751577568 Chr.16: 687541-687541 on Build GRCh38
rs775327153 Chr.16: 687847-687847 on Build GRCh38
rs374228480 Chr.16: 687756-687756 on Build GRCh38
rs201231442 Chr.16: 687726-687726 on Build GRCh38
rs143792602 Chr.16: 687594-687594 on Build GRCh38
rs766121733 Chr.16: 687520-687520 on Build GRCh38
rs762011921 Chr.16: 687849-687849 on Build GRCh38
rs770361774 Chr.16: 687796-687796 on Build GRCh38
rs756661315 Chr.16: 687665-687665 on Build GRCh38
rs548521612 Chr.16: 687717-687717 on Build GRCh38
rs755486817 Chr.16: 687867-687867 on Build GRCh38
rs767340433 Chr.16: 687515-687515 on Build GRCh38
rs575160798 Chr.16: 687942-687942 on Build GRCh38
rs775968417 Chr.16: 687897-687898 on Build GRCh38
rs761239460 Chr.16: 687593-687593 on Build GRCh38
rs1028383286 Chr.16: 687881-687881 on Build GRCh38
rs762076686 Chr.16: 687733-687733 on Build GRCh38
rs919426079 Chr.16: 687939-687939 on Build GRCh38
rs541577098 Chr.16: 687926-687926 on Build GRCh38
rs946293658 Chr.16: 687554-687554 on Build GRCh38
rs758631903 Chr.16: 687882-687882 on Build GRCh38
rs765560157 Chr.16: 687868-687868 on Build GRCh38
rs760465482 Chr.16: 687518-687518 on Build GRCh38
rs758595550 Chr.16: 687527-687527 on Build GRCh38
rs763218597 Chr.16: 687627-687627 on Build GRCh38
rs772604936 Chr.16: 687776-687777 on Build GRCh38
rs980338976 Chr.16: 687509-687509 on Build GRCh38
rs113262479 Chr.16: 687811-687811 on Build GRCh38
rs778432225 Chr.16: 687690-687690 on Build GRCh38
rs889564244 Chr.16: 687633-687633 on Build GRCh38
rs151060301 Chr.16: 687768-687768 on Build GRCh38
rs373953202 Chr.16: 687943-687943 on Build GRCh38
rs905209017 Chr.16: 687798-687798 on Build GRCh38
rs754326685 Chr.16: 687757-687757 on Build GRCh38
rs756738972 Chr.16: 687755-687755 on Build GRCh38
rs566209193 Chr.16: 687701-687701 on Build GRCh38
rs773965462 Chr.16: 687591-687591 on Build GRCh38
rs938064583 Chr.16: 687800-687800 on Build GRCh38
rs766210278 Chr.16: 687621-687621 on Build GRCh38
rs753100984 Chr.16: 687877-687877 on Build GRCh38
rs374125425 Chr.16: 687600-687600 on Build GRCh38
rs764236841 Chr.16: 687632-687632 on Build GRCh38
rs754980768 Chr.16: 687841-687841 on Build GRCh38
rs372034079 Chr.16: 687744-687744 on Build GRCh38
rs371109368 Chr.16: 687556-687556 on Build GRCh38
rs760838601 Chr.16: 687858-687858 on Build GRCh38
rs919541912 Chr.16: 687792-687792 on Build GRCh38
rs774461953 Chr.16: 687732-687732 on Build GRCh38
rs745770518 Chr.16: 687825-687825 on Build GRCh38
rs780548755 Chr.16: 687567-687567 on Build GRCh38
rs747802777 Chr.16: 687772-687772 on Build GRCh38
rs755326725 Chr.16: 687763-687763 on Build GRCh38
rs556891132 Chr.16: 687937-687937 on Build GRCh38
rs887925830 Chr.16: 687892-687892 on Build GRCh38
rs553207972 Chr.16: 687830-687830 on Build GRCh38
rs1024263678 Chr.16: 687752-687752 on Build GRCh38
rs777331548 Chr.16: 687777-687777 on Build GRCh38
rs952248168 Chr.16: 687957-687957 on Build GRCh38
rs374792149 Chr.16: 687550-687550 on Build GRCh38
rs771502736 Chr.16: 687698-687698 on Build GRCh38
rs746206445 Chr.16: 687745-687745 on Build GRCh38
rs767750219 Chr.16: 687743-687743 on Build GRCh38
rs746988584 Chr.16: 687705-687705 on Build GRCh38
rs547474455 Chr.16: 687642-687642 on Build GRCh38
rs941119995 Chr.16: 687910-687910 on Build GRCh38
rs754321773 Chr.16: 687866-687866 on Build GRCh38
rs746625345 Chr.16: 687914-687914 on Build GRCh38
rs770307856 Chr.16: 687561-687561 on Build GRCh38
rs768185333 Chr.16: 687582-687582 on Build GRCh38
rs903746996 Chr.16: 687865-687865 on Build GRCh38
rs111553767 Chr.16: 687553-687553 on Build GRCh38
rs781286465 Chr.16: 687658-687658 on Build GRCh38
rs777976939 Chr.16: 687540-687540 on Build GRCh38
rs752938193 Chr.16: 687521-687521 on Build GRCh38
rs750721897 Chr.16: 687635-687635 on Build GRCh38
rs750530483 Chr.16: 687746-687746 on Build GRCh38
rs777306449 Chr.16: 687901-687901 on Build GRCh38
rs953888430 Chr.16: 687672-687672 on Build GRCh38
rs570644015 Chr.16: 687806-687806 on Build GRCh38
rs749653258 Chr.16: 687681-687681 on Build GRCh38
rs376879524 Chr.16: 687695-687695 on Build GRCh38
rs751019760 Chr.16: 687660-687660 on Build GRCh38
rs761874293 Chr.16: 687512-687512 on Build GRCh38
rs777407917 Chr.16: 687897-687897 on Build GRCh38
rs1008382899 Chr.16: 687907-687907 on Build GRCh38
rs765044529 Chr.16: 687728-687728 on Build GRCh38
rs913243960 Chr.16: 687803-687803 on Build GRCh38
rs758037400 Chr.16: 687773-687773 on Build GRCh38
rs149004720 Chr.16: 687570-687570 on Build GRCh38
rs773506352 Chr.16: 687854-687854 on Build GRCh38

View Full Product Details