Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2044524-2044959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTHL1
Entrez Gene ID
4913
Gene Name
nth-like DNA glycosylase 1
Gene Aliases
FAP3, NTH1, OCTS3, hNTH1
UniGene
Hs.66196
Gene Chromosome Location
Chr.16: 2039815-2047872 on Build GRCh38
Transcripts
NM_001318193.1 NM_001318194.1 NM_002528.6 XM_017023253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298189 c.416C>G p.S139C Chr.16: 2044763-2044763 on Build GRCh38
6297022 c.479C>A p.T160K Chr.16: 2044700-2044700 on Build GRCh38
6201529 c.488G>A p.S163N Chr.16: 2044691-2044691 on Build GRCh38
5513141 c.528C>A p.I176I Chr.16: 2044651-2044651 on Build GRCh38
5462512 c.424A>C p.T142P Chr.16: 2044755-2044755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372848463 Chr.16: 2044906-2044906 on Build GRCh38
rs757412628 Chr.16: 2044775-2044775 on Build GRCh38
rs573165403 Chr.16: 2044560-2044560 on Build GRCh38
rs746940807 Chr.16: 2044721-2044721 on Build GRCh38
rs968060928 Chr.16: 2044679-2044679 on Build GRCh38
rs755309344 Chr.16: 2044830-2044830 on Build GRCh38
rs370384669 Chr.16: 2044602-2044602 on Build GRCh38
rs895589930 Chr.16: 2044598-2044598 on Build GRCh38
rs750798417 Chr.16: 2044699-2044699 on Build GRCh38
rs775138715 Chr.16: 2044811-2044811 on Build GRCh38
rs377517542 Chr.16: 2044607-2044607 on Build GRCh38
rs774894275 Chr.16: 2044734-2044736 on Build GRCh38
rs766774887 Chr.16: 2044743-2044743 on Build GRCh38
rs902449328 Chr.16: 2044640-2044640 on Build GRCh38
rs764702496 Chr.16: 2044674-2044674 on Build GRCh38
rs769348183 Chr.16: 2044807-2044807 on Build GRCh38
rs777988627 Chr.16: 2044793-2044793 on Build GRCh38
rs1047427072 Chr.16: 2044930-2044930 on Build GRCh38
rs758667255 Chr.16: 2044746-2044746 on Build GRCh38
rs962452543 Chr.16: 2044705-2044705 on Build GRCh38
rs768014084 Chr.16: 2044735-2044735 on Build GRCh38
rs369651668 Chr.16: 2044834-2044834 on Build GRCh38
rs112724577 Chr.16: 2044672-2044672 on Build GRCh38
rs200359356 Chr.16: 2044608-2044608 on Build GRCh38
rs760491144 Chr.16: 2044666-2044666 on Build GRCh38
rs754383091 Chr.16: 2044606-2044606 on Build GRCh38
rs930291472 Chr.16: 2044928-2044928 on Build GRCh38
rs201389795 Chr.16: 2044826-2044826 on Build GRCh38
rs927975974 Chr.16: 2044564-2044564 on Build GRCh38
rs990230297 Chr.16: 2044845-2044845 on Build GRCh38
rs750992242 Chr.16: 2044736-2044736 on Build GRCh38
rs772100661 Chr.16: 2044656-2044656 on Build GRCh38
rs545441027 Chr.16: 2044936-2044937 on Build GRCh38
rs755185485 Chr.16: 2044771-2044771 on Build GRCh38
rs746192138 Chr.16: 2044630-2044630 on Build GRCh38
rs566860680 Chr.16: 2044795-2044795 on Build GRCh38
rs891039223 Chr.16: 2044857-2044857 on Build GRCh38
rs1805378 Chr.16: 2044652-2044652 on Build GRCh38
rs764198509 Chr.16: 2044576-2044576 on Build GRCh38
rs1012994792 Chr.16: 2044600-2044600 on Build GRCh38
rs956016009 Chr.16: 2044872-2044872 on Build GRCh38
rs756597137 Chr.16: 2044739-2044739 on Build GRCh38
rs374489979 Chr.16: 2044722-2044722 on Build GRCh38
rs773824795 Chr.16: 2044822-2044822 on Build GRCh38
rs1044127089 Chr.16: 2044738-2044738 on Build GRCh38
rs113462940 Chr.16: 2044756-2044756 on Build GRCh38
rs145366761 Chr.16: 2044711-2044711 on Build GRCh38
rs35691379 Chr.16: 2044608-2044609 on Build GRCh38
rs950917950 Chr.16: 2044616-2044616 on Build GRCh38
rs371328106 Chr.16: 2044789-2044789 on Build GRCh38
rs370228590 Chr.16: 2044797-2044797 on Build GRCh38
rs752220394 Chr.16: 2044687-2044687 on Build GRCh38
rs759156239 Chr.16: 2044727-2044727 on Build GRCh38
rs542117101 Chr.16: 2044909-2044909 on Build GRCh38
rs748183280 Chr.16: 2044783-2044783 on Build GRCh38
rs993161596 Chr.16: 2044878-2044878 on Build GRCh38
rs190313689 Chr.16: 2044558-2044558 on Build GRCh38
rs776464671 Chr.16: 2044725-2044725 on Build GRCh38
rs757736619 Chr.16: 2044695-2044695 on Build GRCh38
rs1032004313 Chr.16: 2044860-2044860 on Build GRCh38
rs116596661 Chr.16: 2044908-2044908 on Build GRCh38
rs920372543 Chr.16: 2044926-2044926 on Build GRCh38
rs990799280 Chr.16: 2044818-2044818 on Build GRCh38
rs1035275685 Chr.16: 2044925-2044925 on Build GRCh38
rs773170479 Chr.16: 2044658-2044658 on Build GRCh38
rs561923127 Chr.16: 2044644-2044644 on Build GRCh38
rs764907191 Chr.16: 2044731-2044731 on Build GRCh38
rs939321761 Chr.16: 2044569-2044569 on Build GRCh38
rs749527022 Chr.16: 2044707-2044707 on Build GRCh38
rs781578064 Chr.16: 2044609-2044609 on Build GRCh38
rs1039338012 Chr.16: 2044871-2044871 on Build GRCh38
rs917049468 Chr.16: 2044886-2044886 on Build GRCh38
rs766136810 Chr.16: 2044667-2044667 on Build GRCh38
rs555337484 Chr.16: 2044935-2044935 on Build GRCh38
rs201521100 Chr.16: 2044717-2044717 on Build GRCh38
rs760441908 Chr.16: 2044604-2044604 on Build GRCh38
rs766868081 Chr.16: 2044824-2044824 on Build GRCh38
rs3087466 Chr.16: 2044837-2044837 on Build GRCh38
rs774887697 Chr.16: 2044732-2044732 on Build GRCh38
rs536140679 Chr.16: 2044800-2044800 on Build GRCh38
rs781629683 Chr.16: 2044698-2044698 on Build GRCh38
rs1057270844 Chr.16: 2044574-2044574 on Build GRCh38
rs201980301 Chr.16: 2044645-2044645 on Build GRCh38
rs111484173 Chr.16: 2044888-2044888 on Build GRCh38
rs2073637 Chr.16: 2044812-2044812 on Build GRCh38
rs376048896 Chr.16: 2044710-2044710 on Build GRCh38
rs374077464 Chr.16: 2044835-2044835 on Build GRCh38
rs776325627 Chr.16: 2044798-2044798 on Build GRCh38
rs761211463 Chr.16: 2044823-2044823 on Build GRCh38
rs551068959 Chr.16: 2044647-2044647 on Build GRCh38
rs747049417 Chr.16: 2044841-2044841 on Build GRCh38
rs1025386529 Chr.16: 2044897-2044897 on Build GRCh38
rs184425926 Chr.16: 2044849-2044849 on Build GRCh38
rs11543268 Chr.16: 2044762-2044762 on Build GRCh38
rs767269308 Chr.16: 2044601-2044601 on Build GRCh38
rs759955745 Chr.16: 2044795-2044796 on Build GRCh38
rs745657333 Chr.16: 2044852-2044852 on Build GRCh38
rs886114582 Chr.16: 2044937-2044937 on Build GRCh38
rs374683930 Chr.16: 2044605-2044605 on Build GRCh38
rs140211154 Chr.16: 2044642-2044642 on Build GRCh38
rs748576083 Chr.16: 2044712-2044712 on Build GRCh38
rs756403102 Chr.16: 2044700-2044700 on Build GRCh38
rs539481355 Chr.16: 2044858-2044858 on Build GRCh38
rs765517336 Chr.16: 2044832-2044832 on Build GRCh38
rs377505085 Chr.16: 2044592-2044592 on Build GRCh38
rs372232458 Chr.16: 2044820-2044820 on Build GRCh38
rs983470671 Chr.16: 2044559-2044559 on Build GRCh38
rs201515357 Chr.16: 2044669-2044669 on Build GRCh38
rs377360166 Chr.16: 2044760-2044760 on Build GRCh38
rs150437839 Chr.16: 2044709-2044709 on Build GRCh38
rs768198453 Chr.16: 2044824-2044825 on Build GRCh38
rs1048431740 Chr.16: 2044792-2044792 on Build GRCh38
rs762393763 Chr.16: 2044734-2044734 on Build GRCh38

View Full Product Details