Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31915501-31916015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF267
Entrez Gene ID
10308
Gene Name
zinc finger protein 267
Gene Aliases
HZF2
UniGene
Hs.460645
Gene Chromosome Location
Chr.16: 31873758-31917951 on Build GRCh38
Transcripts
NM_001265588.1 NM_003414.5 NR_049749.1 XM_017022865.1 XM_017022866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5076790 c.1525C>A p.Q509K Chr.16: 31915774-31915774 on Build GRCh38
77103 c.1679G>C p.S560T Chr.16: 31915928-31915928 on Build GRCh38
5054872 c.1508G>A p.R503H Chr.16: 31915757-31915757 on Build GRCh38
1377682 c.1397G>A p.C466Y Chr.16: 31915646-31915646 on Build GRCh38
3509319 c.1415C>T p.S472F Chr.16: 31915664-31915664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541312659 Chr.16: 31915745-31915745 on Build GRCh38
rs903750705 Chr.16: 31915925-31915925 on Build GRCh38
rs759085650 Chr.16: 31915668-31915668 on Build GRCh38
rs142848475 Chr.16: 31915644-31915644 on Build GRCh38
rs901916662 Chr.16: 31915972-31915972 on Build GRCh38
rs778811812 Chr.16: 31915945-31915945 on Build GRCh38
rs746131922 Chr.16: 31915564-31915564 on Build GRCh38
rs748124802 Chr.16: 31915774-31915774 on Build GRCh38
rs779756174 Chr.16: 31915564-31915564 on Build GRCh38
rs751674405 Chr.16: 31915756-31915756 on Build GRCh38
rs755025685 Chr.16: 31915764-31915764 on Build GRCh38
rs754297644 Chr.16: 31915532-31915532 on Build GRCh38
rs749033519 Chr.16: 31915786-31915786 on Build GRCh38
rs754420435 Chr.16: 31915682-31915682 on Build GRCh38
rs769934396 Chr.16: 31915758-31915758 on Build GRCh38
rs755129791 Chr.16: 31915892-31915892 on Build GRCh38
rs909059556 Chr.16: 31915952-31915952 on Build GRCh38
rs775124056 Chr.16: 31915961-31915961 on Build GRCh38
rs771790142 Chr.16: 31915957-31915957 on Build GRCh38
rs768643998 Chr.16: 31915596-31915596 on Build GRCh38
rs757142983 Chr.16: 31915938-31915938 on Build GRCh38
rs750695362 Chr.16: 31915730-31915730 on Build GRCh38
rs148534327 Chr.16: 31915585-31915585 on Build GRCh38
rs749158015 Chr.16: 31915926-31915926 on Build GRCh38
rs774072165 Chr.16: 31915657-31915657 on Build GRCh38
rs763192014 Chr.16: 31915856-31915856 on Build GRCh38
rs777997681 Chr.16: 31915922-31915932 on Build GRCh38
rs781073898 Chr.16: 31915604-31915604 on Build GRCh38
rs556332192 Chr.16: 31915784-31915784 on Build GRCh38
rs868493070 Chr.16: 31915949-31915949 on Build GRCh38
rs372450625 Chr.16: 31915821-31915821 on Build GRCh38
rs747939925 Chr.16: 31915632-31915632 on Build GRCh38
rs765648644 Chr.16: 31915842-31915842 on Build GRCh38
rs190018640 Chr.16: 31915638-31915638 on Build GRCh38
rs762185591 Chr.16: 31915697-31915697 on Build GRCh38
rs369182518 Chr.16: 31915823-31915823 on Build GRCh38
rs770747607 Chr.16: 31915793-31915793 on Build GRCh38
rs374777310 Chr.16: 31915794-31915794 on Build GRCh38
rs759217317 Chr.16: 31915822-31915822 on Build GRCh38
rs183588326 Chr.16: 31915557-31915557 on Build GRCh38
rs774745782 Chr.16: 31915780-31915780 on Build GRCh38
rs766634128 Chr.16: 31915888-31915888 on Build GRCh38
rs772346336 Chr.16: 31915900-31915900 on Build GRCh38
rs559723367 Chr.16: 31915773-31915773 on Build GRCh38
rs751799225 Chr.16: 31915889-31915889 on Build GRCh38
rs769664773 Chr.16: 31915633-31915633 on Build GRCh38
rs781266280 Chr.16: 31915895-31915895 on Build GRCh38
rs868002676 Chr.16: 31915844-31915844 on Build GRCh38
rs369487547 Chr.16: 31915531-31915531 on Build GRCh38
rs746782142 Chr.16: 31915685-31915687 on Build GRCh38
rs762395235 Chr.16: 31915837-31915837 on Build GRCh38
rs201255582 Chr.16: 31915985-31915985 on Build GRCh38
rs745519301 Chr.16: 31915951-31915951 on Build GRCh38
rs200494347 Chr.16: 31915653-31915653 on Build GRCh38
rs756146826 Chr.16: 31915910-31915910 on Build GRCh38
rs371928927 Chr.16: 31915681-31915681 on Build GRCh38
rs118056264 Chr.16: 31915987-31915987 on Build GRCh38
rs750862618 Chr.16: 31915855-31915855 on Build GRCh38
rs376119387 Chr.16: 31915914-31915914 on Build GRCh38
rs867997720 Chr.16: 31915586-31915586 on Build GRCh38
rs868237473 Chr.16: 31915805-31915805 on Build GRCh38
rs533144236 Chr.16: 31915831-31915831 on Build GRCh38
rs780246168 Chr.16: 31915752-31915752 on Build GRCh38
rs576166037 Chr.16: 31915787-31915787 on Build GRCh38
rs199631232 Chr.16: 31915942-31915942 on Build GRCh38
rs757620592 Chr.16: 31915551-31915551 on Build GRCh38
rs746496766 Chr.16: 31915969-31915969 on Build GRCh38
rs575364062 Chr.16: 31915588-31915588 on Build GRCh38
rs146250449 Chr.16: 31915672-31915672 on Build GRCh38
rs962943372 Chr.16: 31915725-31915725 on Build GRCh38
rs911299636 Chr.16: 31915887-31915887 on Build GRCh38
rs777542126 Chr.16: 31915922-31915922 on Build GRCh38
rs1017105248 Chr.16: 31915670-31915670 on Build GRCh38
rs536399270 Chr.16: 31915781-31915781 on Build GRCh38
rs542618323 Chr.16: 31915589-31915589 on Build GRCh38
rs774793766 Chr.16: 31915834-31915834 on Build GRCh38
rs373938717 Chr.16: 31915867-31915867 on Build GRCh38
rs773484903 Chr.16: 31915953-31915953 on Build GRCh38
rs765637763 Chr.16: 31915706-31915706 on Build GRCh38

View Full Product Details