Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71648447-71648992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
UniGene
Hs.709458
Gene Chromosome Location
Chr.16: 71644924-71724701 on Build GRCh38
Transcripts
NM_001289003.1 NM_015020.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379626 c.3904C>T p.R1302W Chr.16: 71648958-71648958 on Build GRCh38
4566493 c.3946_3947CC>GT p.P1316V Chr.16: 71648915-71648916 on Build GRCh38
472061 c.3915T>C p.P1305P Chr.16: 71648947-71648947 on Build GRCh38
3795102 c.3966A>T p.A1322A Chr.16: 71648896-71648896 on Build GRCh38
273107 c.3964G>T p.A1322S Chr.16: 71648898-71648898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565549181 Chr.16: 71648493-71648493 on Build GRCh38
rs907171916 Chr.16: 71648562-71648562 on Build GRCh38
rs11301738 Chr.16: 71648605-71648605 on Build GRCh38
rs748258512 Chr.16: 71648952-71648952 on Build GRCh38
rs371565551 Chr.16: 71648883-71648883 on Build GRCh38
rs898284467 Chr.16: 71648837-71648837 on Build GRCh38
rs61733124 Chr.16: 71648927-71648927 on Build GRCh38
rs978826344 Chr.16: 71648494-71648494 on Build GRCh38
rs540816929 Chr.16: 71648822-71648822 on Build GRCh38
rs147202140 Chr.16: 71648953-71648953 on Build GRCh38
rs940830080 Chr.16: 71648599-71648599 on Build GRCh38
rs1030690152 Chr.16: 71648644-71648644 on Build GRCh38
rs751206937 Chr.16: 71648917-71648917 on Build GRCh38
rs765770499 Chr.16: 71648908-71648908 on Build GRCh38
rs372040747 Chr.16: 71648766-71648766 on Build GRCh38
rs1056304642 Chr.16: 71648587-71648587 on Build GRCh38
rs146034837 Chr.16: 71648956-71648956 on Build GRCh38
rs958371869 Chr.16: 71648794-71648794 on Build GRCh38
rs538701229 Chr.16: 71648765-71648765 on Build GRCh38
rs13333988 Chr.16: 71648687-71648687 on Build GRCh38
rs994289427 Chr.16: 71648660-71648660 on Build GRCh38
rs181074659 Chr.16: 71648577-71648577 on Build GRCh38
rs555083461 Chr.16: 71648797-71648797 on Build GRCh38
rs371375607 Chr.16: 71648899-71648899 on Build GRCh38
rs139626439 Chr.16: 71648923-71648923 on Build GRCh38
rs558609247 Chr.16: 71648750-71648750 on Build GRCh38
rs938673151 Chr.16: 71648784-71648784 on Build GRCh38
rs760864609 Chr.16: 71648958-71648958 on Build GRCh38
rs752265941 Chr.16: 71648909-71648909 on Build GRCh38
rs532056276 Chr.16: 71648511-71648511 on Build GRCh38
rs566812443 Chr.16: 71648793-71648793 on Build GRCh38
rs192113290 Chr.16: 71648957-71648957 on Build GRCh38
rs533974235 Chr.16: 71648745-71648745 on Build GRCh38
rs61733125 Chr.16: 71648893-71648893 on Build GRCh38
rs141296681 Chr.16: 71648915-71648915 on Build GRCh38
rs777229982 Chr.16: 71648847-71648847 on Build GRCh38
rs965178375 Chr.16: 71648479-71648479 on Build GRCh38
rs946153668 Chr.16: 71648751-71648751 on Build GRCh38
rs147433033 Chr.16: 71648922-71648922 on Build GRCh38
rs955016269 Chr.16: 71648686-71648686 on Build GRCh38
rs201990639 Chr.16: 71648887-71648887 on Build GRCh38
rs545916742 Chr.16: 71648761-71648761 on Build GRCh38
rs770587173 Chr.16: 71648853-71648853 on Build GRCh38
rs917011352 Chr.16: 71648598-71648598 on Build GRCh38
rs573383735 Chr.16: 71648804-71648804 on Build GRCh38
rs547852955 Chr.16: 71648679-71648679 on Build GRCh38
rs760184822 Chr.16: 71648905-71648905 on Build GRCh38
rs749241618 Chr.16: 71648649-71648649 on Build GRCh38
rs756924434 Chr.16: 71648919-71648919 on Build GRCh38
rs746839361 Chr.16: 71648849-71648849 on Build GRCh38
rs1012066384 Chr.16: 71648615-71648615 on Build GRCh38
rs759096889 Chr.16: 71648883-71648884 on Build GRCh38
rs201211632 Chr.16: 71648901-71648901 on Build GRCh38
rs958683829 Chr.16: 71648559-71648559 on Build GRCh38
rs968610236 Chr.16: 71648785-71648785 on Build GRCh38
rs201141453 Chr.16: 71648900-71648900 on Build GRCh38
rs977504865 Chr.16: 71648932-71648932 on Build GRCh38
rs961802089 Chr.16: 71648869-71648869 on Build GRCh38
rs557846154 Chr.16: 71648872-71648873 on Build GRCh38
rs1018519875 Chr.16: 71648699-71648699 on Build GRCh38
rs398029862 Chr.16: 71648616-71648616 on Build GRCh38
rs930922656 Chr.16: 71648639-71648639 on Build GRCh38
rs374906434 Chr.16: 71648783-71648784 on Build GRCh38
rs752747649 Chr.16: 71648899-71648902 on Build GRCh38
rs745867400 Chr.16: 71648880-71648880 on Build GRCh38
rs575679600 Chr.16: 71648787-71648787 on Build GRCh38
rs866860107 Chr.16: 71648528-71648528 on Build GRCh38
rs1000141575 Chr.16: 71648792-71648792 on Build GRCh38
rs758126134 Chr.16: 71648845-71648845 on Build GRCh38
rs892555449 Chr.16: 71648720-71648720 on Build GRCh38
rs144133911 Chr.16: 71648914-71648914 on Build GRCh38
rs146231024 Chr.16: 71648629-71648629 on Build GRCh38
rs988650721 Chr.16: 71648483-71648483 on Build GRCh38
rs200189816 Chr.16: 71648904-71648904 on Build GRCh38
rs534044390 Chr.16: 71648839-71648839 on Build GRCh38
rs550540021 Chr.16: 71648539-71648539 on Build GRCh38
rs773267489 Chr.16: 71648898-71648898 on Build GRCh38
rs756088786 Chr.16: 71648905-71648907 on Build GRCh38
rs964618493 Chr.16: 71648732-71648732 on Build GRCh38
rs1036526704 Chr.16: 71648612-71648612 on Build GRCh38
rs776468659 Chr.16: 71648872-71648872 on Build GRCh38
rs889098877 Chr.16: 71648652-71648652 on Build GRCh38
rs779920332 Chr.16: 71648928-71648928 on Build GRCh38
rs1020175914 Chr.16: 71648490-71648490 on Build GRCh38
rs771178310 Chr.16: 71648968-71648968 on Build GRCh38
rs1050837431 Chr.16: 71648643-71648643 on Build GRCh38
rs766925624 Chr.16: 71648885-71648886 on Build GRCh38

View Full Product Details