Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88037836-88038333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANP
Entrez Gene ID
54971
Gene Name
BTG3 associated nuclear protein
Gene Aliases
BEND1, SMAR1, SMARBP1
UniGene
Hs.461705
Gene Chromosome Location
Chr.16: 87949216-88077318 on Build GRCh38
Transcripts
NM_001173539.1 NM_001173540.1 NM_001173541.1 NM_001173542.1 NM_001173543.1 NM_017869.3 NM_079837.2 XM_005256008.3 XM_005256009.3 XM_011523170.2 XM_011523172.2 XM_011523174.2 XM_011523175.2 XM_011523176.2 XM_011523177.2 XM_011523178.2 XM_011523179.2 XM_011523180.2 XM_011523181.2 XM_011523182.1 XM_011523183.2 XM_011523184.2 XM_011523186.2 XM_017023378.1 XM_017023379.1 XM_017023380.1 XM_017023381.1 XM_017023382.1 XM_017023383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5458596 c.1207G>A p.G403S Chr.16: 88038006-88038006 on Build GRCh38
5458595 c.1273G>A p.G425S Chr.16: 88038006-88038006 on Build GRCh38
5458597 c.1189G>A p.G397S Chr.16: 88038006-88038006 on Build GRCh38
974670 c.1176C>T p.H392H Chr.16: 88037993-88037993 on Build GRCh38
1153016 c.1260C>T p.H420H Chr.16: 88037993-88037993 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs72631418 Chr.16: 88038266-88038266 on Build GRCh38
rs868453674 Chr.16: 88038309-88038309 on Build GRCh38
rs941360581 Chr.16: 88038070-88038070 on Build GRCh38
rs575753866 Chr.16: 88038179-88038179 on Build GRCh38
rs1037444885 Chr.16: 88038257-88038257 on Build GRCh38
rs1043836889 Chr.16: 88038287-88038287 on Build GRCh38
rs200430799 Chr.16: 88037961-88037961 on Build GRCh38
rs978634500 Chr.16: 88037900-88037900 on Build GRCh38
rs370698328 Chr.16: 88038055-88038055 on Build GRCh38
rs759554913 Chr.16: 88038027-88038027 on Build GRCh38
rs767090616 Chr.16: 88038040-88038040 on Build GRCh38
rs117133887 Chr.16: 88038295-88038295 on Build GRCh38
rs528439672 Chr.16: 88038193-88038193 on Build GRCh38
rs138252049 Chr.16: 88037994-88037994 on Build GRCh38
rs769450599 Chr.16: 88038028-88038028 on Build GRCh38
rs754272801 Chr.16: 88038043-88038043 on Build GRCh38
rs369335103 Chr.16: 88038030-88038030 on Build GRCh38
rs771103166 Chr.16: 88038058-88038058 on Build GRCh38
rs113007783 Chr.16: 88038247-88038247 on Build GRCh38
rs368481691 Chr.16: 88037962-88037962 on Build GRCh38
rs998410532 Chr.16: 88038112-88038112 on Build GRCh38
rs754395198 Chr.16: 88037981-88037981 on Build GRCh38
rs139447252 Chr.16: 88037883-88037883 on Build GRCh38
rs149196674 Chr.16: 88038005-88038005 on Build GRCh38
rs917098195 Chr.16: 88038239-88038239 on Build GRCh38
rs564972663 Chr.16: 88038221-88038221 on Build GRCh38
rs924490857 Chr.16: 88037903-88037903 on Build GRCh38
rs185920071 Chr.16: 88037860-88037860 on Build GRCh38
rs890011142 Chr.16: 88038282-88038282 on Build GRCh38
rs1007023561 Chr.16: 88038283-88038283 on Build GRCh38
rs377374842 Chr.16: 88038048-88038048 on Build GRCh38
rs934271905 Chr.16: 88038260-88038260 on Build GRCh38
rs979908881 Chr.16: 88038246-88038246 on Build GRCh38
rs546566316 Chr.16: 88038208-88038208 on Build GRCh38
rs776387973 Chr.16: 88038014-88038014 on Build GRCh38
rs992750577 Chr.16: 88038238-88038238 on Build GRCh38
rs561437669 Chr.16: 88038189-88038189 on Build GRCh38
rs763578662 Chr.16: 88037951-88037951 on Build GRCh38
rs763670799 Chr.16: 88038038-88038038 on Build GRCh38
rs1007285397 Chr.16: 88038137-88038137 on Build GRCh38
rs752256629 Chr.16: 88037948-88037948 on Build GRCh38
rs918594597 Chr.16: 88038258-88038258 on Build GRCh38
rs948792908 Chr.16: 88037915-88037916 on Build GRCh38
rs909908201 Chr.16: 88038068-88038068 on Build GRCh38
rs532457837 Chr.16: 88038235-88038235 on Build GRCh38
rs368943863 Chr.16: 88037954-88037954 on Build GRCh38
rs1014611432 Chr.16: 88038296-88038296 on Build GRCh38
rs372806525 Chr.16: 88038044-88038044 on Build GRCh38
rs753035881 Chr.16: 88038049-88038049 on Build GRCh38
rs1031295357 Chr.16: 88038223-88038223 on Build GRCh38
rs755258976 Chr.16: 88037993-88037993 on Build GRCh38
rs999823743 Chr.16: 88038209-88038209 on Build GRCh38
rs1051742335 Chr.16: 88038113-88038113 on Build GRCh38
rs563904210 Chr.16: 88038118-88038118 on Build GRCh38
rs758843447 Chr.16: 88038052-88038052 on Build GRCh38
rs770923492 Chr.16: 88037926-88037926 on Build GRCh38
rs760683336 Chr.16: 88037922-88037922 on Build GRCh38
rs762591107 Chr.16: 88038036-88038036 on Build GRCh38
rs969862068 Chr.16: 88038243-88038243 on Build GRCh38
rs375732500 Chr.16: 88037940-88037940 on Build GRCh38
rs149669486 Chr.16: 88038106-88038106 on Build GRCh38
rs889954895 Chr.16: 88038136-88038136 on Build GRCh38
rs941394754 Chr.16: 88038250-88038250 on Build GRCh38
rs999945329 Chr.16: 88038294-88038294 on Build GRCh38
rs373953863 Chr.16: 88038057-88038057 on Build GRCh38
rs372847394 Chr.16: 88037911-88037911 on Build GRCh38
rs778144977 Chr.16: 88038003-88038003 on Build GRCh38
rs751226491 Chr.16: 88038039-88038039 on Build GRCh38
rs1024245582 Chr.16: 88038300-88038300 on Build GRCh38
rs188770212 Chr.16: 88037910-88037910 on Build GRCh38
rs1037007077 Chr.16: 88038071-88038072 on Build GRCh38
rs772483207 Chr.16: 88037996-88037996 on Build GRCh38
rs536869710 Chr.16: 88038288-88038288 on Build GRCh38
rs776643413 Chr.16: 88037939-88037939 on Build GRCh38
rs895896768 Chr.16: 88037914-88037914 on Build GRCh38
rs968408802 Chr.16: 88038176-88038176 on Build GRCh38
rs1017324486 Chr.16: 88038169-88038169 on Build GRCh38
rs765107975 Chr.16: 88037945-88037945 on Build GRCh38
rs1015836594 Chr.16: 88037956-88037956 on Build GRCh38
rs181160027 Chr.16: 88038246-88038246 on Build GRCh38
rs771156378 Chr.16: 88038010-88038010 on Build GRCh38
rs778088228 Chr.16: 88038054-88038054 on Build GRCh38
rs559457962 Chr.16: 88038293-88038293 on Build GRCh38
rs969934531 Chr.16: 88038305-88038305 on Build GRCh38
rs1005836265 Chr.16: 88037943-88037943 on Build GRCh38
rs112204663 Chr.16: 88038180-88038180 on Build GRCh38
rs762712156 Chr.16: 88037949-88037949 on Build GRCh38

View Full Product Details