Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88716120-88716627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4722
Entrez Gene ID
100616167
Gene Name
microRNA 4722
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.16: 88716278-88716337 on Build GRCh38
Transcripts
NR_039873.1 NM_001142864.3

Gene Symbol
PIEZO1
Entrez Gene ID
9780
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
DHS, FAM38A, LMPH3, Mib
UniGene
Hs.377001
Gene Chromosome Location
Chr.16: 88715338-88785220 on Build GRCh38
Transcripts
NR_039873.1 NM_001142864.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206319 c.6929A>G p.D2310G Chr.16: 88716481-88716481 on Build GRCh38
4129489 c.7059T>C p.P2353P Chr.16: 88716268-88716268 on Build GRCh38
4129490 c.663T>C p.P221P Chr.16: 88716268-88716268 on Build GRCh38
4623450 c.685C>T p.R229C Chr.16: 88716246-88716246 on Build GRCh38
3818696 c.6861G>C p.Q2287H Chr.16: 88716624-88716624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575843344 Chr.16: 88716345-88716345 on Build GRCh38
rs895132366 Chr.16: 88716175-88716175 on Build GRCh38
rs767518948 Chr.16: 88716346-88716346 on Build GRCh38
rs758880704 Chr.16: 88716298-88716298 on Build GRCh38
rs534495716 Chr.16: 88716231-88716231 on Build GRCh38
rs147479380 Chr.16: 88716582-88716582 on Build GRCh38
rs778431128 Chr.16: 88716555-88716555 on Build GRCh38
rs143153481 Chr.16: 88716503-88716503 on Build GRCh38
rs995439313 Chr.16: 88716344-88716344 on Build GRCh38
rs1011783029 Chr.16: 88716536-88716536 on Build GRCh38
rs922211056 Chr.16: 88716229-88716229 on Build GRCh38
rs1027668127 Chr.16: 88716187-88716187 on Build GRCh38
rs1052413244 Chr.16: 88716513-88716513 on Build GRCh38
rs371049428 Chr.16: 88716249-88716249 on Build GRCh38
rs570953388 Chr.16: 88716579-88716579 on Build GRCh38
rs545326364 Chr.16: 88716420-88716420 on Build GRCh38
rs760709820 Chr.16: 88716475-88716475 on Build GRCh38
rs746444593 Chr.16: 88716417-88716417 on Build GRCh38
rs553165820 Chr.16: 88716372-88716372 on Build GRCh38
rs953643803 Chr.16: 88716235-88716235 on Build GRCh38
rs775703527 Chr.16: 88716311-88716311 on Build GRCh38
rs528203278 Chr.16: 88716483-88716483 on Build GRCh38
rs889317135 Chr.16: 88716531-88716531 on Build GRCh38
rs774561483 Chr.16: 88716446-88716446 on Build GRCh38
rs766084250 Chr.16: 88716220-88716220 on Build GRCh38
rs768952742 Chr.16: 88716445-88716445 on Build GRCh38
rs993658851 Chr.16: 88716586-88716586 on Build GRCh38
rs765355284 Chr.16: 88716258-88716258 on Build GRCh38
rs536821291 Chr.16: 88716597-88716597 on Build GRCh38
rs566985926 Chr.16: 88716474-88716474 on Build GRCh38
rs930119085 Chr.16: 88716283-88716283 on Build GRCh38
rs1006990165 Chr.16: 88716151-88716151 on Build GRCh38
rs568552611 Chr.16: 88716602-88716602 on Build GRCh38
rs774516383 Chr.16: 88716335-88716335 on Build GRCh38
rs892485332 Chr.16: 88716543-88716543 on Build GRCh38
rs374203151 Chr.16: 88716404-88716404 on Build GRCh38
rs1013412353 Chr.16: 88716236-88716236 on Build GRCh38
rs766262024 Chr.16: 88716477-88716477 on Build GRCh38
rs761248105 Chr.16: 88716276-88716276 on Build GRCh38
rs749571599 Chr.16: 88716312-88716312 on Build GRCh38
rs936661021 Chr.16: 88716510-88716510 on Build GRCh38
rs768834483 Chr.16: 88716327-88716327 on Build GRCh38
rs781582005 Chr.16: 88716405-88716405 on Build GRCh38
rs2290902 Chr.16: 88716268-88716268 on Build GRCh38
rs759540004 Chr.16: 88716604-88716604 on Build GRCh38
rs772199276 Chr.16: 88716448-88716448 on Build GRCh38
rs776821033 Chr.16: 88716512-88716513 on Build GRCh38
rs750302950 Chr.16: 88716199-88716199 on Build GRCh38
rs903839506 Chr.16: 88716341-88716341 on Build GRCh38
rs938540081 Chr.16: 88716325-88716325 on Build GRCh38
rs1046326232 Chr.16: 88716554-88716554 on Build GRCh38
rs749646104 Chr.16: 88716440-88716440 on Build GRCh38
rs530682022 Chr.16: 88716426-88716426 on Build GRCh38
rs767465849 Chr.16: 88716195-88716195 on Build GRCh38
rs942739465 Chr.16: 88716315-88716315 on Build GRCh38
rs1039689538 Chr.16: 88716154-88716154 on Build GRCh38
rs368483543 Chr.16: 88716291-88716291 on Build GRCh38
rs781766948 Chr.16: 88716277-88716277 on Build GRCh38
rs928486875 Chr.16: 88716318-88716318 on Build GRCh38
rs1015058705 Chr.16: 88716210-88716210 on Build GRCh38
rs762678343 Chr.16: 88716523-88716523 on Build GRCh38
rs969205484 Chr.16: 88716373-88716373 on Build GRCh38
rs1048374764 Chr.16: 88716455-88716455 on Build GRCh38
rs557265812 Chr.16: 88716324-88716324 on Build GRCh38
rs751000081 Chr.16: 88716535-88716535 on Build GRCh38
rs747694984 Chr.16: 88716256-88716256 on Build GRCh38
rs1026404950 Chr.16: 88716435-88716435 on Build GRCh38
rs763707151 Chr.16: 88716528-88716528 on Build GRCh38
rs558982192 Chr.16: 88716606-88716606 on Build GRCh38
rs991295243 Chr.16: 88716437-88716437 on Build GRCh38
rs538983076 Chr.16: 88716290-88716290 on Build GRCh38
rs773146648 Chr.16: 88716452-88716452 on Build GRCh38
rs765053602 Chr.16: 88716508-88716508 on Build GRCh38
rs753588420 Chr.16: 88716225-88716225 on Build GRCh38
rs751616957 Chr.16: 88716185-88716185 on Build GRCh38
rs1002786139 Chr.16: 88716156-88716156 on Build GRCh38
rs980506949 Chr.16: 88716246-88716246 on Build GRCh38
rs1029084282 Chr.16: 88716522-88716522 on Build GRCh38
rs554886303 Chr.16: 88716598-88716598 on Build GRCh38
rs34325377 Chr.16: 88716594-88716594 on Build GRCh38
rs887784032 Chr.16: 88716350-88716350 on Build GRCh38
rs997612565 Chr.16: 88716520-88716520 on Build GRCh38
rs770497231 Chr.16: 88716551-88716551 on Build GRCh38
rs563334408 Chr.16: 88716424-88716424 on Build GRCh38
rs956728685 Chr.16: 88716216-88716216 on Build GRCh38
rs573391046 Chr.16: 88716384-88716384 on Build GRCh38
rs969069987 Chr.16: 88716245-88716245 on Build GRCh38
rs908840664 Chr.16: 88716253-88716253 on Build GRCh38
rs755943845 Chr.16: 88716201-88716201 on Build GRCh38
rs1041893774 Chr.16: 88716313-88716313 on Build GRCh38
rs953592517 Chr.16: 88716377-88716378 on Build GRCh38
rs754305193 Chr.16: 88716570-88716570 on Build GRCh38
rs549106702 Chr.16: 88716149-88716149 on Build GRCh38
rs1024673286 Chr.16: 88716241-88716241 on Build GRCh38
rs1016417537 Chr.16: 88716581-88716581 on Build GRCh38
rs764746748 Chr.16: 88716303-88716304 on Build GRCh38
rs764214848 Chr.16: 88716165-88716165 on Build GRCh38
rs565307908 Chr.16: 88716403-88716403 on Build GRCh38
rs768287647 Chr.16: 88716281-88716281 on Build GRCh38
rs770258207 Chr.16: 88716308-88716308 on Build GRCh38
rs35917730 Chr.16: 88716447-88716447 on Build GRCh38
rs754766447 Chr.16: 88716381-88716381 on Build GRCh38
rs534277817 Chr.16: 88716205-88716205 on Build GRCh38
rs552562531 Chr.16: 88716228-88716228 on Build GRCh38
rs200555745 Chr.16: 88716580-88716580 on Build GRCh38
rs760744612 Chr.16: 88716603-88716603 on Build GRCh38
rs761778012 Chr.16: 88716186-88716186 on Build GRCh38
rs912707774 Chr.16: 88716467-88716467 on Build GRCh38
rs1051232477 Chr.16: 88716287-88716287 on Build GRCh38
rs751878651 Chr.16: 88716352-88716352 on Build GRCh38
rs760099159 Chr.16: 88716593-88716593 on Build GRCh38
rs951572334 Chr.16: 88716441-88716441 on Build GRCh38
rs752462348 Chr.16: 88716518-88716518 on Build GRCh38
rs745394304 Chr.16: 88716556-88716556 on Build GRCh38
rs573353481 Chr.16: 88716599-88716599 on Build GRCh38
rs192028757 Chr.16: 88716150-88716150 on Build GRCh38
rs780507176 Chr.16: 88716422-88716422 on Build GRCh38
rs34352064 Chr.16: 88716369-88716369 on Build GRCh38

View Full Product Details