Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89279244-89279743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4941391 c.6868C>T p.P2290S Chr.16: 89279674-89279674 on Build GRCh38
6952509 c.6821C>T p.P2274L Chr.16: 89279721-89279721 on Build GRCh38
974850 c.7212G>A p.T2404T Chr.16: 89279330-89279330 on Build GRCh38
6953192 c.6883G>A p.E2295K Chr.16: 89279659-89279659 on Build GRCh38
4986444 c.6826C>T p.P2276S Chr.16: 89279716-89279716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759740258 Chr.16: 89279344-89279344 on Build GRCh38
rs888918193 Chr.16: 89279610-89279610 on Build GRCh38
rs767106606 Chr.16: 89279725-89279725 on Build GRCh38
rs754298757 Chr.16: 89279559-89279559 on Build GRCh38
rs954622305 Chr.16: 89279670-89279670 on Build GRCh38
rs773056661 Chr.16: 89279330-89279330 on Build GRCh38
rs532817153 Chr.16: 89279311-89279311 on Build GRCh38
rs760933701 Chr.16: 89279557-89279557 on Build GRCh38
rs763041579 Chr.16: 89279332-89279332 on Build GRCh38
rs761715813 Chr.16: 89279321-89279321 on Build GRCh38
rs918154229 Chr.16: 89279616-89279616 on Build GRCh38
rs779428707 Chr.16: 89279381-89279381 on Build GRCh38
rs548304688 Chr.16: 89279360-89279360 on Build GRCh38
rs1013086722 Chr.16: 89279664-89279664 on Build GRCh38
rs1024160802 Chr.16: 89279496-89279496 on Build GRCh38
rs1036666745 Chr.16: 89279410-89279410 on Build GRCh38
rs776557887 Chr.16: 89279319-89279319 on Build GRCh38
rs4031012 Chr.16: 89279520-89279520 on Build GRCh38
rs772115194 Chr.16: 89279680-89279680 on Build GRCh38
rs775584042 Chr.16: 89279683-89279683 on Build GRCh38
rs972009866 Chr.16: 89279563-89279563 on Build GRCh38
rs749176560 Chr.16: 89279481-89279481 on Build GRCh38
rs993290174 Chr.16: 89279662-89279662 on Build GRCh38
rs753291494 Chr.16: 89279407-89279409 on Build GRCh38
rs756552353 Chr.16: 89279294-89279294 on Build GRCh38
rs764436784 Chr.16: 89279714-89279714 on Build GRCh38
rs747911603 Chr.16: 89279431-89279431 on Build GRCh38
rs1018954790 Chr.16: 89279439-89279439 on Build GRCh38
rs565608120 Chr.16: 89279309-89279309 on Build GRCh38
rs999284780 Chr.16: 89279646-89279646 on Build GRCh38
rs1023089135 Chr.16: 89279666-89279666 on Build GRCh38
rs538103968 Chr.16: 89279600-89279600 on Build GRCh38
rs201957371 Chr.16: 89279716-89279716 on Build GRCh38
rs146575027 Chr.16: 89279618-89279618 on Build GRCh38
rs918068073 Chr.16: 89279723-89279723 on Build GRCh38
rs1007876265 Chr.16: 89279672-89279672 on Build GRCh38
rs903253722 Chr.16: 89279643-89279643 on Build GRCh38
rs561404796 Chr.16: 89279691-89279691 on Build GRCh38
rs756497363 Chr.16: 89279352-89279352 on Build GRCh38
rs745339361 Chr.16: 89279303-89279303 on Build GRCh38
rs113621769 Chr.16: 89279279-89279279 on Build GRCh38
rs550312255 Chr.16: 89279336-89279336 on Build GRCh38
rs991655950 Chr.16: 89279508-89279508 on Build GRCh38
rs994703276 Chr.16: 89279413-89279413 on Build GRCh38
rs999130123 Chr.16: 89279452-89279452 on Build GRCh38
rs897670799 Chr.16: 89279644-89279644 on Build GRCh38
rs377447921 Chr.16: 89279678-89279678 on Build GRCh38
rs755860697 Chr.16: 89279582-89279582 on Build GRCh38
rs982226775 Chr.16: 89279538-89279538 on Build GRCh38
rs767739621 Chr.16: 89279351-89279351 on Build GRCh38
rs747782477 Chr.16: 89279579-89279579 on Build GRCh38
rs148730499 Chr.16: 89279720-89279720 on Build GRCh38
rs927673241 Chr.16: 89279549-89279549 on Build GRCh38
rs957598792 Chr.16: 89279482-89279482 on Build GRCh38
rs746579362 Chr.16: 89279409-89279409 on Build GRCh38
rs780945512 Chr.16: 89279571-89279571 on Build GRCh38
rs893421448 Chr.16: 89279656-89279656 on Build GRCh38
rs1013550925 Chr.16: 89279487-89279487 on Build GRCh38
rs1033011026 Chr.16: 89279655-89279655 on Build GRCh38
rs916069779 Chr.16: 89279509-89279509 on Build GRCh38
rs978514758 Chr.16: 89279690-89279690 on Build GRCh38
rs967759428 Chr.16: 89279444-89279444 on Build GRCh38
rs896510600 Chr.16: 89279483-89279483 on Build GRCh38
rs764046639 Chr.16: 89279309-89279310 on Build GRCh38
rs778144283 Chr.16: 89279299-89279299 on Build GRCh38
rs370897874 Chr.16: 89279281-89279281 on Build GRCh38
rs774236725 Chr.16: 89279507-89279507 on Build GRCh38
rs575642464 Chr.16: 89279623-89279623 on Build GRCh38
rs746615405 Chr.16: 89279315-89279315 on Build GRCh38
rs1006297836 Chr.16: 89279525-89279525 on Build GRCh38
rs749121556 Chr.16: 89279607-89279607 on Build GRCh38
rs34069598 Chr.16: 89279627-89279627 on Build GRCh38
rs533765590 Chr.16: 89279510-89279510 on Build GRCh38
rs4785560 Chr.16: 89279414-89279414 on Build GRCh38
rs927582667 Chr.16: 89279375-89279375 on Build GRCh38
rs863225296 Chr.16: 89279362-89279362 on Build GRCh38
rs558301893 Chr.16: 89279564-89279564 on Build GRCh38
rs768492741 Chr.16: 89279318-89279318 on Build GRCh38
rs886042019 Chr.16: 89279308-89279308 on Build GRCh38
rs769501453 Chr.16: 89279455-89279455 on Build GRCh38
rs770957459 Chr.16: 89279501-89279501 on Build GRCh38
rs905177395 Chr.16: 89279632-89279632 on Build GRCh38
rs1053121119 Chr.16: 89279580-89279580 on Build GRCh38
rs535866230 Chr.16: 89279358-89279358 on Build GRCh38
rs753116112 Chr.16: 89279280-89279280 on Build GRCh38
rs889244275 Chr.16: 89279426-89279426 on Build GRCh38
rs750990942 Chr.16: 89279719-89279719 on Build GRCh38
rs886041822 Chr.16: 89279458-89279459 on Build GRCh38
rs576742682 Chr.16: 89279565-89279565 on Build GRCh38
rs781533044 Chr.16: 89279277-89279277 on Build GRCh38
rs756783131 Chr.16: 89279414-89279415 on Build GRCh38
rs947802091 Chr.16: 89279340-89279340 on Build GRCh38
rs986291618 Chr.16: 89279401-89279401 on Build GRCh38
rs867650813 Chr.16: 89279484-89279484 on Build GRCh38
rs767682539 Chr.16: 89279528-89279528 on Build GRCh38
rs1006348798 Chr.16: 89279635-89279635 on Build GRCh38
rs773110185 Chr.16: 89279471-89279471 on Build GRCh38
rs573355746 Chr.16: 89279715-89279715 on Build GRCh38
rs754567522 Chr.16: 89279568-89279568 on Build GRCh38
rs899013747 Chr.16: 89279411-89279411 on Build GRCh38
rs377374681 Chr.16: 89279356-89279356 on Build GRCh38
rs886041942 Chr.16: 89279480-89279480 on Build GRCh38
rs374668457 Chr.16: 89279674-89279674 on Build GRCh38
rs904015120 Chr.16: 89279437-89279437 on Build GRCh38
rs374714506 Chr.16: 89279335-89279335 on Build GRCh38
rs940623226 Chr.16: 89279406-89279406 on Build GRCh38
rs969115786 Chr.16: 89279515-89279515 on Build GRCh38
rs755232209 Chr.16: 89279266-89279266 on Build GRCh38
rs780996696 Chr.16: 89279427-89279427 on Build GRCh38
rs961871894 Chr.16: 89279561-89279561 on Build GRCh38
rs775708950 Chr.16: 89279546-89279546 on Build GRCh38

View Full Product Details