Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4996721-4997218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INCA1
Entrez Gene ID
388324
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Gene Aliases
HSD45
UniGene
Hs.658926
Gene Chromosome Location
Chr.17: 4988130-4997933 on Build GRCh38
Transcripts
NM_001167985.1 NM_001167986.1 NM_001167987.1 NM_213726.2 XM_005256627.3 XM_005256628.4 XM_005256629.1 XM_006721517.1 XM_006721518.2 XM_006721519.3 XM_011523832.2 XM_011523834.2

SNP Detail

SNP ID Chromosome Location
rs529591884 Chr.17: 4996930-4996930 on Build GRCh38
rs1003082665 Chr.17: 4996924-4996924 on Build GRCh38
rs894291335 Chr.17: 4997182-4997182 on Build GRCh38
rs1011404969 Chr.17: 4997189-4997189 on Build GRCh38
rs993533962 Chr.17: 4996906-4996906 on Build GRCh38
rs914253946 Chr.17: 4996782-4996782 on Build GRCh38
rs889343431 Chr.17: 4996875-4996875 on Build GRCh38
rs952262983 Chr.17: 4996747-4996747 on Build GRCh38
rs958654728 Chr.17: 4996753-4996753 on Build GRCh38
rs757286926 Chr.17: 4996987-4996987 on Build GRCh38
rs928496612 Chr.17: 4996900-4996900 on Build GRCh38
rs1021489159 Chr.17: 4997190-4997190 on Build GRCh38
rs1021521642 Chr.17: 4996980-4996980 on Build GRCh38
rs185196036 Chr.17: 4996931-4996931 on Build GRCh38
rs954503668 Chr.17: 4997050-4997050 on Build GRCh38
rs1025053774 Chr.17: 4996914-4996914 on Build GRCh38
rs923064279 Chr.17: 4996832-4996832 on Build GRCh38
rs1036932575 Chr.17: 4997199-4997199 on Build GRCh38
rs1050618638 Chr.17: 4996854-4996854 on Build GRCh38
rs990096041 Chr.17: 4996804-4996805 on Build GRCh38
rs750236636 Chr.17: 4997065-4997065 on Build GRCh38
rs554236839 Chr.17: 4997093-4997093 on Build GRCh38
rs189275307 Chr.17: 4996836-4996836 on Build GRCh38
rs551325731 Chr.17: 4997054-4997054 on Build GRCh38
rs1046437789 Chr.17: 4997163-4997163 on Build GRCh38
rs527577293 Chr.17: 4996957-4996957 on Build GRCh38
rs928540943 Chr.17: 4997134-4997134 on Build GRCh38
rs990170099 Chr.17: 4997059-4997059 on Build GRCh38
rs1044253836 Chr.17: 4996871-4996871 on Build GRCh38
rs977123441 Chr.17: 4996805-4996805 on Build GRCh38
rs1048136595 Chr.17: 4997193-4997193 on Build GRCh38
rs945746175 Chr.17: 4996790-4996790 on Build GRCh38
rs1055649313 Chr.17: 4997181-4997181 on Build GRCh38
rs922984343 Chr.17: 4997028-4997028 on Build GRCh38
rs1037985330 Chr.17: 4996893-4996893 on Build GRCh38
rs563531905 Chr.17: 4996912-4996912 on Build GRCh38
rs571389667 Chr.17: 4997081-4997081 on Build GRCh38
rs1018263410 Chr.17: 4996965-4996965 on Build GRCh38
rs924685646 Chr.17: 4996794-4996794 on Build GRCh38
rs577694708 Chr.17: 4996845-4996845 on Build GRCh38
rs906589733 Chr.17: 4997164-4997164 on Build GRCh38
rs986674880 Chr.17: 4997073-4997073 on Build GRCh38
rs1037954030 Chr.17: 4997101-4997101 on Build GRCh38
rs35584694 Chr.17: 4997152-4997152 on Build GRCh38
rs906628642 Chr.17: 4996921-4996921 on Build GRCh38
rs889433646 Chr.17: 4996940-4996940 on Build GRCh38
rs967270567 Chr.17: 4997192-4997192 on Build GRCh38
rs146336867 Chr.17: 4997169-4997169 on Build GRCh38
rs976224319 Chr.17: 4996774-4996774 on Build GRCh38
rs183788742 Chr.17: 4996762-4996762 on Build GRCh38
rs536258955 Chr.17: 4997019-4997019 on Build GRCh38
rs966574195 Chr.17: 4996773-4996773 on Build GRCh38
rs919540582 Chr.17: 4997139-4997139 on Build GRCh38
rs371859357 Chr.17: 4996928-4996928 on Build GRCh38
rs929255176 Chr.17: 4997143-4997143 on Build GRCh38
rs969998556 Chr.17: 4997091-4997091 on Build GRCh38
rs942308721 Chr.17: 4996879-4996879 on Build GRCh38
rs995202226 Chr.17: 4996939-4996939 on Build GRCh38
rs998074075 Chr.17: 4997009-4997009 on Build GRCh38
rs181983649 Chr.17: 4996842-4996842 on Build GRCh38
rs149787223 Chr.17: 4996898-4996898 on Build GRCh38
rs528092860 Chr.17: 4997014-4997014 on Build GRCh38
rs956087775 Chr.17: 4996795-4996795 on Build GRCh38
rs765736353 Chr.17: 4996815-4996816 on Build GRCh38
rs566773078 Chr.17: 4997116-4997116 on Build GRCh38

View Full Product Details