Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5394801-5395265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP88
Entrez Gene ID
4927
Gene Name
nucleoporin 88
Gene Aliases
-
UniGene
Hs.584784
Gene Chromosome Location
Chr.17: 5384833-5419739 on Build GRCh38
Transcripts
NM_001320653.1 NM_002532.5 XM_011523893.2 XM_017024694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7148860 c.1342T>C p.F448L Chr.17: 5394931-5394931 on Build GRCh38
1563753 c.1302T>G p.D434E Chr.17: 5394971-5394971 on Build GRCh38
5755077 c.1327A>G p.T443A Chr.17: 5394946-5394946 on Build GRCh38
3519696 c.1356C>T p.I452I Chr.17: 5394917-5394917 on Build GRCh38
1663185 c.1375C>A p.P459T Chr.17: 5394898-5394898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765175531 Chr.17: 5395002-5395002 on Build GRCh38
rs746678761 Chr.17: 5394964-5394964 on Build GRCh38
rs138136065 Chr.17: 5395126-5395126 on Build GRCh38
rs190617828 Chr.17: 5395227-5395227 on Build GRCh38
rs748207088 Chr.17: 5395173-5395173 on Build GRCh38
rs150439515 Chr.17: 5395044-5395044 on Build GRCh38
rs768620677 Chr.17: 5394843-5394843 on Build GRCh38
rs2309372 Chr.17: 5395076-5395076 on Build GRCh38
rs892700361 Chr.17: 5395222-5395222 on Build GRCh38
rs766133225 Chr.17: 5395018-5395018 on Build GRCh38
rs984161738 Chr.17: 5395064-5395064 on Build GRCh38
rs771229029 Chr.17: 5394988-5394988 on Build GRCh38
rs147533029 Chr.17: 5394968-5394968 on Build GRCh38
rs1036232735 Chr.17: 5394830-5394830 on Build GRCh38
rs376528237 Chr.17: 5394939-5394939 on Build GRCh38
rs770218779 Chr.17: 5394970-5394970 on Build GRCh38
rs971875578 Chr.17: 5395098-5395098 on Build GRCh38
rs2871203 Chr.17: 5395170-5395170 on Build GRCh38
rs750179023 Chr.17: 5394936-5394936 on Build GRCh38
rs574618342 Chr.17: 5394917-5394917 on Build GRCh38
rs775648901 Chr.17: 5394999-5394999 on Build GRCh38
rs892752715 Chr.17: 5394938-5394938 on Build GRCh38
rs776904261 Chr.17: 5394998-5395001 on Build GRCh38
rs755189254 Chr.17: 5395089-5395089 on Build GRCh38
rs757330790 Chr.17: 5394929-5394929 on Build GRCh38
rs369692555 Chr.17: 5394985-5394985 on Build GRCh38
rs114441146 Chr.17: 5395138-5395138 on Build GRCh38
rs376624528 Chr.17: 5395025-5395025 on Build GRCh38
rs532651292 Chr.17: 5395221-5395221 on Build GRCh38
rs1001039477 Chr.17: 5394924-5394924 on Build GRCh38
rs367700903 Chr.17: 5394855-5394855 on Build GRCh38
rs536804168 Chr.17: 5395177-5395177 on Build GRCh38
rs1043269678 Chr.17: 5395162-5395162 on Build GRCh38
rs567755038 Chr.17: 5395171-5395171 on Build GRCh38
rs891860376 Chr.17: 5394909-5394909 on Build GRCh38
rs371621216 Chr.17: 5394839-5394839 on Build GRCh38
rs758178530 Chr.17: 5395013-5395013 on Build GRCh38
rs753759158 Chr.17: 5394949-5394949 on Build GRCh38
rs750361574 Chr.17: 5395012-5395012 on Build GRCh38
rs987416588 Chr.17: 5395139-5395139 on Build GRCh38
rs766563515 Chr.17: 5394899-5394899 on Build GRCh38
rs975066821 Chr.17: 5395125-5395125 on Build GRCh38
rs371452378 Chr.17: 5394871-5394871 on Build GRCh38
rs774457202 Chr.17: 5394902-5394902 on Build GRCh38
rs945649523 Chr.17: 5395230-5395230 on Build GRCh38
rs767756522 Chr.17: 5394912-5394912 on Build GRCh38
rs369859181 Chr.17: 5394875-5394875 on Build GRCh38
rs763230266 Chr.17: 5394877-5394877 on Build GRCh38
rs773605064 Chr.17: 5394980-5394980 on Build GRCh38
rs1020225250 Chr.17: 5394960-5394960 on Build GRCh38
rs913349980 Chr.17: 5395150-5395150 on Build GRCh38
rs765274280 Chr.17: 5394935-5394935 on Build GRCh38
rs751206489 Chr.17: 5395019-5395019 on Build GRCh38
rs937492993 Chr.17: 5395175-5395175 on Build GRCh38
rs138380831 Chr.17: 5394908-5394908 on Build GRCh38
rs199978623 Chr.17: 5394866-5394866 on Build GRCh38
rs564960543 Chr.17: 5395136-5395136 on Build GRCh38
rs936332585 Chr.17: 5395176-5395176 on Build GRCh38
rs749614981 Chr.17: 5394983-5394983 on Build GRCh38
rs771691521 Chr.17: 5394852-5394852 on Build GRCh38
rs904871528 Chr.17: 5394904-5394904 on Build GRCh38
rs1806240 Chr.17: 5395000-5395000 on Build GRCh38
rs759896374 Chr.17: 5394997-5394997 on Build GRCh38
rs747599946 Chr.17: 5395030-5395030 on Build GRCh38
rs774611314 Chr.17: 5394993-5394993 on Build GRCh38
rs946128164 Chr.17: 5395151-5395151 on Build GRCh38
rs779719274 Chr.17: 5394943-5394943 on Build GRCh38
rs746481575 Chr.17: 5394862-5394862 on Build GRCh38
rs767701288 Chr.17: 5394998-5394998 on Build GRCh38
rs185861715 Chr.17: 5394873-5394873 on Build GRCh38
rs143358064 Chr.17: 5394967-5394967 on Build GRCh38
rs754689198 Chr.17: 5395024-5395024 on Build GRCh38
rs917637463 Chr.17: 5394824-5394824 on Build GRCh38
rs777088428 Chr.17: 5394868-5394868 on Build GRCh38
rs933291232 Chr.17: 5394892-5394892 on Build GRCh38

View Full Product Details