Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29264145-29264654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUFIP2
Entrez Gene ID
57532
Gene Name
NUFIP2, FMR1 interacting protein 2
Gene Aliases
182-FIP, 82-FIP, FIP-82, PIG1
UniGene
Hs.462598
Gene Chromosome Location
Chr.17: 29255836-29294148 on Build GRCh38
Transcripts
NM_020772.2 XM_017024896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778484 c.2050A>G p.I684V Chr.17: 29264577-29264577 on Build GRCh38
4769275 c.2058C>A p.Y686* Chr.17: 29264569-29264569 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757731261 Chr.17: 29264486-29264486 on Build GRCh38
rs999568376 Chr.17: 29264345-29264345 on Build GRCh38
rs749514785 Chr.17: 29264620-29264620 on Build GRCh38
rs780876515 Chr.17: 29264498-29264498 on Build GRCh38
rs903256730 Chr.17: 29264311-29264311 on Build GRCh38
rs1053669913 Chr.17: 29264214-29264214 on Build GRCh38
rs529020414 Chr.17: 29264368-29264368 on Build GRCh38
rs1003394824 Chr.17: 29264333-29264333 on Build GRCh38
rs761053643 Chr.17: 29264542-29264542 on Build GRCh38
rs200721235 Chr.17: 29264404-29264405 on Build GRCh38
rs894451074 Chr.17: 29264361-29264361 on Build GRCh38
rs774470686 Chr.17: 29264541-29264541 on Build GRCh38
rs944833218 Chr.17: 29264171-29264171 on Build GRCh38
rs778653778 Chr.17: 29264415-29264415 on Build GRCh38
rs377244438 Chr.17: 29264453-29264453 on Build GRCh38
rs1011015804 Chr.17: 29264256-29264259 on Build GRCh38
rs763549861 Chr.17: 29264566-29264566 on Build GRCh38
rs757184414 Chr.17: 29264495-29264495 on Build GRCh38
rs757961967 Chr.17: 29264458-29264458 on Build GRCh38
rs368319103 Chr.17: 29264544-29264544 on Build GRCh38
rs201190853 Chr.17: 29264519-29264519 on Build GRCh38
rs759737105 Chr.17: 29264602-29264603 on Build GRCh38
rs371560511 Chr.17: 29264492-29264492 on Build GRCh38
rs112392860 Chr.17: 29264379-29264379 on Build GRCh38
rs773725068 Chr.17: 29264536-29264536 on Build GRCh38
rs879152719 Chr.17: 29264404-29264404 on Build GRCh38
rs568177581 Chr.17: 29264360-29264360 on Build GRCh38
rs200428740 Chr.17: 29264585-29264585 on Build GRCh38
rs181872571 Chr.17: 29264625-29264625 on Build GRCh38
rs993034128 Chr.17: 29264376-29264377 on Build GRCh38
rs751036631 Chr.17: 29264567-29264567 on Build GRCh38
rs189825211 Chr.17: 29264514-29264514 on Build GRCh38
rs1057233338 Chr.17: 29264338-29264338 on Build GRCh38
rs770011613 Chr.17: 29264510-29264510 on Build GRCh38
rs368710244 Chr.17: 29264547-29264547 on Build GRCh38
rs762259195 Chr.17: 29264525-29264525 on Build GRCh38
rs374136415 Chr.17: 29264508-29264508 on Build GRCh38
rs3785961 Chr.17: 29264307-29264307 on Build GRCh38
rs781045101 Chr.17: 29264590-29264590 on Build GRCh38
rs755963790 Chr.17: 29264599-29264599 on Build GRCh38
rs767779688 Chr.17: 29264607-29264610 on Build GRCh38
rs944990655 Chr.17: 29264553-29264553 on Build GRCh38
rs565691805 Chr.17: 29264448-29264448 on Build GRCh38
rs537276840 Chr.17: 29264624-29264624 on Build GRCh38
rs774460218 Chr.17: 29264520-29264520 on Build GRCh38
rs1016428621 Chr.17: 29264384-29264384 on Build GRCh38
rs752767473 Chr.17: 29264377-29264378 on Build GRCh38
rs745804161 Chr.17: 29264596-29264596 on Build GRCh38
rs150387941 Chr.17: 29264376-29264377 on Build GRCh38
rs945189909 Chr.17: 29264299-29264299 on Build GRCh38
rs566461350 Chr.17: 29264393-29264393 on Build GRCh38
rs946898676 Chr.17: 29264617-29264617 on Build GRCh38
rs768019365 Chr.17: 29264534-29264534 on Build GRCh38
rs1039211250 Chr.17: 29264177-29264177 on Build GRCh38
rs757160286 Chr.17: 29264569-29264569 on Build GRCh38
rs140777493 Chr.17: 29264245-29264245 on Build GRCh38
rs758043741 Chr.17: 29264556-29264556 on Build GRCh38
rs370091001 Chr.17: 29264457-29264457 on Build GRCh38
rs779804093 Chr.17: 29264610-29264610 on Build GRCh38
rs1022766511 Chr.17: 29264405-29264405 on Build GRCh38
rs943030050 Chr.17: 29264213-29264213 on Build GRCh38
rs373146104 Chr.17: 29264375-29264376 on Build GRCh38

View Full Product Details