Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44374906-44375406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA2B
Entrez Gene ID
3674
Gene Name
integrin subunit alpha 2b
Gene Aliases
BDPLT16, BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, PPP1R93
UniGene
Hs.411312
Gene Chromosome Location
Chr.17: 44372181-44389601 on Build GRCh38
Transcripts
NM_000419.4 XM_011524749.1 XM_011524750.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402655 c.2772G>T p.Q924H Chr.17: 44375067-44375067 on Build GRCh38
4629125 c.2735A>G p.D912G Chr.17: 44375104-44375104 on Build GRCh38
4484657 c.2835C>T p.L945L Chr.17: 44375004-44375004 on Build GRCh38
1383666 c.2739G>A p.S913S Chr.17: 44375100-44375100 on Build GRCh38
4881555 c.2741C>T p.A914V Chr.17: 44375098-44375098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557632223 Chr.17: 44374942-44374942 on Build GRCh38
rs77458039 Chr.17: 44375039-44375039 on Build GRCh38
rs868106253 Chr.17: 44375106-44375106 on Build GRCh38
rs776470835 Chr.17: 44374950-44374950 on Build GRCh38
rs753790795 Chr.17: 44375174-44375174 on Build GRCh38
rs911592439 Chr.17: 44375295-44375295 on Build GRCh38
rs540296465 Chr.17: 44375126-44375126 on Build GRCh38
rs1028948312 Chr.17: 44374983-44374983 on Build GRCh38
rs564223176 Chr.17: 44375209-44375209 on Build GRCh38
rs2521613 Chr.17: 44374956-44374956 on Build GRCh38
rs914498453 Chr.17: 44375151-44375151 on Build GRCh38
rs1004292813 Chr.17: 44375170-44375170 on Build GRCh38
rs932875924 Chr.17: 44375343-44375343 on Build GRCh38
rs924329423 Chr.17: 44375171-44375171 on Build GRCh38
rs76572092 Chr.17: 44375082-44375091 on Build GRCh38
rs1015399230 Chr.17: 44375085-44375085 on Build GRCh38
rs1006186368 Chr.17: 44375025-44375025 on Build GRCh38
rs759486426 Chr.17: 44374957-44374957 on Build GRCh38
rs897885305 Chr.17: 44375346-44375346 on Build GRCh38
rs201904682 Chr.17: 44375143-44375143 on Build GRCh38
rs763637665 Chr.17: 44375057-44375057 on Build GRCh38
rs536034015 Chr.17: 44374927-44374927 on Build GRCh38
rs371546196 Chr.17: 44375144-44375144 on Build GRCh38
rs977134206 Chr.17: 44375169-44375169 on Build GRCh38
rs541221026 Chr.17: 44375156-44375156 on Build GRCh38
rs868549069 Chr.17: 44375246-44375246 on Build GRCh38
rs774105556 Chr.17: 44375054-44375055 on Build GRCh38
rs974789703 Chr.17: 44375338-44375338 on Build GRCh38
rs199961276 Chr.17: 44375140-44375140 on Build GRCh38
rs532951276 Chr.17: 44375157-44375157 on Build GRCh38
rs944396673 Chr.17: 44375321-44375321 on Build GRCh38
rs1050603825 Chr.17: 44375227-44375227 on Build GRCh38
rs558502381 Chr.17: 44374985-44374985 on Build GRCh38
rs944327941 Chr.17: 44375045-44375045 on Build GRCh38
rs1002995837 Chr.17: 44375377-44375377 on Build GRCh38
rs573244991 Chr.17: 44375082-44375082 on Build GRCh38
rs1040925788 Chr.17: 44375192-44375192 on Build GRCh38
rs562196345 Chr.17: 44375133-44375133 on Build GRCh38
rs977808919 Chr.17: 44375271-44375271 on Build GRCh38
rs546476765 Chr.17: 44374973-44374973 on Build GRCh38
rs551295330 Chr.17: 44375199-44375199 on Build GRCh38
rs751210971 Chr.17: 44375073-44375073 on Build GRCh38
rs762404002 Chr.17: 44374988-44374988 on Build GRCh38
rs1033613373 Chr.17: 44375184-44375184 on Build GRCh38
rs867277020 Chr.17: 44375048-44375048 on Build GRCh38
rs766712350 Chr.17: 44375100-44375100 on Build GRCh38
rs777941686 Chr.17: 44375269-44375269 on Build GRCh38
rs943833152 Chr.17: 44375179-44375179 on Build GRCh38
rs79808582 Chr.17: 44375207-44375207 on Build GRCh38
rs921552973 Chr.17: 44375342-44375342 on Build GRCh38
rs555353533 Chr.17: 44374939-44374939 on Build GRCh38
rs113397452 Chr.17: 44375345-44375345 on Build GRCh38
rs1041303475 Chr.17: 44375049-44375052 on Build GRCh38
rs200677976 Chr.17: 44374970-44374970 on Build GRCh38
rs753923342 Chr.17: 44374953-44374971 on Build GRCh38
rs772192587 Chr.17: 44375175-44375175 on Build GRCh38
rs888047077 Chr.17: 44375233-44375233 on Build GRCh38
rs962164705 Chr.17: 44375101-44375101 on Build GRCh38

View Full Product Details