Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58998079-58998612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
UniGene
Hs.579079
Gene Chromosome Location
Chr.17: 58968068-59106921 on Build GRCh38
Transcripts
NM_001005207.3 NM_001320987.1 NM_001320988.1 NM_001320989.1 NM_001320990.1 NM_015294.4 XM_005257385.1 XM_011524831.1 XM_011524832.2 XM_011524833.1 XM_011524834.1 XM_011524836.1 XM_017024662.1 XM_017024663.1 XM_017024664.1 XM_017024665.1 XM_017024667.1 XM_017024668.1 XM_017024670.1 XM_017024672.1 XM_017024673.1

SNP Detail

SNP ID Chromosome Location
rs12451581 Chr.17: 58998293-58998293 on Build GRCh38
rs146140916 Chr.17: 58998288-58998288 on Build GRCh38
rs542550520 Chr.17: 58998196-58998196 on Build GRCh38
rs964107817 Chr.17: 58998377-58998377 on Build GRCh38
rs911199869 Chr.17: 58998551-58998551 on Build GRCh38
rs918443515 Chr.17: 58998483-58998483 on Build GRCh38
rs942696454 Chr.17: 58998576-58998576 on Build GRCh38
rs567981472 Chr.17: 58998552-58998552 on Build GRCh38
rs550696270 Chr.17: 58998517-58998517 on Build GRCh38
rs182050495 Chr.17: 58998141-58998141 on Build GRCh38
rs897812975 Chr.17: 58998163-58998164 on Build GRCh38
rs1056454129 Chr.17: 58998157-58998157 on Build GRCh38
rs867901457 Chr.17: 58998443-58998443 on Build GRCh38
rs1041100171 Chr.17: 58998577-58998577 on Build GRCh38
rs1041545874 Chr.17: 58998221-58998221 on Build GRCh38
rs1035856048 Chr.17: 58998470-58998470 on Build GRCh38
rs926744685 Chr.17: 58998502-58998502 on Build GRCh38
rs952616121 Chr.17: 58998312-58998312 on Build GRCh38
rs914 Chr.17: 58998352-58998352 on Build GRCh38
rs879406619 Chr.17: 58998284-58998284 on Build GRCh38
rs1000109205 Chr.17: 58998292-58998292 on Build GRCh38
rs972976089 Chr.17: 58998481-58998481 on Build GRCh38
rs764131923 Chr.17: 58998452-58998452 on Build GRCh38
rs142111745 Chr.17: 58998541-58998541 on Build GRCh38
rs368742554 Chr.17: 58998354-58998354 on Build GRCh38
rs550928857 Chr.17: 58998121-58998121 on Build GRCh38
rs186389005 Chr.17: 58998347-58998347 on Build GRCh38
rs773584114 Chr.17: 58998127-58998127 on Build GRCh38
rs779235712 Chr.17: 58998540-58998543 on Build GRCh38
rs1001267233 Chr.17: 58998449-58998449 on Build GRCh38
rs752596150 Chr.17: 58998255-58998255 on Build GRCh38
rs1045612852 Chr.17: 58998493-58998493 on Build GRCh38
rs529727378 Chr.17: 58998539-58998539 on Build GRCh38
rs988389887 Chr.17: 58998471-58998471 on Build GRCh38
rs1021286743 Chr.17: 58998414-58998414 on Build GRCh38
rs3826354 Chr.17: 58998416-58998416 on Build GRCh38
rs939441140 Chr.17: 58998516-58998516 on Build GRCh38
rs1005760604 Chr.17: 58998348-58998348 on Build GRCh38
rs959852232 Chr.17: 58998480-58998480 on Build GRCh38
rs112918162 Chr.17: 58998400-58998400 on Build GRCh38
rs994115654 Chr.17: 58998164-58998164 on Build GRCh38
rs532079099 Chr.17: 58998464-58998464 on Build GRCh38
rs562334292 Chr.17: 58998538-58998538 on Build GRCh38
rs946912901 Chr.17: 58998487-58998487 on Build GRCh38
rs561272285 Chr.17: 58998282-58998282 on Build GRCh38
rs116695008 Chr.17: 58998140-58998140 on Build GRCh38
rs1031200749 Chr.17: 58998331-58998331 on Build GRCh38
rs753733313 Chr.17: 58998514-58998514 on Build GRCh38

View Full Product Details