Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59234475-59234978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD1
Entrez Gene ID
284161
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Gene Aliases
GDE4
UniGene
Hs.631744
Gene Chromosome Location
Chr.17: 59220467-59275970 on Build GRCh38
Transcripts
NM_001165993.1 NM_001165994.1 NM_182569.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
131213 c.183G>T p.Q61H Chr.17: 59234532-59234532 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764348959 Chr.17: 59234562-59234562 on Build GRCh38
rs927984604 Chr.17: 59234647-59234647 on Build GRCh38
rs910769642 Chr.17: 59234780-59234780 on Build GRCh38
rs557885019 Chr.17: 59234803-59234803 on Build GRCh38
rs956444849 Chr.17: 59234735-59234735 on Build GRCh38
rs747492981 Chr.17: 59234552-59234552 on Build GRCh38
rs946638903 Chr.17: 59234777-59234777 on Build GRCh38
rs560627497 Chr.17: 59234814-59234814 on Build GRCh38
rs750236960 Chr.17: 59234630-59234630 on Build GRCh38
rs969342359 Chr.17: 59234613-59234613 on Build GRCh38
rs1018359850 Chr.17: 59234706-59234706 on Build GRCh38
rs745946710 Chr.17: 59234531-59234531 on Build GRCh38
rs539795600 Chr.17: 59234756-59234756 on Build GRCh38
rs952285562 Chr.17: 59234673-59234673 on Build GRCh38
rs1049275973 Chr.17: 59234866-59234866 on Build GRCh38
rs765955063 Chr.17: 59234585-59234585 on Build GRCh38
rs182177921 Chr.17: 59234727-59234727 on Build GRCh38
rs987947903 Chr.17: 59234745-59234745 on Build GRCh38
rs897940195 Chr.17: 59234860-59234860 on Build GRCh38
rs1050678965 Chr.17: 59234774-59234774 on Build GRCh38
rs993689068 Chr.17: 59234619-59234619 on Build GRCh38
rs568362633 Chr.17: 59234635-59234635 on Build GRCh38
rs762184779 Chr.17: 59234575-59234575 on Build GRCh38
rs779137228 Chr.17: 59234522-59234522 on Build GRCh38
rs776886710 Chr.17: 59234564-59234564 on Build GRCh38
rs151029801 Chr.17: 59234591-59234592 on Build GRCh38
rs972563052 Chr.17: 59234715-59234715 on Build GRCh38
rs114944626 Chr.17: 59234862-59234862 on Build GRCh38
rs939404789 Chr.17: 59234845-59234845 on Build GRCh38
rs892958425 Chr.17: 59234936-59234936 on Build GRCh38
rs982054900 Chr.17: 59234643-59234643 on Build GRCh38
rs769102530 Chr.17: 59234554-59234554 on Build GRCh38
rs188037593 Chr.17: 59234591-59234591 on Build GRCh38
rs905154952 Chr.17: 59234551-59234551 on Build GRCh38
rs776195386 Chr.17: 59234546-59234546 on Build GRCh38
rs547521038 Chr.17: 59234710-59234710 on Build GRCh38
rs1054648054 Chr.17: 59234931-59234931 on Build GRCh38
rs762792524 Chr.17: 59234707-59234707 on Build GRCh38
rs749071257 Chr.17: 59234499-59234499 on Build GRCh38
rs745558097 Chr.17: 59234578-59234578 on Build GRCh38
rs902853152 Chr.17: 59234854-59234854 on Build GRCh38
rs1007785453 Chr.17: 59234680-59234680 on Build GRCh38
rs772272278 Chr.17: 59234543-59234543 on Build GRCh38
rs915053873 Chr.17: 59234768-59234768 on Build GRCh38
rs529337278 Chr.17: 59234650-59234650 on Build GRCh38
rs537348659 Chr.17: 59234836-59234836 on Build GRCh38
rs9895821 Chr.17: 59234851-59234851 on Build GRCh38
rs976822126 Chr.17: 59234721-59234721 on Build GRCh38
rs757443617 Chr.17: 59234517-59234517 on Build GRCh38
rs957470978 Chr.17: 59234588-59234588 on Build GRCh38
rs549915099 Chr.17: 59234614-59234614 on Build GRCh38
rs527244093 Chr.17: 59234863-59234863 on Build GRCh38
rs563100888 Chr.17: 59234520-59234520 on Build GRCh38
rs887905754 Chr.17: 59234923-59234923 on Build GRCh38
rs531921701 Chr.17: 59234603-59234603 on Build GRCh38
rs533649819 Chr.17: 59234624-59234624 on Build GRCh38
rs1004047124 Chr.17: 59234582-59234582 on Build GRCh38

View Full Product Details