Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61943635-61944178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED13
Entrez Gene ID
9969
Gene Name
mediator complex subunit 13
Gene Aliases
ARC250, DRIP250, HSPC221, THRAP1, TRAP240
UniGene
Hs.282678
Gene Chromosome Location
Chr.17: 61942605-62065983 on Build GRCh38
Transcripts
NM_005121.2 XM_011525551.2 XM_011525553.2

SNP Detail

SNP ID Chromosome Location
rs908390203 Chr.17: 61944000-61944000 on Build GRCh38
rs979976597 Chr.17: 61943878-61943878 on Build GRCh38
rs143989721 Chr.17: 61943851-61943851 on Build GRCh38
rs889282880 Chr.17: 61943762-61943762 on Build GRCh38
rs992737736 Chr.17: 61943903-61943903 on Build GRCh38
rs956727433 Chr.17: 61943846-61943847 on Build GRCh38
rs1018830181 Chr.17: 61943749-61943749 on Build GRCh38
rs927079575 Chr.17: 61943899-61943902 on Build GRCh38
rs868751272 Chr.17: 61943820-61943820 on Build GRCh38
rs145960183 Chr.17: 61943965-61943965 on Build GRCh38
rs552080124 Chr.17: 61943975-61943975 on Build GRCh38
rs916035836 Chr.17: 61943855-61943855 on Build GRCh38
rs996519932 Chr.17: 61943689-61943689 on Build GRCh38
rs919587302 Chr.17: 61944046-61944046 on Build GRCh38
rs938619707 Chr.17: 61943900-61943900 on Build GRCh38
rs999124119 Chr.17: 61943829-61943829 on Build GRCh38
rs1009093651 Chr.17: 61943767-61943767 on Build GRCh38
rs918502289 Chr.17: 61943906-61943906 on Build GRCh38
rs971618589 Chr.17: 61943936-61943936 on Build GRCh38
rs981327062 Chr.17: 61943967-61943967 on Build GRCh38
rs537458098 Chr.17: 61944063-61944063 on Build GRCh38
rs551068455 Chr.17: 61944111-61944111 on Build GRCh38
rs1031017586 Chr.17: 61943841-61943841 on Build GRCh38
rs1049927413 Chr.17: 61944082-61944082 on Build GRCh38
rs866689715 Chr.17: 61944023-61944023 on Build GRCh38
rs967624850 Chr.17: 61943809-61943809 on Build GRCh38
rs755660658 Chr.17: 61944012-61944012 on Build GRCh38
rs370237483 Chr.17: 61943990-61943990 on Build GRCh38
rs56328668 Chr.17: 61944017-61944017 on Build GRCh38
rs148391388 Chr.17: 61943848-61943848 on Build GRCh38
rs529398391 Chr.17: 61943838-61943838 on Build GRCh38
rs1008409105 Chr.17: 61944153-61944153 on Build GRCh38
rs1032970962 Chr.17: 61943844-61943844 on Build GRCh38
rs1040521028 Chr.17: 61944155-61944155 on Build GRCh38
rs1039877997 Chr.17: 61944079-61944079 on Build GRCh38
rs185065407 Chr.17: 61943668-61943668 on Build GRCh38
rs892707910 Chr.17: 61944141-61944141 on Build GRCh38
rs939902821 Chr.17: 61944016-61944016 on Build GRCh38
rs1031280113 Chr.17: 61944126-61944126 on Build GRCh38
rs879490222 Chr.17: 61943776-61943776 on Build GRCh38
rs1028064936 Chr.17: 61943725-61943725 on Build GRCh38
rs866193586 Chr.17: 61943840-61943840 on Build GRCh38
rs528778109 Chr.17: 61943941-61943941 on Build GRCh38

View Full Product Details