Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76683763-76684262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXRA7
Entrez Gene ID
439921
Gene Name
matrix remodeling associated 7
Gene Aliases
PS1TP1, TMAP1
UniGene
Hs.250723
Gene Chromosome Location
Chr.17: 76673650-76711005 on Build GRCh38
Transcripts
NM_001008528.2 NM_001008529.2 NM_198530.3 NR_130926.1 NR_130927.1 NR_130928.1 XM_005257382.2 XM_005257383.2 XM_005257384.2 XM_011524830.1

SNP Detail

SNP ID Chromosome Location
rs867657389 Chr.17: 76684085-76684085 on Build GRCh38
rs566545238 Chr.17: 76684183-76684183 on Build GRCh38
rs750068207 Chr.17: 76683894-76683894 on Build GRCh38
rs764906211 Chr.17: 76683886-76683886 on Build GRCh38
rs977883410 Chr.17: 76684159-76684159 on Build GRCh38
rs758173345 Chr.17: 76683896-76683896 on Build GRCh38
rs3803741 Chr.17: 76684081-76684081 on Build GRCh38
rs1025775474 Chr.17: 76684101-76684101 on Build GRCh38
rs760615448 Chr.17: 76683874-76683874 on Build GRCh38
rs536020993 Chr.17: 76684102-76684102 on Build GRCh38
rs373506079 Chr.17: 76683861-76683861 on Build GRCh38
rs763970523 Chr.17: 76683877-76683877 on Build GRCh38
rs183901071 Chr.17: 76683859-76683859 on Build GRCh38
rs939435255 Chr.17: 76684042-76684042 on Build GRCh38
rs939444108 Chr.17: 76684171-76684171 on Build GRCh38
rs775659083 Chr.17: 76684173-76684173 on Build GRCh38
rs73996672 Chr.17: 76684032-76684032 on Build GRCh38
rs759610519 Chr.17: 76684150-76684150 on Build GRCh38
rs1026437324 Chr.17: 76684169-76684169 on Build GRCh38
rs112400888 Chr.17: 76683994-76683994 on Build GRCh38
rs765583326 Chr.17: 76684158-76684158 on Build GRCh38
rs916568584 Chr.17: 76684182-76684182 on Build GRCh38
rs1042818553 Chr.17: 76684087-76684087 on Build GRCh38
rs189597500 Chr.17: 76684094-76684094 on Build GRCh38
rs75635193 Chr.17: 76684126-76684126 on Build GRCh38
rs768583274 Chr.17: 76683965-76683965 on Build GRCh38
rs778154298 Chr.17: 76683927-76683927 on Build GRCh38
rs946774423 Chr.17: 76683783-76683783 on Build GRCh38
rs918173449 Chr.17: 76684080-76684080 on Build GRCh38
rs756573832 Chr.17: 76683919-76683919 on Build GRCh38
rs1009100146 Chr.17: 76684108-76684108 on Build GRCh38
rs1034929 Chr.17: 76684041-76684041 on Build GRCh38
rs1007417179 Chr.17: 76684139-76684139 on Build GRCh38
rs772307542 Chr.17: 76683953-76683953 on Build GRCh38
rs1038346055 Chr.17: 76684234-76684234 on Build GRCh38
rs993169036 Chr.17: 76684162-76684162 on Build GRCh38
rs774475928 Chr.17: 76683956-76683957 on Build GRCh38
rs746072682 Chr.17: 76683946-76683946 on Build GRCh38
rs1018602745 Chr.17: 76684113-76684113 on Build GRCh38
rs543277932 Chr.17: 76683991-76683991 on Build GRCh38
rs375701830 Chr.17: 76683792-76683792 on Build GRCh38
rs1000026299 Chr.17: 76683916-76683916 on Build GRCh38
rs911548134 Chr.17: 76683996-76683996 on Build GRCh38
rs138127502 Chr.17: 76683918-76683918 on Build GRCh38
rs551804004 Chr.17: 76684040-76684040 on Build GRCh38
rs761245244 Chr.17: 76683885-76683885 on Build GRCh38
rs776140505 Chr.17: 76683881-76683881 on Build GRCh38
rs965958289 Chr.17: 76684161-76684161 on Build GRCh38
rs187642578 Chr.17: 76683935-76683935 on Build GRCh38
rs374101668 Chr.17: 76684220-76684220 on Build GRCh38
rs544256738 Chr.17: 76683785-76683785 on Build GRCh38
rs776420702 Chr.17: 76683969-76683969 on Build GRCh38
rs748773919 Chr.17: 76684001-76684001 on Build GRCh38
rs370041559 Chr.17: 76683906-76683906 on Build GRCh38
rs771825265 Chr.17: 76683864-76683864 on Build GRCh38
rs529503826 Chr.17: 76683957-76683957 on Build GRCh38
rs768560278 Chr.17: 76683801-76683801 on Build GRCh38
rs35684284 Chr.17: 76684033-76684034 on Build GRCh38
rs896464929 Chr.17: 76683793-76683793 on Build GRCh38
rs1056620167 Chr.17: 76684073-76684073 on Build GRCh38
rs558102556 Chr.17: 76683800-76683800 on Build GRCh38
rs1021000923 Chr.17: 76683839-76683839 on Build GRCh38
rs879651081 Chr.17: 76684208-76684208 on Build GRCh38
rs888923034 Chr.17: 76684127-76684127 on Build GRCh38
rs753031180 Chr.17: 76683913-76683913 on Build GRCh38
rs972141795 Chr.17: 76683992-76683992 on Build GRCh38
rs926395640 Chr.17: 76684235-76684235 on Build GRCh38
rs375660221 Chr.17: 76683866-76683866 on Build GRCh38
rs114504194 Chr.17: 76684038-76684038 on Build GRCh38
rs749330065 Chr.17: 76683856-76683856 on Build GRCh38
rs984971910 Chr.17: 76684039-76684039 on Build GRCh38
rs113035134 Chr.17: 76684086-76684086 on Build GRCh38
rs116096243 Chr.17: 76683928-76683928 on Build GRCh38
rs779006499 Chr.17: 76683941-76683941 on Build GRCh38
rs895361267 Chr.17: 76683817-76683817 on Build GRCh38
rs746785149 Chr.17: 76683964-76683964 on Build GRCh38
rs938197770 Chr.17: 76684061-76684061 on Build GRCh38
rs948025872 Chr.17: 76684208-76684209 on Build GRCh38
rs774539240 Chr.17: 76683860-76683860 on Build GRCh38
rs964653911 Chr.17: 76684009-76684009 on Build GRCh38
rs547465137 Chr.17: 76684092-76684092 on Build GRCh38
rs774314674 Chr.17: 76683945-76683945 on Build GRCh38

View Full Product Details