Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36139950-36140480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC3
Entrez Gene ID
27095
Gene Name
trafficking protein particle complex 3
Gene Aliases
BET3
UniGene
Hs.523131
Gene Chromosome Location
Chr.1: 36136569-36156053 on Build GRCh38
Transcripts
NM_001270894.1 NM_001270895.1 NM_001270896.1 NM_001270897.1 NM_014408.4 NR_073097.1 NR_073098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5159485 c.139delA p.M47fs*9 Chr.1: 36140070-36140070 on Build GRCh38
4932971 c.66C>G p.T22T Chr.1: 36140143-36140143 on Build GRCh38
7182323 c.103G>A p.E35K Chr.1: 36140106-36140106 on Build GRCh38
7182322 c.103G>A p.E35K Chr.1: 36140106-36140106 on Build GRCh38
4165177 c.110A>T p.D37V Chr.1: 36140099-36140099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553462390 Chr.1: 36140426-36140426 on Build GRCh38
rs776312501 Chr.1: 36140192-36140192 on Build GRCh38
rs915527755 Chr.1: 36140347-36140347 on Build GRCh38
rs762824143 Chr.1: 36140081-36140081 on Build GRCh38
rs979832751 Chr.1: 36140004-36140004 on Build GRCh38
rs574799798 Chr.1: 36140133-36140133 on Build GRCh38
rs998676222 Chr.1: 36140137-36140137 on Build GRCh38
rs913419027 Chr.1: 36140151-36140151 on Build GRCh38
rs926999125 Chr.1: 36140323-36140323 on Build GRCh38
rs369075133 Chr.1: 36140197-36140197 on Build GRCh38
rs764659729 Chr.1: 36140003-36140003 on Build GRCh38
rs145641137 Chr.1: 36140115-36140115 on Build GRCh38
rs752648678 Chr.1: 36140401-36140401 on Build GRCh38
rs757532491 Chr.1: 36140118-36140118 on Build GRCh38
rs148705774 Chr.1: 36140141-36140141 on Build GRCh38
rs369744066 Chr.1: 36140109-36140109 on Build GRCh38
rs375966178 Chr.1: 36140443-36140443 on Build GRCh38
rs201359010 Chr.1: 36140056-36140056 on Build GRCh38
rs775943264 Chr.1: 36140024-36140024 on Build GRCh38
rs759829777 Chr.1: 36140211-36140211 on Build GRCh38
rs560528823 Chr.1: 36139989-36139989 on Build GRCh38
rs375815667 Chr.1: 36140175-36140175 on Build GRCh38
rs2147448 Chr.1: 36140332-36140332 on Build GRCh38
rs749963001 Chr.1: 36140076-36140076 on Build GRCh38
rs957042442 Chr.1: 36140328-36140328 on Build GRCh38
rs764764352 Chr.1: 36140058-36140058 on Build GRCh38
rs889793699 Chr.1: 36140195-36140195 on Build GRCh38
rs751289379 Chr.1: 36140100-36140100 on Build GRCh38
rs551907286 Chr.1: 36139979-36139979 on Build GRCh38
rs74066051 Chr.1: 36140427-36140427 on Build GRCh38
rs151272539 Chr.1: 36140148-36140148 on Build GRCh38
rs1036304733 Chr.1: 36140417-36140417 on Build GRCh38
rs538141722 Chr.1: 36140099-36140099 on Build GRCh38
rs766300044 Chr.1: 36140218-36140218 on Build GRCh38
rs751223513 Chr.1: 36140303-36140303 on Build GRCh38
rs375229055 Chr.1: 36140384-36140384 on Build GRCh38
rs774949357 Chr.1: 36139977-36139977 on Build GRCh38
rs561050377 Chr.1: 36140009-36140009 on Build GRCh38
rs945613780 Chr.1: 36140397-36140397 on Build GRCh38
rs772495843 Chr.1: 36140329-36140329 on Build GRCh38
rs769455514 Chr.1: 36140206-36140206 on Build GRCh38
rs776207212 Chr.1: 36140052-36140052 on Build GRCh38
rs994048867 Chr.1: 36140276-36140276 on Build GRCh38
rs768953925 Chr.1: 36140188-36140188 on Build GRCh38
rs767326003 Chr.1: 36140388-36140388 on Build GRCh38
rs979854860 Chr.1: 36140311-36140311 on Build GRCh38
rs536473647 Chr.1: 36140437-36140437 on Build GRCh38
rs372007648 Chr.1: 36140210-36140210 on Build GRCh38
rs772791960 Chr.1: 36140042-36140042 on Build GRCh38
rs2231311 Chr.1: 36140001-36140001 on Build GRCh38
rs760956575 Chr.1: 36140025-36140025 on Build GRCh38
rs376110188 Chr.1: 36140186-36140186 on Build GRCh38
rs961571956 Chr.1: 36140232-36140232 on Build GRCh38
rs560456901 Chr.1: 36140343-36140343 on Build GRCh38
rs988908410 Chr.1: 36140117-36140117 on Build GRCh38
rs572469695 Chr.1: 36140448-36140448 on Build GRCh38
rs945591638 Chr.1: 36140113-36140113 on Build GRCh38
rs935620637 Chr.1: 36140019-36140019 on Build GRCh38
rs570910507 Chr.1: 36140295-36140295 on Build GRCh38
rs772644730 Chr.1: 36140173-36140173 on Build GRCh38

View Full Product Details