Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5394977-5395473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7198677 c.2616C>A p.I872I Chr.18: 5395097-5395097 on Build GRCh38
4876325 c.2579G>A p.G860D Chr.18: 5395134-5395134 on Build GRCh38
6083578 c.3127G>T p.G1043W Chr.18: 5395093-5395093 on Build GRCh38
4859128 c.2635G>A p.D879N Chr.18: 5395078-5395078 on Build GRCh38
989053 c.3073-1G>T p.? Chr.18: 5395148-5395148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368752856 Chr.18: 5395080-5395080 on Build GRCh38
rs749098606 Chr.18: 5395010-5395010 on Build GRCh38
rs1020291039 Chr.18: 5395067-5395067 on Build GRCh38
rs758086349 Chr.18: 5395363-5395363 on Build GRCh38
rs767310973 Chr.18: 5395199-5395199 on Build GRCh38
rs188833555 Chr.18: 5395430-5395430 on Build GRCh38
rs999714124 Chr.18: 5395196-5395196 on Build GRCh38
rs376613231 Chr.18: 5395159-5395159 on Build GRCh38
rs967743121 Chr.18: 5395081-5395081 on Build GRCh38
rs370686158 Chr.18: 5395061-5395061 on Build GRCh38
rs182897640 Chr.18: 5395426-5395426 on Build GRCh38
rs144676596 Chr.18: 5395094-5395094 on Build GRCh38
rs745342175 Chr.18: 5395004-5395004 on Build GRCh38
rs760696640 Chr.18: 5395055-5395055 on Build GRCh38
rs140104598 Chr.18: 5395249-5395249 on Build GRCh38
rs372063784 Chr.18: 5395173-5395173 on Build GRCh38
rs778610232 Chr.18: 5395127-5395127 on Build GRCh38
rs765902022 Chr.18: 5395427-5395427 on Build GRCh38
rs116042930 Chr.18: 5395252-5395252 on Build GRCh38
rs200752548 Chr.18: 5395045-5395045 on Build GRCh38
rs777928624 Chr.18: 5395155-5395155 on Build GRCh38
rs1031240788 Chr.18: 5395200-5395200 on Build GRCh38
rs909318269 Chr.18: 5395262-5395262 on Build GRCh38
rs774180947 Chr.18: 5395022-5395022 on Build GRCh38
rs966348627 Chr.18: 5395215-5395215 on Build GRCh38
rs566876033 Chr.18: 5395305-5395305 on Build GRCh38
rs903994499 Chr.18: 5395174-5395174 on Build GRCh38
rs552076207 Chr.18: 5395447-5395447 on Build GRCh38
rs750687392 Chr.18: 5395310-5395310 on Build GRCh38
rs767256031 Chr.18: 5395075-5395075 on Build GRCh38
rs1046538398 Chr.18: 5395184-5395184 on Build GRCh38
rs185170221 Chr.18: 5395026-5395026 on Build GRCh38
rs147930423 Chr.18: 5395095-5395095 on Build GRCh38
rs1000182197 Chr.18: 5395404-5395404 on Build GRCh38
rs368350627 Chr.18: 5395050-5395050 on Build GRCh38
rs548448511 Chr.18: 5395185-5395185 on Build GRCh38
rs570664288 Chr.18: 5395158-5395158 on Build GRCh38
rs140548684 Chr.18: 5395099-5395099 on Build GRCh38
rs756170568 Chr.18: 5395005-5395005 on Build GRCh38
rs780091318 Chr.18: 5395008-5395008 on Build GRCh38
rs369441193 Chr.18: 5395255-5395255 on Build GRCh38
rs929673630 Chr.18: 5395201-5395201 on Build GRCh38
rs754937234 Chr.18: 5395107-5395107 on Build GRCh38
rs1020827255 Chr.18: 5395321-5395321 on Build GRCh38
rs150071835 Chr.18: 5395116-5395116 on Build GRCh38
rs538615463 Chr.18: 5395334-5395334 on Build GRCh38
rs997923547 Chr.18: 5395244-5395244 on Build GRCh38
rs747949084 Chr.18: 5395137-5395137 on Build GRCh38
rs766905896 Chr.18: 5395051-5395051 on Build GRCh38
rs998282604 Chr.18: 5395315-5395315 on Build GRCh38
rs768277416 Chr.18: 5395018-5395018 on Build GRCh38
rs200047347 Chr.18: 5395160-5395160 on Build GRCh38
rs780992773 Chr.18: 5395192-5395192 on Build GRCh38
rs1039906740 Chr.18: 5395275-5395275 on Build GRCh38
rs765580895 Chr.18: 5395060-5395060 on Build GRCh38
rs1802388 Chr.18: 5395073-5395073 on Build GRCh38
rs763330611 Chr.18: 5395198-5395198 on Build GRCh38
rs7229903 Chr.18: 5395431-5395431 on Build GRCh38
rs754741995 Chr.18: 5395187-5395187 on Build GRCh38
rs145398752 Chr.18: 5395145-5395145 on Build GRCh38
rs1036884062 Chr.18: 5395052-5395052 on Build GRCh38
rs916216085 Chr.18: 5395438-5395438 on Build GRCh38
rs772304378 Chr.18: 5395041-5395041 on Build GRCh38

View Full Product Details