Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 69401189-69401690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK6
Entrez Gene ID
220164
Gene Name
docking protein 6
Gene Aliases
DOK5L, HsT3226
UniGene
Hs.278285
Gene Chromosome Location
Chr.18: 69401027-69849087 on Build GRCh38
Transcripts
NM_152721.5 XM_011525875.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4497588 c.49C>T p.R17C Chr.18: 69401293-69401293 on Build GRCh38
6149129 c.8C>G p.S3C Chr.18: 69401252-69401252 on Build GRCh38
5701892 c.8_9CC>TA p.S3L Chr.18: 69401252-69401253 on Build GRCh38
5670391 c.66+1G>T p.? Chr.18: 69401311-69401311 on Build GRCh38
4553082 c.57G>A p.R19R Chr.18: 69401301-69401301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932703865 Chr.18: 69401670-69401670 on Build GRCh38
rs997294549 Chr.18: 69401349-69401349 on Build GRCh38
rs762209004 Chr.18: 69401324-69401324 on Build GRCh38
rs533047144 Chr.18: 69401372-69401372 on Build GRCh38
rs779104236 Chr.18: 69401313-69401314 on Build GRCh38
rs868130286 Chr.18: 69401541-69401541 on Build GRCh38
rs993333014 Chr.18: 69401331-69401331 on Build GRCh38
rs369720344 Chr.18: 69401583-69401583 on Build GRCh38
rs765465397 Chr.18: 69401356-69401356 on Build GRCh38
rs568416381 Chr.18: 69401485-69401485 on Build GRCh38
rs1014874768 Chr.18: 69401521-69401521 on Build GRCh38
rs981133224 Chr.18: 69401335-69401335 on Build GRCh38
rs1035314836 Chr.18: 69401377-69401377 on Build GRCh38
rs944233404 Chr.18: 69401439-69401439 on Build GRCh38
rs148639698 Chr.18: 69401512-69401512 on Build GRCh38
rs951807704 Chr.18: 69401390-69401390 on Build GRCh38
rs11659170 Chr.18: 69401353-69401353 on Build GRCh38
rs1041131669 Chr.18: 69401480-69401480 on Build GRCh38
rs760777559 Chr.18: 69401215-69401215 on Build GRCh38
rs893415202 Chr.18: 69401252-69401252 on Build GRCh38
rs899813341 Chr.18: 69401343-69401343 on Build GRCh38
rs981198062 Chr.18: 69401373-69401373 on Build GRCh38
rs984396215 Chr.18: 69401429-69401429 on Build GRCh38
rs564058132 Chr.18: 69401241-69401241 on Build GRCh38
rs777659023 Chr.18: 69401245-69401245 on Build GRCh38
rs958393495 Chr.18: 69401375-69401375 on Build GRCh38
rs546562709 Chr.18: 69401224-69401224 on Build GRCh38
rs955904265 Chr.18: 69401378-69401378 on Build GRCh38
rs201119676 Chr.18: 69401321-69401321 on Build GRCh38
rs1035538977 Chr.18: 69401545-69401545 on Build GRCh38
rs996946230 Chr.18: 69401484-69401484 on Build GRCh38
rs574819480 Chr.18: 69401582-69401582 on Build GRCh38
rs1041829156 Chr.18: 69401319-69401319 on Build GRCh38
rs924768294 Chr.18: 69401431-69401431 on Build GRCh38
rs375656195 Chr.18: 69401334-69401334 on Build GRCh38
rs78889924 Chr.18: 69401553-69401553 on Build GRCh38
rs1048833148 Chr.18: 69401496-69401496 on Build GRCh38
rs914242477 Chr.18: 69401396-69401396 on Build GRCh38
rs947616796 Chr.18: 69401423-69401423 on Build GRCh38
rs546983461 Chr.18: 69401483-69401483 on Build GRCh38
rs528990882 Chr.18: 69401364-69401364 on Build GRCh38
rs539803963 Chr.18: 69401652-69401652 on Build GRCh38
rs1056050363 Chr.18: 69401243-69401243 on Build GRCh38
rs753919096 Chr.18: 69401210-69401210 on Build GRCh38
rs985484531 Chr.18: 69401367-69401367 on Build GRCh38
rs1026013130 Chr.18: 69401342-69401342 on Build GRCh38
rs534680590 Chr.18: 69401402-69401402 on Build GRCh38
rs911306592 Chr.18: 69401489-69401489 on Build GRCh38
rs201712085 Chr.18: 69401305-69401305 on Build GRCh38
rs766936694 Chr.18: 69401352-69401352 on Build GRCh38
rs540246608 Chr.18: 69401329-69401329 on Build GRCh38
rs1041572327 Chr.18: 69401635-69401635 on Build GRCh38
rs924904250 Chr.18: 69401597-69401597 on Build GRCh38
rs550453237 Chr.18: 69401370-69401370 on Build GRCh38
rs375390067 Chr.18: 69401525-69401525 on Build GRCh38
rs568001551 Chr.18: 69401632-69401662 on Build GRCh38
rs929987431 Chr.18: 69401542-69401542 on Build GRCh38
rs1022739397 Chr.18: 69401308-69401308 on Build GRCh38
rs755095040 Chr.18: 69401355-69401355 on Build GRCh38
rs540118413 Chr.18: 69401567-69401567 on Build GRCh38
rs1010124117 Chr.18: 69401422-69401422 on Build GRCh38
rs1021527752 Chr.18: 69401666-69401666 on Build GRCh38
rs977624141 Chr.18: 69401584-69401584 on Build GRCh38
rs944212398 Chr.18: 69401599-69401599 on Build GRCh38
rs991527840 Chr.18: 69401379-69401379 on Build GRCh38
rs1010337717 Chr.18: 69401665-69401665 on Build GRCh38
rs1005941675 Chr.18: 69401232-69401232 on Build GRCh38
rs761103309 Chr.18: 69401348-69401348 on Build GRCh38
rs921444701 Chr.18: 69401649-69401649 on Build GRCh38
rs375895200 Chr.18: 69401395-69401395 on Build GRCh38
rs775057063 Chr.18: 69401322-69401322 on Build GRCh38
rs747951101 Chr.18: 69401228-69401228 on Build GRCh38
rs773638016 Chr.18: 69401338-69401338 on Build GRCh38
rs755185103 Chr.18: 69401214-69401214 on Build GRCh38
rs764541066 Chr.18: 69401493-69401493 on Build GRCh38
rs117832633 Chr.18: 69401561-69401561 on Build GRCh38
rs779057964 Chr.18: 69401217-69401217 on Build GRCh38
rs951906400 Chr.18: 69401354-69401354 on Build GRCh38
rs33984456 Chr.18: 69401370-69401370 on Build GRCh38
rs1003156234 Chr.18: 69401544-69401544 on Build GRCh38
rs111462164 Chr.18: 69401374-69401374 on Build GRCh38
rs878974799 Chr.18: 69401369-69401370 on Build GRCh38
rs947220638 Chr.18: 69401366-69401366 on Build GRCh38
rs570103532 Chr.18: 69401371-69401371 on Build GRCh38
rs570490351 Chr.18: 69401376-69401376 on Build GRCh38
rs1002589222 Chr.18: 69401593-69401593 on Build GRCh38
rs771010944 Chr.18: 69401285-69401285 on Build GRCh38
rs746875087 Chr.18: 69401281-69401281 on Build GRCh38
rs113970522 Chr.18: 69401316-69401316 on Build GRCh38
rs1006111580 Chr.18: 69401518-69401518 on Build GRCh38
rs748407106 Chr.18: 69401358-69401374 on Build GRCh38
rs947647774 Chr.18: 69401275-69401275 on Build GRCh38

View Full Product Details