Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 76980326-76980866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBP
Entrez Gene ID
4155
Gene Name
myelin basic protein
Gene Aliases
-
UniGene
Hs.551713
Gene Chromosome Location
Chr.18: 76978833-77133708 on Build GRCh38
Transcripts
NM_001025081.1 NM_001025090.1 NM_001025092.1 NM_001025101.1 NM_002385.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133775 c.871-1G>T p.? Chr.18: 76980472-76980472 on Build GRCh38
990006 c.883A>G p.S295G Chr.18: 76980459-76980459 on Build GRCh38
6709212 c.589C>T p.R197C Chr.18: 76980432-76980432 on Build GRCh38
5900371 c.565C>T p.R189C Chr.18: 76980456-76980456 on Build GRCh38
5955832 c.912C>A p.R304R Chr.18: 76980430-76980430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72987247 Chr.18: 76980534-76980534 on Build GRCh38
rs757900784 Chr.18: 76980496-76980496 on Build GRCh38
rs902680479 Chr.18: 76980715-76980715 on Build GRCh38
rs753749041 Chr.18: 76980369-76980369 on Build GRCh38
rs112118220 Chr.18: 76980828-76980828 on Build GRCh38
rs528363202 Chr.18: 76980730-76980730 on Build GRCh38
rs921712379 Chr.18: 76980395-76980395 on Build GRCh38
rs373574609 Chr.18: 76980719-76980719 on Build GRCh38
rs907744218 Chr.18: 76980807-76980807 on Build GRCh38
rs1013689782 Chr.18: 76980726-76980726 on Build GRCh38
rs142115897 Chr.18: 76980815-76980815 on Build GRCh38
rs751156656 Chr.18: 76980500-76980500 on Build GRCh38
rs544919259 Chr.18: 76980750-76980750 on Build GRCh38
rs931551107 Chr.18: 76980678-76980678 on Build GRCh38
rs998555165 Chr.18: 76980659-76980659 on Build GRCh38
rs138972622 Chr.18: 76980603-76980603 on Build GRCh38
rs761965498 Chr.18: 76980454-76980454 on Build GRCh38
rs780429241 Chr.18: 76980504-76980504 on Build GRCh38
rs373454610 Chr.18: 76980377-76980377 on Build GRCh38
rs940394109 Chr.18: 76980651-76980651 on Build GRCh38
rs371134243 Chr.18: 76980386-76980386 on Build GRCh38
rs927665739 Chr.18: 76980545-76980545 on Build GRCh38
rs201862296 Chr.18: 76980699-76980699 on Build GRCh38
rs145141322 Chr.18: 76980410-76980410 on Build GRCh38
rs995705170 Chr.18: 76980508-76980508 on Build GRCh38
rs1011137815 Chr.18: 76980677-76980678 on Build GRCh38
rs553780605 Chr.18: 76980685-76980685 on Build GRCh38
rs934025803 Chr.18: 76980844-76980844 on Build GRCh38
rs974579177 Chr.18: 76980605-76980605 on Build GRCh38
rs958017251 Chr.18: 76980753-76980753 on Build GRCh38
rs112023970 Chr.18: 76980839-76980839 on Build GRCh38
rs564077793 Chr.18: 76980666-76980666 on Build GRCh38
rs201571232 Chr.18: 76980440-76980440 on Build GRCh38
rs748238952 Chr.18: 76980519-76980519 on Build GRCh38
rs760814414 Chr.18: 76980482-76980483 on Build GRCh38
rs999560054 Chr.18: 76980749-76980749 on Build GRCh38
rs148716184 Chr.18: 76980459-76980459 on Build GRCh38
rs768236319 Chr.18: 76980501-76980501 on Build GRCh38
rs373942491 Chr.18: 76980499-76980499 on Build GRCh38
rs773107996 Chr.18: 76980403-76980404 on Build GRCh38
rs890012065 Chr.18: 76980674-76980674 on Build GRCh38
rs766227267 Chr.18: 76980364-76980364 on Build GRCh38
rs1024988359 Chr.18: 76980570-76980570 on Build GRCh38
rs984694702 Chr.18: 76980760-76980760 on Build GRCh38
rs544017750 Chr.18: 76980415-76980415 on Build GRCh38
rs778445201 Chr.18: 76980379-76980379 on Build GRCh38
rs78091324 Chr.18: 76980606-76980606 on Build GRCh38
rs542371917 Chr.18: 76980706-76980706 on Build GRCh38
rs749588391 Chr.18: 76980509-76980509 on Build GRCh38
rs752261542 Chr.18: 76980495-76980495 on Build GRCh38
rs760556766 Chr.18: 76980462-76980462 on Build GRCh38
rs376676611 Chr.18: 76980471-76980471 on Build GRCh38
rs533561240 Chr.18: 76980679-76980679 on Build GRCh38
rs1034690174 Chr.18: 76980725-76980725 on Build GRCh38
rs187126799 Chr.18: 76980571-76980571 on Build GRCh38
rs776402815 Chr.18: 76980567-76980567 on Build GRCh38
rs978432167 Chr.18: 76980580-76980580 on Build GRCh38
rs758095224 Chr.18: 76980385-76980385 on Build GRCh38
rs772485459 Chr.18: 76980455-76980455 on Build GRCh38
rs575940831 Chr.18: 76980741-76980741 on Build GRCh38
rs9199 Chr.18: 76980380-76980380 on Build GRCh38
rs901213921 Chr.18: 76980645-76980645 on Build GRCh38
rs547064265 Chr.18: 76980527-76980527 on Build GRCh38
rs749756430 Chr.18: 76980432-76980432 on Build GRCh38
rs987154784 Chr.18: 76980628-76980628 on Build GRCh38
rs907553076 Chr.18: 76980634-76980634 on Build GRCh38
rs548927106 Chr.18: 76980589-76980589 on Build GRCh38
rs1037761698 Chr.18: 76980660-76980660 on Build GRCh38
rs750295461 Chr.18: 76980357-76980357 on Build GRCh38
rs998482397 Chr.18: 76980399-76980399 on Build GRCh38
rs1037111283 Chr.18: 76980632-76980632 on Build GRCh38
rs144660620 Chr.18: 76980470-76980470 on Build GRCh38
rs999268205 Chr.18: 76980723-76980723 on Build GRCh38
rs760538407 Chr.18: 76980363-76980363 on Build GRCh38
rs191984186 Chr.18: 76980623-76980623 on Build GRCh38
rs1051415090 Chr.18: 76980431-76980431 on Build GRCh38
rs960357950 Chr.18: 76980549-76980549 on Build GRCh38
rs895281577 Chr.18: 76980688-76980688 on Build GRCh38
rs371262081 Chr.18: 76980480-76980480 on Build GRCh38
rs765062745 Chr.18: 76980491-76980491 on Build GRCh38
rs537726560 Chr.18: 76980604-76980604 on Build GRCh38
rs770077639 Chr.18: 76980405-76980405 on Build GRCh38
rs779279135 Chr.18: 76980511-76980511 on Build GRCh38
rs774533624 Chr.18: 76980442-76980442 on Build GRCh38
rs1028799283 Chr.18: 76980784-76980784 on Build GRCh38
rs113673334 Chr.18: 76980473-76980473 on Build GRCh38
rs996008617 Chr.18: 76980779-76980779 on Build GRCh38

View Full Product Details