Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1272987-1273482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIRBP
Entrez Gene ID
1153
Gene Name
cold inducible RNA binding protein
Gene Aliases
CIRP
UniGene
Hs.618145
Gene Chromosome Location
Chr.19: 1269266-1274810 on Build GRCh38
Transcripts
NM_001280.2 NM_001300815.1 NM_001300829.1 NR_023312.2 NR_023313.2 XM_006722637.1 XM_011527668.1 XM_017026237.1

SNP Detail

SNP ID Chromosome Location
rs988413257 Chr.19: 1273069-1273069 on Build GRCh38
rs534700548 Chr.19: 1273268-1273268 on Build GRCh38
rs543498474 Chr.19: 1273411-1273411 on Build GRCh38
rs995598869 Chr.19: 1273125-1273125 on Build GRCh38
rs180889146 Chr.19: 1273457-1273457 on Build GRCh38
rs925415043 Chr.19: 1273095-1273095 on Build GRCh38
rs570295493 Chr.19: 1273122-1273122 on Build GRCh38
rs1049572741 Chr.19: 1273352-1273352 on Build GRCh38
rs553230465 Chr.19: 1273269-1273269 on Build GRCh38
rs1009616676 Chr.19: 1273412-1273412 on Build GRCh38
rs907836925 Chr.19: 1273297-1273297 on Build GRCh38
rs966416798 Chr.19: 1273242-1273242 on Build GRCh38
rs575448772 Chr.19: 1273363-1273363 on Build GRCh38
rs1023743978 Chr.19: 1273421-1273421 on Build GRCh38
rs761520102 Chr.19: 1273116-1273116 on Build GRCh38
rs1032598637 Chr.19: 1273244-1273247 on Build GRCh38
rs533473961 Chr.19: 1273015-1273015 on Build GRCh38
rs1028092385 Chr.19: 1273025-1273025 on Build GRCh38
rs762768732 Chr.19: 1273180-1273180 on Build GRCh38
rs944239896 Chr.19: 1273350-1273350 on Build GRCh38
rs970444922 Chr.19: 1273427-1273427 on Build GRCh38
rs1032966798 Chr.19: 1273388-1273388 on Build GRCh38
rs533031113 Chr.19: 1273315-1273315 on Build GRCh38
rs949452462 Chr.19: 1273285-1273285 on Build GRCh38
rs753974324 Chr.19: 1273358-1273358 on Build GRCh38
rs544746239 Chr.19: 1273118-1273118 on Build GRCh38
rs970559672 Chr.19: 1273260-1273260 on Build GRCh38
rs981978444 Chr.19: 1273428-1273428 on Build GRCh38
rs1019838107 Chr.19: 1273209-1273209 on Build GRCh38
rs140842163 Chr.19: 1273243-1273244 on Build GRCh38
rs997116216 Chr.19: 1273377-1273377 on Build GRCh38
rs753479520 Chr.19: 1273283-1273283 on Build GRCh38
rs538800117 Chr.19: 1273314-1273314 on Build GRCh38
rs898407320 Chr.19: 1273324-1273324 on Build GRCh38
rs555722524 Chr.19: 1273447-1273447 on Build GRCh38
rs995633555 Chr.19: 1273006-1273006 on Build GRCh38
rs1046378714 Chr.19: 1273146-1273146 on Build GRCh38
rs953662094 Chr.19: 1273062-1273062 on Build GRCh38
rs973348441 Chr.19: 1273448-1273448 on Build GRCh38
rs913742086 Chr.19: 1273071-1273071 on Build GRCh38
rs990825075 Chr.19: 1273111-1273111 on Build GRCh38
rs957906432 Chr.19: 1273237-1273237 on Build GRCh38
rs189305570 Chr.19: 1273219-1273219 on Build GRCh38
rs995339547 Chr.19: 1273173-1273173 on Build GRCh38
rs766196411 Chr.19: 1273231-1273231 on Build GRCh38
rs887164561 Chr.19: 1273349-1273349 on Build GRCh38
rs757494114 Chr.19: 1273420-1273420 on Build GRCh38
rs562490573 Chr.19: 1273160-1273160 on Build GRCh38
rs756138840 Chr.19: 1273307-1273307 on Build GRCh38
rs957808713 Chr.19: 1273048-1273048 on Build GRCh38
rs751568903 Chr.19: 1273251-1273251 on Build GRCh38
rs1032456813 Chr.19: 1273405-1273405 on Build GRCh38
rs975020665 Chr.19: 1273012-1273012 on Build GRCh38
rs1021098775 Chr.19: 1273255-1273255 on Build GRCh38
rs919947952 Chr.19: 1273303-1273303 on Build GRCh38
rs370375138 Chr.19: 1273019-1273019 on Build GRCh38
rs959160843 Chr.19: 1273432-1273432 on Build GRCh38
rs143564147 Chr.19: 1273042-1273042 on Build GRCh38
rs931259623 Chr.19: 1273346-1273346 on Build GRCh38
rs893489350 Chr.19: 1273394-1273394 on Build GRCh38
rs1033506716 Chr.19: 1273429-1273429 on Build GRCh38
rs1047162642 Chr.19: 1273326-1273326 on Build GRCh38
rs889653645 Chr.19: 1273354-1273354 on Build GRCh38
rs377699492 Chr.19: 1273288-1273288 on Build GRCh38
rs199651756 Chr.19: 1273267-1273267 on Build GRCh38
rs772943370 Chr.19: 1273161-1273161 on Build GRCh38
rs1007953496 Chr.19: 1273356-1273356 on Build GRCh38
rs1046363601 Chr.19: 1273081-1273081 on Build GRCh38
rs1028196914 Chr.19: 1273208-1273208 on Build GRCh38
rs886463857 Chr.19: 1273157-1273157 on Build GRCh38
rs768262250 Chr.19: 1273075-1273075 on Build GRCh38
rs776133146 Chr.19: 1273100-1273100 on Build GRCh38
rs926495963 Chr.19: 1273087-1273087 on Build GRCh38
rs939918320 Chr.19: 1273159-1273159 on Build GRCh38
rs374014782 Chr.19: 1273038-1273038 on Build GRCh38
rs898616423 Chr.19: 1273123-1273123 on Build GRCh38
rs147165041 Chr.19: 1273189-1273189 on Build GRCh38
rs900127638 Chr.19: 1273359-1273359 on Build GRCh38
rs1803282 Chr.19: 1273080-1273080 on Build GRCh38
rs996508377 Chr.19: 1273230-1273230 on Build GRCh38
rs1008327835 Chr.19: 1273195-1273195 on Build GRCh38

View Full Product Details