Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40807729-40808223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGLN2
Entrez Gene ID
112398
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
UniGene
Hs.515417
Gene Chromosome Location
Chr.19: 40799143-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Gene Symbol
RAB4B-EGLN2
Entrez Gene ID
100529264
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
Gene Aliases
RERT-lncRNA
UniGene
Hs.631539
Gene Chromosome Location
Chr.19: 40778219-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1393898 c.1199T>C p.V400A Chr.19: 40807839-40807839 on Build GRCh38
1205058 c.1208C>A p.P403Q Chr.19: 40807848-40807848 on Build GRCh38
996836 c.1214C>T p.T405M Chr.19: 40807854-40807854 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906864411 Chr.19: 40808163-40808163 on Build GRCh38
rs1004562975 Chr.19: 40807930-40807930 on Build GRCh38
rs982733205 Chr.19: 40808078-40808078 on Build GRCh38
rs867216902 Chr.19: 40807831-40807831 on Build GRCh38
rs759650902 Chr.19: 40807796-40807796 on Build GRCh38
rs546812287 Chr.19: 40808145-40808145 on Build GRCh38
rs369625239 Chr.19: 40807785-40807785 on Build GRCh38
rs757656607 Chr.19: 40807796-40807797 on Build GRCh38
rs374000050 Chr.19: 40807882-40807882 on Build GRCh38
rs747824587 Chr.19: 40807867-40807867 on Build GRCh38
rs769400397 Chr.19: 40807873-40807873 on Build GRCh38
rs920211641 Chr.19: 40807824-40807824 on Build GRCh38
rs906834115 Chr.19: 40807978-40807978 on Build GRCh38
rs1020105853 Chr.19: 40808083-40808083 on Build GRCh38
rs796455436 Chr.19: 40808073-40808074 on Build GRCh38
rs966060484 Chr.19: 40808095-40808095 on Build GRCh38
rs761239810 Chr.19: 40808189-40808189 on Build GRCh38
rs377024627 Chr.19: 40807793-40807793 on Build GRCh38
rs756595762 Chr.19: 40807809-40807809 on Build GRCh38
rs749027019 Chr.19: 40807881-40807881 on Build GRCh38
rs746413418 Chr.19: 40808121-40808121 on Build GRCh38
rs764146764 Chr.19: 40807768-40807770 on Build GRCh38
rs551227905 Chr.19: 40808047-40808047 on Build GRCh38
rs772998542 Chr.19: 40807874-40807874 on Build GRCh38
rs946298805 Chr.19: 40808160-40808160 on Build GRCh38
rs759575898 Chr.19: 40807915-40807915 on Build GRCh38
rs533067495 Chr.19: 40808012-40808012 on Build GRCh38
rs761746320 Chr.19: 40808073-40808074 on Build GRCh38
rs62108990 Chr.19: 40807919-40807919 on Build GRCh38
rs967217590 Chr.19: 40808084-40808084 on Build GRCh38
rs767700388 Chr.19: 40807800-40807800 on Build GRCh38
rs780859020 Chr.19: 40807861-40807861 on Build GRCh38
rs779415979 Chr.19: 40807848-40807848 on Build GRCh38
rs995328439 Chr.19: 40808067-40808067 on Build GRCh38
rs1041975014 Chr.19: 40808161-40808161 on Build GRCh38
rs868685290 Chr.19: 40808016-40808016 on Build GRCh38
rs141698095 Chr.19: 40807854-40807854 on Build GRCh38
rs573597851 Chr.19: 40807950-40807950 on Build GRCh38
rs775403804 Chr.19: 40808022-40808022 on Build GRCh38
rs202046460 Chr.19: 40807828-40807828 on Build GRCh38
rs562298934 Chr.19: 40807996-40807996 on Build GRCh38
rs1011397487 Chr.19: 40808039-40808039 on Build GRCh38
rs769543429 Chr.19: 40807784-40807784 on Build GRCh38
rs751674826 Chr.19: 40807782-40807785 on Build GRCh38
rs568530112 Chr.19: 40808194-40808194 on Build GRCh38
rs1001610245 Chr.19: 40808005-40808005 on Build GRCh38
rs748000680 Chr.19: 40807780-40807780 on Build GRCh38
rs544193492 Chr.19: 40807951-40807951 on Build GRCh38
rs1045468086 Chr.19: 40807963-40807963 on Build GRCh38
rs376964412 Chr.19: 40807763-40807763 on Build GRCh38
rs551912605 Chr.19: 40808096-40808096 on Build GRCh38
rs770467475 Chr.19: 40808185-40808185 on Build GRCh38
rs914758331 Chr.19: 40808134-40808134 on Build GRCh38
rs781739072 Chr.19: 40807802-40807802 on Build GRCh38
rs774342029 Chr.19: 40807907-40807907 on Build GRCh38
rs181478340 Chr.19: 40808114-40808114 on Build GRCh38
rs754503213 Chr.19: 40807855-40807855 on Build GRCh38
rs201243991 Chr.19: 40808074-40808075 on Build GRCh38
rs774310360 Chr.19: 40807794-40807794 on Build GRCh38
rs377069095 Chr.19: 40807795-40807795 on Build GRCh38
rs772947190 Chr.19: 40807789-40807789 on Build GRCh38
rs1002558940 Chr.19: 40808029-40808029 on Build GRCh38
rs369273747 Chr.19: 40808111-40808112 on Build GRCh38
rs946308202 Chr.19: 40807959-40807959 on Build GRCh38
rs370322331 Chr.19: 40807844-40807844 on Build GRCh38
rs371429105 Chr.19: 40808130-40808130 on Build GRCh38
rs559937401 Chr.19: 40808107-40808107 on Build GRCh38
rs764574945 Chr.19: 40807825-40807825 on Build GRCh38
rs749135378 Chr.19: 40808025-40808025 on Build GRCh38

View Full Product Details