Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44710331-44710824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM16
Entrez Gene ID
388551
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Gene Aliases
CEAL2, DFNA4B
UniGene
Hs.456381
Gene Chromosome Location
Chr.19: 44699151-44710714 on Build GRCh38
Transcripts
NM_001039213.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4855869 c.1470G>T p.G490G Chr.19: 44710503-44710503 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749073842 Chr.19: 44710715-44710715 on Build GRCh38
rs77791595 Chr.19: 44710408-44710408 on Build GRCh38
rs369172258 Chr.19: 44710596-44710596 on Build GRCh38
rs888974890 Chr.19: 44710417-44710417 on Build GRCh38
rs755012844 Chr.19: 44710756-44710756 on Build GRCh38
rs986296424 Chr.19: 44710590-44710590 on Build GRCh38
rs143656680 Chr.19: 44710398-44710398 on Build GRCh38
rs375895764 Chr.19: 44710511-44710511 on Build GRCh38
rs368670088 Chr.19: 44710477-44710477 on Build GRCh38
rs775671781 Chr.19: 44710519-44710519 on Build GRCh38
rs746231278 Chr.19: 44710533-44710535 on Build GRCh38
rs1040267459 Chr.19: 44710660-44710660 on Build GRCh38
rs752427705 Chr.19: 44710550-44710550 on Build GRCh38
rs1051276695 Chr.19: 44710698-44710698 on Build GRCh38
rs757379447 Chr.19: 44710788-44710790 on Build GRCh38
rs766219029 Chr.19: 44710545-44710545 on Build GRCh38
rs755657554 Chr.19: 44710551-44710551 on Build GRCh38
rs780961627 Chr.19: 44710472-44710472 on Build GRCh38
rs527795648 Chr.19: 44710525-44710525 on Build GRCh38
rs761936990 Chr.19: 44710478-44710478 on Build GRCh38
rs751154314 Chr.19: 44710546-44710546 on Build GRCh38
rs914904079 Chr.19: 44710394-44710394 on Build GRCh38
rs765114269 Chr.19: 44710524-44710524 on Build GRCh38
rs199656542 Chr.19: 44710510-44710510 on Build GRCh38
rs796138165 Chr.19: 44710788-44710788 on Build GRCh38
rs57882224 Chr.19: 44710786-44710786 on Build GRCh38
rs897713350 Chr.19: 44710460-44710460 on Build GRCh38
rs758200407 Chr.19: 44710462-44710462 on Build GRCh38
rs754222528 Chr.19: 44710787-44710787 on Build GRCh38
rs561412468 Chr.19: 44710589-44710589 on Build GRCh38
rs146757817 Chr.19: 44710506-44710506 on Build GRCh38
rs1052015664 Chr.19: 44710369-44710369 on Build GRCh38
rs930216872 Chr.19: 44710493-44710493 on Build GRCh38
rs779834338 Chr.19: 44710622-44710622 on Build GRCh38
rs529177756 Chr.19: 44710786-44710786 on Build GRCh38
rs969203097 Chr.19: 44710641-44710641 on Build GRCh38
rs763002767 Chr.19: 44710534-44710534 on Build GRCh38
rs972238524 Chr.19: 44710669-44710669 on Build GRCh38
rs867407868 Chr.19: 44710682-44710682 on Build GRCh38
rs560993240 Chr.19: 44710761-44710761 on Build GRCh38
rs980640418 Chr.19: 44710657-44710657 on Build GRCh38
rs1012897827 Chr.19: 44710770-44710770 on Build GRCh38
rs959785442 Chr.19: 44710742-44710742 on Build GRCh38
rs777355717 Chr.19: 44710569-44710569 on Build GRCh38
rs763388555 Chr.19: 44710786-44710787 on Build GRCh38
rs750399818 Chr.19: 44710453-44710453 on Build GRCh38
rs1024925180 Chr.19: 44710624-44710626 on Build GRCh38
rs1040319650 Chr.19: 44710434-44710434 on Build GRCh38
rs904007881 Chr.19: 44710754-44710754 on Build GRCh38
rs909041979 Chr.19: 44710658-44710658 on Build GRCh38
rs929232014 Chr.19: 44710686-44710686 on Build GRCh38
rs765140094 Chr.19: 44710447-44710447 on Build GRCh38
rs781675289 Chr.19: 44710511-44710511 on Build GRCh38
rs779626566 Chr.19: 44710467-44710467 on Build GRCh38
rs1050162329 Chr.19: 44710410-44710410 on Build GRCh38
rs897662446 Chr.19: 44710665-44710665 on Build GRCh38
rs907249729 Chr.19: 44710774-44710774 on Build GRCh38
rs574525847 Chr.19: 44710795-44710795 on Build GRCh38
rs780798424 Chr.19: 44710549-44710549 on Build GRCh38
rs374135385 Chr.19: 44710469-44710469 on Build GRCh38
rs1048958791 Chr.19: 44710691-44710691 on Build GRCh38
rs910264024 Chr.19: 44710595-44710595 on Build GRCh38
rs754131979 Chr.19: 44710461-44710461 on Build GRCh38
rs57684876 Chr.19: 44710429-44710429 on Build GRCh38
rs918908054 Chr.19: 44710670-44710670 on Build GRCh38
rs62120572 Chr.19: 44710480-44710480 on Build GRCh38
rs754741174 Chr.19: 44710470-44710470 on Build GRCh38
rs528890286 Chr.19: 44710638-44710638 on Build GRCh38
rs200994313 Chr.19: 44710788-44710789 on Build GRCh38
rs547345424 Chr.19: 44710692-44710692 on Build GRCh38
rs956187507 Chr.19: 44710794-44710794 on Build GRCh38
rs760762646 Chr.19: 44710521-44710521 on Build GRCh38
rs971989791 Chr.19: 44710503-44710503 on Build GRCh38
rs540979481 Chr.19: 44710709-44710709 on Build GRCh38
rs369749061 Chr.19: 44710757-44710757 on Build GRCh38
rs946388747 Chr.19: 44710568-44710568 on Build GRCh38
rs1039837603 Chr.19: 44710724-44710724 on Build GRCh38
rs143934888 Chr.19: 44710483-44710483 on Build GRCh38
rs1030971713 Chr.19: 44710557-44710557 on Build GRCh38
rs374369312 Chr.19: 44710785-44710785 on Build GRCh38
rs772325768 Chr.19: 44710513-44710513 on Build GRCh38
rs771286099 Chr.19: 44710488-44710488 on Build GRCh38
rs549041281 Chr.19: 44710793-44710793 on Build GRCh38
rs1023472556 Chr.19: 44710773-44710773 on Build GRCh38

View Full Product Details