Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46493734-46494234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNMAL2
Entrez Gene ID
57469
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
-
UniGene
Hs.7193
Gene Chromosome Location
Chr.19: 46491191-46495912 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Gene Symbol
PPP5D1
Entrez Gene ID
100506012
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
Gene Aliases
-
UniGene
Hs.671661
Gene Chromosome Location
Chr.19: 46480788-46601200 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5604553 c.1597C>T p.P533S Chr.19: 46493869-46493869 on Build GRCh38
4716196 c.1466C>T p.A489V Chr.19: 46494000-46494000 on Build GRCh38
4716197 c.1413C>A p.A471A Chr.19: 46494053-46494053 on Build GRCh38
3105360 c.1477G>A p.A493T Chr.19: 46493989-46493989 on Build GRCh38
4716194 c.1706_1707insAGGCCG p.G570_V571insRG Chr.19: 46493759-46493760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753108325 Chr.19: 46494100-46494100 on Build GRCh38
rs370754573 Chr.19: 46494127-46494127 on Build GRCh38
rs753524991 Chr.19: 46493944-46493944 on Build GRCh38
rs184536656 Chr.19: 46494046-46494046 on Build GRCh38
rs764329002 Chr.19: 46494161-46494161 on Build GRCh38
rs370168747 Chr.19: 46493850-46493850 on Build GRCh38
rs201500682 Chr.19: 46494140-46494140 on Build GRCh38
rs773732551 Chr.19: 46493776-46493777 on Build GRCh38
rs982195629 Chr.19: 46494005-46494005 on Build GRCh38
rs778069315 Chr.19: 46494104-46494104 on Build GRCh38
rs778358222 Chr.19: 46493875-46493875 on Build GRCh38
rs748793703 Chr.19: 46493781-46493781 on Build GRCh38
rs955053105 Chr.19: 46493911-46493911 on Build GRCh38
rs570360507 Chr.19: 46494152-46494152 on Build GRCh38
rs774063939 Chr.19: 46494203-46494203 on Build GRCh38
rs757819278 Chr.19: 46493783-46493783 on Build GRCh38
rs887299416 Chr.19: 46493887-46493887 on Build GRCh38
rs543308728 Chr.19: 46493922-46493922 on Build GRCh38
rs376968267 Chr.19: 46494038-46494038 on Build GRCh38
rs779093120 Chr.19: 46494178-46494178 on Build GRCh38
rs767267019 Chr.19: 46493849-46493849 on Build GRCh38
rs755790334 Chr.19: 46493858-46493858 on Build GRCh38
rs760569792 Chr.19: 46494047-46494047 on Build GRCh38
rs757400859 Chr.19: 46494171-46494171 on Build GRCh38
rs779409662 Chr.19: 46493803-46493803 on Build GRCh38
rs755398353 Chr.19: 46493994-46493994 on Build GRCh38
rs372657510 Chr.19: 46494054-46494054 on Build GRCh38
rs986557996 Chr.19: 46493755-46493756 on Build GRCh38
rs776390928 Chr.19: 46494049-46494049 on Build GRCh38
rs534649722 Chr.19: 46493776-46493776 on Build GRCh38
rs775208262 Chr.19: 46493925-46493925 on Build GRCh38
rs748008910 Chr.19: 46494189-46494189 on Build GRCh38
rs747244251 Chr.19: 46494190-46494190 on Build GRCh38
rs748402323 Chr.19: 46494001-46494001 on Build GRCh38
rs370979395 Chr.19: 46494134-46494134 on Build GRCh38
rs772922156 Chr.19: 46494141-46494141 on Build GRCh38
rs765934844 Chr.19: 46494149-46494149 on Build GRCh38
rs913317636 Chr.19: 46493798-46493798 on Build GRCh38
rs772103004 Chr.19: 46493775-46493776 on Build GRCh38
rs765740558 Chr.19: 46494069-46494069 on Build GRCh38
rs927875573 Chr.19: 46493773-46493773 on Build GRCh38
rs576671060 Chr.19: 46493942-46493942 on Build GRCh38
rs778812819 Chr.19: 46494125-46494125 on Build GRCh38
rs946544571 Chr.19: 46494066-46494066 on Build GRCh38
rs767274682 Chr.19: 46493770-46493770 on Build GRCh38
rs768127021 Chr.19: 46494159-46494159 on Build GRCh38
rs1045803981 Chr.19: 46493877-46493877 on Build GRCh38
rs748314343 Chr.19: 46493902-46493902 on Build GRCh38
rs187904708 Chr.19: 46493903-46493903 on Build GRCh38
rs751458508 Chr.19: 46493758-46493758 on Build GRCh38
rs762473728 Chr.19: 46494146-46494146 on Build GRCh38
rs748233775 Chr.19: 46493760-46493765 on Build GRCh38
rs545570021 Chr.19: 46493946-46493946 on Build GRCh38
rs781654636 Chr.19: 46494000-46494000 on Build GRCh38
rs377681319 Chr.19: 46494133-46494133 on Build GRCh38
rs771077994 Chr.19: 46494115-46494115 on Build GRCh38
rs749955547 Chr.19: 46494062-46494062 on Build GRCh38
rs199644087 Chr.19: 46494202-46494202 on Build GRCh38
rs749915322 Chr.19: 46493956-46493956 on Build GRCh38
rs756464861 Chr.19: 46494103-46494103 on Build GRCh38
rs925172549 Chr.19: 46493795-46493795 on Build GRCh38
rs551063520 Chr.19: 46493894-46493894 on Build GRCh38
rs369049949 Chr.19: 46494042-46494042 on Build GRCh38
rs758890399 Chr.19: 46493961-46493961 on Build GRCh38
rs768964744 Chr.19: 46493825-46493825 on Build GRCh38
rs761221819 Chr.19: 46494125-46494126 on Build GRCh38
rs747241096 Chr.19: 46493823-46493823 on Build GRCh38
rs753198145 Chr.19: 46494160-46494160 on Build GRCh38
rs746386665 Chr.19: 46493876-46493876 on Build GRCh38
rs368708147 Chr.19: 46494196-46494196 on Build GRCh38
rs752138437 Chr.19: 46494077-46494077 on Build GRCh38
rs751952034 Chr.19: 46493967-46493967 on Build GRCh38
rs766409853 Chr.19: 46493755-46493755 on Build GRCh38
rs746322356 Chr.19: 46493805-46493805 on Build GRCh38
rs969044325 Chr.19: 46493954-46493954 on Build GRCh38
rs759475429 Chr.19: 46493847-46493847 on Build GRCh38
rs988007770 Chr.19: 46493793-46493793 on Build GRCh38
rs901428062 Chr.19: 46494180-46494180 on Build GRCh38
rs777005893 Chr.19: 46493899-46493899 on Build GRCh38
rs373309758 Chr.19: 46493974-46493974 on Build GRCh38
rs764774191 Chr.19: 46493953-46493953 on Build GRCh38
rs776913670 Chr.19: 46493829-46493829 on Build GRCh38
rs763723651 Chr.19: 46493866-46493866 on Build GRCh38
rs749404557 Chr.19: 46494108-46494108 on Build GRCh38
rs762534117 Chr.19: 46494067-46494067 on Build GRCh38
rs554234767 Chr.19: 46493867-46493867 on Build GRCh38
rs761407668 Chr.19: 46493949-46493949 on Build GRCh38
rs930250478 Chr.19: 46494191-46494191 on Build GRCh38
rs772357024 Chr.19: 46493817-46493817 on Build GRCh38
rs780592952 Chr.19: 46493962-46493962 on Build GRCh38
rs765349767 Chr.19: 46493838-46493838 on Build GRCh38
rs888176334 Chr.19: 46493927-46493927 on Build GRCh38
rs749375279 Chr.19: 46494041-46494041 on Build GRCh38
rs745940906 Chr.19: 46494183-46494183 on Build GRCh38
rs758913922 Chr.19: 46493885-46493885 on Build GRCh38
rs913751532 Chr.19: 46493759-46493759 on Build GRCh38
rs756768436 Chr.19: 46493869-46493869 on Build GRCh38
rs144644044 Chr.19: 46494128-46494128 on Build GRCh38
rs889764091 Chr.19: 46493842-46493842 on Build GRCh38
rs777306940 Chr.19: 46493778-46493778 on Build GRCh38
rs773291456 Chr.19: 46493906-46493906 on Build GRCh38
rs775872838 Chr.19: 46493821-46493821 on Build GRCh38
rs969475898 Chr.19: 46493761-46493761 on Build GRCh38
rs771437872 Chr.19: 46494156-46494156 on Build GRCh38
rs147338403 Chr.19: 46493759-46493760 on Build GRCh38
rs761971396 Chr.19: 46493834-46493834 on Build GRCh38
rs781723065 Chr.19: 46494088-46494088 on Build GRCh38
rs199560010 Chr.19: 46494048-46494048 on Build GRCh38
rs201060790 Chr.19: 46493892-46493892 on Build GRCh38
rs375434413 Chr.19: 46494187-46494187 on Build GRCh38
rs764935818 Chr.19: 46494055-46494055 on Build GRCh38
rs754116968 Chr.19: 46494170-46494170 on Build GRCh38
rs373911812 Chr.19: 46493935-46493935 on Build GRCh38
rs558385656 Chr.19: 46493819-46493819 on Build GRCh38
rs898717014 Chr.19: 46493896-46493896 on Build GRCh38
rs565071242 Chr.19: 46494009-46494009 on Build GRCh38
rs754742737 Chr.19: 46493760-46493760 on Build GRCh38
rs759433142 Chr.19: 46493912-46493912 on Build GRCh38
rs759132492 Chr.19: 46494210-46494210 on Build GRCh38
rs1055683527 Chr.19: 46493808-46493808 on Build GRCh38
rs193045013 Chr.19: 46493933-46493933 on Build GRCh38
rs1025806293 Chr.19: 46493917-46493917 on Build GRCh38
rs377156204 Chr.19: 46494080-46494080 on Build GRCh38
rs1000269308 Chr.19: 46494078-46494078 on Build GRCh38
rs769054369 Chr.19: 46493898-46493898 on Build GRCh38
rs760188756 Chr.19: 46494158-46494158 on Build GRCh38
rs369389159 Chr.19: 46493839-46493839 on Build GRCh38

View Full Product Details