Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48596647-48597139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT2B1
Entrez Gene ID
6820
Gene Name
sulfotransferase family 2B member 1
Gene Aliases
HSST2
UniGene
Hs.369331
Gene Chromosome Location
Chr.19: 48552172-48599427 on Build GRCh38
Transcripts
NM_004605.2 NM_177973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999022 c.657C>A p.G219G Chr.19: 48596750-48596750 on Build GRCh38
6085188 c.715T>G p.S239A Chr.19: 48596808-48596808 on Build GRCh38
6842167 c.753delC p.H252fs*>99 Chr.19: 48596891-48596891 on Build GRCh38
1240343 c.772C>A p.L258I Chr.19: 48596910-48596910 on Build GRCh38
3107566 c.742G>A p.A248T Chr.19: 48596835-48596835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779491595 Chr.19: 48596860-48596860 on Build GRCh38
rs199827788 Chr.19: 48596904-48596904 on Build GRCh38
rs758925227 Chr.19: 48596753-48596753 on Build GRCh38
rs567294175 Chr.19: 48596981-48596981 on Build GRCh38
rs146884008 Chr.19: 48596896-48596896 on Build GRCh38
rs2302947 Chr.19: 48596811-48596811 on Build GRCh38
rs151077045 Chr.19: 48596813-48596813 on Build GRCh38
rs771791526 Chr.19: 48596754-48596754 on Build GRCh38
rs746709836 Chr.19: 48596891-48596891 on Build GRCh38
rs781232514 Chr.19: 48596728-48596728 on Build GRCh38
rs899341554 Chr.19: 48596932-48596932 on Build GRCh38
rs373999064 Chr.19: 48596786-48596786 on Build GRCh38
rs534674352 Chr.19: 48597026-48597026 on Build GRCh38
rs546703156 Chr.19: 48596827-48596827 on Build GRCh38
rs1026648315 Chr.19: 48597020-48597020 on Build GRCh38
rs761474582 Chr.19: 48596820-48596820 on Build GRCh38
rs370064077 Chr.19: 48596834-48596834 on Build GRCh38
rs112884737 Chr.19: 48596867-48596867 on Build GRCh38
rs1041356325 Chr.19: 48596686-48596689 on Build GRCh38
rs764247607 Chr.19: 48596845-48596845 on Build GRCh38
rs935576247 Chr.19: 48597112-48597112 on Build GRCh38
rs764129407 Chr.19: 48596691-48596691 on Build GRCh38
rs138458856 Chr.19: 48597106-48597106 on Build GRCh38
rs776545829 Chr.19: 48596819-48596819 on Build GRCh38
rs528536213 Chr.19: 48596814-48596814 on Build GRCh38
rs549027793 Chr.19: 48596923-48596923 on Build GRCh38
rs1012311607 Chr.19: 48596726-48596726 on Build GRCh38
rs781489535 Chr.19: 48596785-48596785 on Build GRCh38
rs774942612 Chr.19: 48596758-48596758 on Build GRCh38
rs192715000 Chr.19: 48597107-48597107 on Build GRCh38
rs1017999475 Chr.19: 48597074-48597074 on Build GRCh38
rs749794091 Chr.19: 48596747-48596747 on Build GRCh38
rs985135712 Chr.19: 48597069-48597069 on Build GRCh38
rs376261406 Chr.19: 48596924-48596924 on Build GRCh38
rs924131600 Chr.19: 48597111-48597111 on Build GRCh38
rs751691295 Chr.19: 48596781-48596781 on Build GRCh38
rs533667517 Chr.19: 48596828-48596828 on Build GRCh38
rs866168990 Chr.19: 48596977-48596977 on Build GRCh38
rs1051549233 Chr.19: 48596701-48596701 on Build GRCh38
rs755251687 Chr.19: 48596714-48596714 on Build GRCh38
rs3815691 Chr.19: 48596698-48596698 on Build GRCh38
rs746349876 Chr.19: 48596864-48596864 on Build GRCh38
rs758811349 Chr.19: 48596961-48596961 on Build GRCh38
rs778313193 Chr.19: 48596746-48596746 on Build GRCh38
rs771429001 Chr.19: 48596810-48596810 on Build GRCh38
rs967321424 Chr.19: 48597052-48597052 on Build GRCh38
rs1038953511 Chr.19: 48596879-48596879 on Build GRCh38
rs543822912 Chr.19: 48596999-48596999 on Build GRCh38
rs757500882 Chr.19: 48596856-48596856 on Build GRCh38
rs780176168 Chr.19: 48596699-48596699 on Build GRCh38
rs760562701 Chr.19: 48596759-48596759 on Build GRCh38
rs199842613 Chr.19: 48596835-48596835 on Build GRCh38
rs957936501 Chr.19: 48597104-48597104 on Build GRCh38
rs753768862 Chr.19: 48596697-48596697 on Build GRCh38
rs201499986 Chr.19: 48596905-48596905 on Build GRCh38
rs756535883 Chr.19: 48596789-48596789 on Build GRCh38
rs766419184 Chr.19: 48596766-48596766 on Build GRCh38
rs16982170 Chr.19: 48596922-48596922 on Build GRCh38
rs763970039 Chr.19: 48596760-48596760 on Build GRCh38
rs553115951 Chr.19: 48597065-48597065 on Build GRCh38
rs749839881 Chr.19: 48596798-48596798 on Build GRCh38
rs751146133 Chr.19: 48596966-48596966 on Build GRCh38
rs757800349 Chr.19: 48596806-48596806 on Build GRCh38
rs143866846 Chr.19: 48596866-48596866 on Build GRCh38
rs770390968 Chr.19: 48596748-48596748 on Build GRCh38
rs542643724 Chr.19: 48596796-48596796 on Build GRCh38
rs747388263 Chr.19: 48596965-48596965 on Build GRCh38
rs16982169 Chr.19: 48596782-48596782 on Build GRCh38
rs140730805 Chr.19: 48596900-48596900 on Build GRCh38
rs765224593 Chr.19: 48596823-48596823 on Build GRCh38
rs762765702 Chr.19: 48596914-48596914 on Build GRCh38
rs978822555 Chr.19: 48597064-48597064 on Build GRCh38
rs777282944 Chr.19: 48596668-48596668 on Build GRCh38
rs745390863 Chr.19: 48596807-48596807 on Build GRCh38
rs370301738 Chr.19: 48596952-48596952 on Build GRCh38
rs141049192 Chr.19: 48596849-48596849 on Build GRCh38
rs773829573 Chr.19: 48596730-48596730 on Build GRCh38
rs376041292 Chr.19: 48596713-48596713 on Build GRCh38
rs780659441 Chr.19: 48596962-48596962 on Build GRCh38
rs756303048 Chr.19: 48596837-48596837 on Build GRCh38
rs754005949 Chr.19: 48596928-48596928 on Build GRCh38
rs370301606 Chr.19: 48596739-48596739 on Build GRCh38
rs994514830 Chr.19: 48596990-48596990 on Build GRCh38
rs773893795 Chr.19: 48596750-48596750 on Build GRCh38
rs148634243 Chr.19: 48596761-48596761 on Build GRCh38
rs1020224471 Chr.19: 48597013-48597013 on Build GRCh38
rs765517576 Chr.19: 48596931-48596931 on Build GRCh38
rs757702000 Chr.19: 48596929-48596929 on Build GRCh38
rs202103681 Chr.19: 48596895-48596895 on Build GRCh38
rs948136772 Chr.19: 48596927-48596927 on Build GRCh38

View Full Product Details