Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57849707-57850201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF587
Entrez Gene ID
84914
Gene Name
zinc finger protein 587
Gene Aliases
ZF6
UniGene
Hs.642598
Gene Chromosome Location
Chr.19: 57849813-57865123 on Build GRCh38
Transcripts
NM_001204817.1 NM_032828.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6152558 c.10G>T p.A4S Chr.19: 57850048-57850048 on Build GRCh38
1326117 c.18G>A p.P6P Chr.19: 57850056-57850056 on Build GRCh38
7109851 c.33G>A p.Q11Q Chr.19: 57850071-57850071 on Build GRCh38
6376304 c.3G>T p.M1I Chr.19: 57850041-57850041 on Build GRCh38
6585442 c.7G>A p.A3T Chr.19: 57850045-57850045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143135812 Chr.19: 57850084-57850084 on Build GRCh38
rs113336552 Chr.19: 57849756-57849756 on Build GRCh38
rs1028044232 Chr.19: 57850140-57850140 on Build GRCh38
rs968245512 Chr.19: 57849978-57849978 on Build GRCh38
rs766005988 Chr.19: 57850038-57850038 on Build GRCh38
rs375933945 Chr.19: 57850001-57850001 on Build GRCh38
rs879170083 Chr.19: 57849767-57849767 on Build GRCh38
rs372730849 Chr.19: 57849992-57849992 on Build GRCh38
rs1022452723 Chr.19: 57849968-57849968 on Build GRCh38
rs541719372 Chr.19: 57849952-57849952 on Build GRCh38
rs755863325 Chr.19: 57849998-57849998 on Build GRCh38
rs781335414 Chr.19: 57850063-57850063 on Build GRCh38
rs528034712 Chr.19: 57849840-57849840 on Build GRCh38
rs774217649 Chr.19: 57850069-57850069 on Build GRCh38
rs528096107 Chr.19: 57850036-57850036 on Build GRCh38
rs150767167 Chr.19: 57849909-57849909 on Build GRCh38
rs754205261 Chr.19: 57850000-57850000 on Build GRCh38
rs28577146 Chr.19: 57849966-57849966 on Build GRCh38
rs933684336 Chr.19: 57849925-57849925 on Build GRCh38
rs58754895 Chr.19: 57850181-57850181 on Build GRCh38
rs377053348 Chr.19: 57850106-57850106 on Build GRCh38
rs572758356 Chr.19: 57849941-57849941 on Build GRCh38
rs757343887 Chr.19: 57850060-57850060 on Build GRCh38
rs775225203 Chr.19: 57850135-57850135 on Build GRCh38
rs771669074 Chr.19: 57850070-57850070 on Build GRCh38
rs117112607 Chr.19: 57849832-57849832 on Build GRCh38
rs887043475 Chr.19: 57849931-57849931 on Build GRCh38
rs377089857 Chr.19: 57850030-57850030 on Build GRCh38
rs780401779 Chr.19: 57850110-57850110 on Build GRCh38
rs111230161 Chr.19: 57849962-57849962 on Build GRCh38
rs576528002 Chr.19: 57849934-57849934 on Build GRCh38
rs535023272 Chr.19: 57850151-57850151 on Build GRCh38
rs764640053 Chr.19: 57850088-57850088 on Build GRCh38
rs1038155615 Chr.19: 57849935-57849935 on Build GRCh38
rs573378306 Chr.19: 57850177-57850177 on Build GRCh38
rs184357272 Chr.19: 57849791-57849791 on Build GRCh38
rs190088441 Chr.19: 57849816-57849816 on Build GRCh38
rs563940898 Chr.19: 57850170-57850170 on Build GRCh38
rs368980517 Chr.19: 57849990-57849990 on Build GRCh38
rs760425658 Chr.19: 57850149-57850149 on Build GRCh38
rs899528107 Chr.19: 57849798-57849798 on Build GRCh38
rs563230480 Chr.19: 57849989-57849989 on Build GRCh38
rs748016117 Chr.19: 57850115-57850115 on Build GRCh38
rs546008824 Chr.19: 57849751-57849751 on Build GRCh38
rs775950883 Chr.19: 57850081-57850081 on Build GRCh38
rs1003954114 Chr.19: 57849954-57849954 on Build GRCh38
rs997019338 Chr.19: 57850122-57850122 on Build GRCh38
rs182311323 Chr.19: 57849852-57849852 on Build GRCh38
rs946433539 Chr.19: 57849916-57849916 on Build GRCh38
rs555920556 Chr.19: 57850108-57850108 on Build GRCh38
rs748130968 Chr.19: 57850022-57850026 on Build GRCh38
rs558120072 Chr.19: 57850152-57850152 on Build GRCh38
rs114721771 Chr.19: 57849875-57849875 on Build GRCh38
rs779359053 Chr.19: 57850005-57850005 on Build GRCh38
rs868231673 Chr.19: 57850127-57850127 on Build GRCh38
rs754524408 Chr.19: 57850043-57850043 on Build GRCh38
rs778219430 Chr.19: 57850054-57850054 on Build GRCh38
rs376758560 Chr.19: 57849800-57849800 on Build GRCh38
rs190896843 Chr.19: 57850032-57850032 on Build GRCh38
rs749422351 Chr.19: 57850058-57850058 on Build GRCh38
rs370293189 Chr.19: 57850090-57850090 on Build GRCh38
rs758738317 Chr.19: 57850086-57850086 on Build GRCh38
rs373907556 Chr.19: 57850099-57850099 on Build GRCh38
rs548612241 Chr.19: 57849878-57849878 on Build GRCh38
rs199622081 Chr.19: 57850052-57850052 on Build GRCh38
rs769850056 Chr.19: 57850068-57850068 on Build GRCh38
rs199739385 Chr.19: 57850028-57850028 on Build GRCh38
rs28369370 Chr.19: 57850082-57850082 on Build GRCh38
rs765650299 Chr.19: 57850044-57850044 on Build GRCh38
rs747137509 Chr.19: 57850057-57850057 on Build GRCh38
rs577250768 Chr.19: 57849735-57849735 on Build GRCh38
rs551221879 Chr.19: 57850037-57850037 on Build GRCh38
rs760331754 Chr.19: 57850078-57850078 on Build GRCh38
rs115379680 Chr.19: 57849917-57849917 on Build GRCh38
rs765920721 Chr.19: 57850079-57850079 on Build GRCh38
rs61030302 Chr.19: 57849845-57849845 on Build GRCh38
rs112900689 Chr.19: 57850061-57850061 on Build GRCh38
rs748695423 Chr.19: 57850013-57850013 on Build GRCh38
rs920936115 Chr.19: 57849920-57849920 on Build GRCh38
rs769811775 Chr.19: 57849901-57849901 on Build GRCh38
rs572439961 Chr.19: 57850111-57850111 on Build GRCh38
rs139873841 Chr.19: 57850003-57850003 on Build GRCh38
rs28374851 Chr.19: 57849960-57849960 on Build GRCh38
rs755429769 Chr.19: 57849775-57849775 on Build GRCh38
rs551137689 Chr.19: 57849896-57849896 on Build GRCh38
rs1039398793 Chr.19: 57849794-57849794 on Build GRCh38
rs987852981 Chr.19: 57849758-57849758 on Build GRCh38
rs1035691039 Chr.19: 57849894-57849894 on Build GRCh38
rs565628771 Chr.19: 57849881-57849881 on Build GRCh38
rs757636292 Chr.19: 57850097-57850097 on Build GRCh38
rs534635237 Chr.19: 57849893-57849893 on Build GRCh38
rs955288876 Chr.19: 57850146-57850146 on Build GRCh38
rs772512985 Chr.19: 57850017-57850017 on Build GRCh38
rs373035916 Chr.19: 57850027-57850027 on Build GRCh38
rs747921611 Chr.19: 57850071-57850071 on Build GRCh38
rs747049604 Chr.19: 57850023-57850023 on Build GRCh38
rs982796854 Chr.19: 57849732-57849732 on Build GRCh38
rs753013896 Chr.19: 57850046-57850046 on Build GRCh38
rs776225152 Chr.19: 57849799-57849799 on Build GRCh38
rs569740754 Chr.19: 57849873-57849873 on Build GRCh38
rs373843081 Chr.19: 57850065-57850065 on Build GRCh38
rs774402947 Chr.19: 57850075-57850075 on Build GRCh38
rs187378277 Chr.19: 57849940-57849940 on Build GRCh38
rs777850179 Chr.19: 57850019-57850019 on Build GRCh38
rs570037570 Chr.19: 57850085-57850085 on Build GRCh38
rs577165076 Chr.19: 57849979-57849979 on Build GRCh38
rs753528255 Chr.19: 57850041-57850041 on Build GRCh38
rs928153504 Chr.19: 57849740-57849740 on Build GRCh38
rs535841966 Chr.19: 57849938-57849938 on Build GRCh38
rs578227686 Chr.19: 57850153-57850153 on Build GRCh38
rs771990138 Chr.19: 57850123-57850123 on Build GRCh38
rs772869871 Chr.19: 57850078-57850080 on Build GRCh38
rs556337792 Chr.19: 57849930-57849930 on Build GRCh38
rs1023711167 Chr.19: 57849947-57849947 on Build GRCh38
rs386811357 Chr.19: 57849960-57849962 on Build GRCh38
rs781346399 Chr.19: 57850098-57850098 on Build GRCh38
rs543643126 Chr.19: 57850155-57850155 on Build GRCh38
rs541624897 Chr.19: 57849841-57849841 on Build GRCh38
rs112151138 Chr.19: 57849853-57849853 on Build GRCh38

View Full Product Details