Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10443117-10443650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ODC1
Entrez Gene ID
4953
Gene Name
ornithine decarboxylase 1
Gene Aliases
ODC
UniGene
Hs.467701
Gene Chromosome Location
Chr.2: 10440371-10448554 on Build GRCh38
Transcripts
NM_001287188.1 NM_001287189.1 NM_001287190.1 NM_002539.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398571 c.705C>T p.G235G Chr.2: 10443275-10443275 on Build GRCh38
6732891 c.669T>G p.A223A Chr.2: 10443311-10443311 on Build GRCh38
5859805 c.657delT p.F219fs*26 Chr.2: 10443499-10443499 on Build GRCh38
5897387 c.610G>A p.D204N Chr.2: 10443546-10443546 on Build GRCh38
4167906 c.666+5G>C p.? Chr.2: 10443485-10443485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767987899 Chr.2: 10443221-10443221 on Build GRCh38
rs770600498 Chr.2: 10443515-10443515 on Build GRCh38
rs28362402 Chr.2: 10443584-10443585 on Build GRCh38
rs762333743 Chr.2: 10443576-10443576 on Build GRCh38
rs200737468 Chr.2: 10443458-10443458 on Build GRCh38
rs755925883 Chr.2: 10443556-10443556 on Build GRCh38
rs994753121 Chr.2: 10443415-10443415 on Build GRCh38
rs764843880 Chr.2: 10443192-10443192 on Build GRCh38
rs199713576 Chr.2: 10443459-10443459 on Build GRCh38
rs773974304 Chr.2: 10443517-10443517 on Build GRCh38
rs762066558 Chr.2: 10443543-10443543 on Build GRCh38
rs138255982 Chr.2: 10443407-10443407 on Build GRCh38
rs759064940 Chr.2: 10443518-10443518 on Build GRCh38
rs13396481 Chr.2: 10443195-10443195 on Build GRCh38
rs775665889 Chr.2: 10443304-10443304 on Build GRCh38
rs201608190 Chr.2: 10443593-10443593 on Build GRCh38
rs747926407 Chr.2: 10443311-10443311 on Build GRCh38
rs575117755 Chr.2: 10443454-10443454 on Build GRCh38
rs1041108783 Chr.2: 10443401-10443401 on Build GRCh38
rs751540790 Chr.2: 10443480-10443480 on Build GRCh38
rs980206699 Chr.2: 10443159-10443159 on Build GRCh38
rs190678797 Chr.2: 10443352-10443352 on Build GRCh38
rs758404248 Chr.2: 10443549-10443549 on Build GRCh38
rs776380566 Chr.2: 10443297-10443297 on Build GRCh38
rs757339527 Chr.2: 10443475-10443475 on Build GRCh38
rs769574543 Chr.2: 10443508-10443508 on Build GRCh38
rs746937871 Chr.2: 10443279-10443279 on Build GRCh38
rs917778049 Chr.2: 10443218-10443218 on Build GRCh38
rs114387410 Chr.2: 10443565-10443565 on Build GRCh38
rs747898520 Chr.2: 10443504-10443504 on Build GRCh38
rs765695722 Chr.2: 10443591-10443591 on Build GRCh38
rs1005776321 Chr.2: 10443468-10443468 on Build GRCh38
rs548065140 Chr.2: 10443344-10443344 on Build GRCh38
rs571097597 Chr.2: 10443402-10443402 on Build GRCh38
rs778007589 Chr.2: 10443262-10443262 on Build GRCh38
rs373811110 Chr.2: 10443198-10443198 on Build GRCh38
rs374811349 Chr.2: 10443623-10443623 on Build GRCh38
rs772853982 Chr.2: 10443336-10443336 on Build GRCh38
rs373045430 Chr.2: 10443495-10443495 on Build GRCh38
rs548307029 Chr.2: 10443252-10443252 on Build GRCh38
rs564731756 Chr.2: 10443609-10443609 on Build GRCh38
rs148268330 Chr.2: 10443190-10443190 on Build GRCh38
rs753137096 Chr.2: 10443450-10443450 on Build GRCh38
rs771087689 Chr.2: 10443198-10443200 on Build GRCh38
rs765352140 Chr.2: 10443546-10443546 on Build GRCh38
rs11538363 Chr.2: 10443275-10443275 on Build GRCh38
rs752188928 Chr.2: 10443325-10443325 on Build GRCh38
rs764510141 Chr.2: 10443529-10443529 on Build GRCh38
rs534291934 Chr.2: 10443340-10443340 on Build GRCh38
rs374970523 Chr.2: 10443350-10443350 on Build GRCh38
rs530142309 Chr.2: 10443214-10443214 on Build GRCh38
rs754889450 Chr.2: 10443482-10443482 on Build GRCh38
rs777348378 Chr.2: 10443509-10443509 on Build GRCh38
rs1052222200 Chr.2: 10443330-10443330 on Build GRCh38
rs193033745 Chr.2: 10443405-10443405 on Build GRCh38
rs756265506 Chr.2: 10443253-10443253 on Build GRCh38
rs752053612 Chr.2: 10443205-10443205 on Build GRCh38
rs768506216 Chr.2: 10443289-10443289 on Build GRCh38
rs754074772 Chr.2: 10443536-10443536 on Build GRCh38
rs761240909 Chr.2: 10443611-10443611 on Build GRCh38
rs766798214 Chr.2: 10443200-10443200 on Build GRCh38
rs765895777 Chr.2: 10443349-10443349 on Build GRCh38
rs770727071 Chr.2: 10443566-10443566 on Build GRCh38
rs768384026 Chr.2: 10443181-10443181 on Build GRCh38
rs772740739 Chr.2: 10443522-10443522 on Build GRCh38
rs778672400 Chr.2: 10443578-10443578 on Build GRCh38
rs745976547 Chr.2: 10443189-10443189 on Build GRCh38
rs1042340617 Chr.2: 10443287-10443287 on Build GRCh38
rs746795611 Chr.2: 10443200-10443201 on Build GRCh38
rs759097637 Chr.2: 10443615-10443615 on Build GRCh38
rs990554190 Chr.2: 10443212-10443212 on Build GRCh38
rs891690955 Chr.2: 10443375-10443375 on Build GRCh38
rs781120730 Chr.2: 10443555-10443555 on Build GRCh38
rs889771030 Chr.2: 10443354-10443354 on Build GRCh38
rs376328527 Chr.2: 10443592-10443592 on Build GRCh38
rs768434315 Chr.2: 10443202-10443205 on Build GRCh38
rs143860094 Chr.2: 10443274-10443274 on Build GRCh38
rs145681887 Chr.2: 10443547-10443547 on Build GRCh38
rs768941724 Chr.2: 10443622-10443622 on Build GRCh38
rs769697899 Chr.2: 10443622-10443622 on Build GRCh38
rs769360745 Chr.2: 10443317-10443317 on Build GRCh38
rs942230769 Chr.2: 10443599-10443600 on Build GRCh38
rs1031591392 Chr.2: 10443368-10443368 on Build GRCh38
rs778959430 Chr.2: 10443479-10443479 on Build GRCh38
rs779149420 Chr.2: 10443199-10443200 on Build GRCh38
rs1006953263 Chr.2: 10443371-10443371 on Build GRCh38
rs776001478 Chr.2: 10443526-10443526 on Build GRCh38
rs768040994 Chr.2: 10443445-10443445 on Build GRCh38
rs777589727 Chr.2: 10443558-10443558 on Build GRCh38
rs776409985 Chr.2: 10443356-10443357 on Build GRCh38
rs747288907 Chr.2: 10443319-10443319 on Build GRCh38
rs760964873 Chr.2: 10443527-10443527 on Build GRCh38

View Full Product Details