Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27212262-27212795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRAID
Entrez Gene ID
51374
Gene Name
all-trans retinoic acid induced differentiation factor
Gene Aliases
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18
UniGene
Hs.9527
Gene Chromosome Location
Chr.2: 27212031-27217178 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3 NM_021095.2 NR_028323.1

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
UniGene
Hs.435735
Gene Chromosome Location
Chr.2: 27199587-27212307 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3 NM_021095.2 NR_028323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380133 c.194C>T p.T65M Chr.2: 27212397-27212397 on Build GRCh38
4847399 c.145G>A p.G49R Chr.2: 27212348-27212348 on Build GRCh38
5417473 c.240G>A p.V80V Chr.2: 27212443-27212443 on Build GRCh38
7290830 c.194C>G p.T65R Chr.2: 27212397-27212397 on Build GRCh38
6429032 c.212C>T p.A71V Chr.2: 27212415-27212415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921323005 Chr.2: 27212525-27212525 on Build GRCh38
rs780717342 Chr.2: 27212350-27212350 on Build GRCh38
rs369544433 Chr.2: 27212345-27212345 on Build GRCh38
rs767986269 Chr.2: 27212388-27212388 on Build GRCh38
rs771834206 Chr.2: 27212374-27212374 on Build GRCh38
rs993655060 Chr.2: 27212584-27212584 on Build GRCh38
rs202067119 Chr.2: 27212288-27212288 on Build GRCh38
rs932640153 Chr.2: 27212526-27212526 on Build GRCh38
rs370704734 Chr.2: 27212504-27212504 on Build GRCh38
rs11608 Chr.2: 27212506-27212506 on Build GRCh38
rs1026448265 Chr.2: 27212586-27212586 on Build GRCh38
rs934494386 Chr.2: 27212444-27212444 on Build GRCh38
rs559815627 Chr.2: 27212331-27212331 on Build GRCh38
rs1052688429 Chr.2: 27212358-27212358 on Build GRCh38
rs1015817858 Chr.2: 27212771-27212771 on Build GRCh38
rs915873588 Chr.2: 27212392-27212392 on Build GRCh38
rs1049476466 Chr.2: 27212746-27212747 on Build GRCh38
rs1019174834 Chr.2: 27212576-27212576 on Build GRCh38
rs765299018 Chr.2: 27212298-27212298 on Build GRCh38
rs185160026 Chr.2: 27212393-27212393 on Build GRCh38
rs775244461 Chr.2: 27212464-27212464 on Build GRCh38
rs773378990 Chr.2: 27212499-27212499 on Build GRCh38
rs754285511 Chr.2: 27212758-27212758 on Build GRCh38
rs139490556 Chr.2: 27212596-27212596 on Build GRCh38
rs201763328 Chr.2: 27212397-27212397 on Build GRCh38
rs911844796 Chr.2: 27212530-27212530 on Build GRCh38
rs766269297 Chr.2: 27212708-27212713 on Build GRCh38
rs774303259 Chr.2: 27212373-27212373 on Build GRCh38
rs188379657 Chr.2: 27212401-27212401 on Build GRCh38
rs892282740 Chr.2: 27212580-27212580 on Build GRCh38
rs201498828 Chr.2: 27212400-27212400 on Build GRCh38
rs780518356 Chr.2: 27212710-27212710 on Build GRCh38
rs946544324 Chr.2: 27212582-27212582 on Build GRCh38
rs1052301070 Chr.2: 27212557-27212557 on Build GRCh38
rs79602377 Chr.2: 27212468-27212468 on Build GRCh38
rs983008746 Chr.2: 27212491-27212491 on Build GRCh38
rs765352518 Chr.2: 27212378-27212378 on Build GRCh38
rs201210592 Chr.2: 27212318-27212318 on Build GRCh38
rs896757153 Chr.2: 27212731-27212731 on Build GRCh38
rs944561840 Chr.2: 27212346-27212346 on Build GRCh38
rs372747928 Chr.2: 27212390-27212390 on Build GRCh38
rs770036360 Chr.2: 27212473-27212473 on Build GRCh38
rs981369967 Chr.2: 27212402-27212402 on Build GRCh38
rs192827384 Chr.2: 27212707-27212707 on Build GRCh38
rs776777155 Chr.2: 27212292-27212292 on Build GRCh38
rs749286000 Chr.2: 27212360-27212360 on Build GRCh38
rs369868002 Chr.2: 27212324-27212324 on Build GRCh38
rs376559066 Chr.2: 27212355-27212355 on Build GRCh38
rs778720867 Chr.2: 27212423-27212423 on Build GRCh38
rs889429012 Chr.2: 27212560-27212560 on Build GRCh38
rs373432775 Chr.2: 27212352-27212352 on Build GRCh38
rs1001834084 Chr.2: 27212611-27212611 on Build GRCh38
rs11556163 Chr.2: 27212297-27212297 on Build GRCh38
rs1049961713 Chr.2: 27212559-27212559 on Build GRCh38
rs970066599 Chr.2: 27212705-27212705 on Build GRCh38
rs751904846 Chr.2: 27212344-27212344 on Build GRCh38
rs755297948 Chr.2: 27212385-27212385 on Build GRCh38
rs904913335 Chr.2: 27212607-27212607 on Build GRCh38
rs750988045 Chr.2: 27212301-27212301 on Build GRCh38
rs752452216 Chr.2: 27212389-27212389 on Build GRCh38
rs762960190 Chr.2: 27212500-27212500 on Build GRCh38
rs200120344 Chr.2: 27212372-27212372 on Build GRCh38
rs755750046 Chr.2: 27212354-27212354 on Build GRCh38
rs768041005 Chr.2: 27212511-27212511 on Build GRCh38
rs952192986 Chr.2: 27212601-27212601 on Build GRCh38
rs1008337095 Chr.2: 27212562-27212562 on Build GRCh38
rs1007788608 Chr.2: 27212329-27212329 on Build GRCh38
rs944619519 Chr.2: 27212548-27212548 on Build GRCh38
rs558107241 Chr.2: 27212755-27212755 on Build GRCh38
rs1275533 Chr.2: 27212382-27212382 on Build GRCh38
rs376508461 Chr.2: 27212516-27212516 on Build GRCh38
rs764253186 Chr.2: 27212290-27212290 on Build GRCh38
rs1019627140 Chr.2: 27212342-27212342 on Build GRCh38
rs995795518 Chr.2: 27212321-27212336 on Build GRCh38
rs762010882 Chr.2: 27212377-27212377 on Build GRCh38
rs986763902 Chr.2: 27212529-27212529 on Build GRCh38
rs1023500601 Chr.2: 27212667-27212667 on Build GRCh38
rs947099019 Chr.2: 27212547-27212549 on Build GRCh38
rs780667170 Chr.2: 27212407-27212431 on Build GRCh38
rs866330510 Chr.2: 27212465-27212465 on Build GRCh38
rs745813413 Chr.2: 27212432-27212432 on Build GRCh38
rs143266715 Chr.2: 27212733-27212733 on Build GRCh38
rs1035238411 Chr.2: 27212419-27212419 on Build GRCh38
rs756742309 Chr.2: 27212407-27212407 on Build GRCh38
rs535243797 Chr.2: 27212524-27212524 on Build GRCh38
rs771821798 Chr.2: 27212442-27212442 on Build GRCh38
rs988694419 Chr.2: 27212720-27212720 on Build GRCh38
rs762557057 Chr.2: 27212295-27212295 on Build GRCh38
rs1027955698 Chr.2: 27212316-27212323 on Build GRCh38
rs768222803 Chr.2: 27212286-27212287 on Build GRCh38
rs773247722 Chr.2: 27212381-27212381 on Build GRCh38
rs1034641410 Chr.2: 27212724-27212724 on Build GRCh38
rs934546786 Chr.2: 27212712-27212713 on Build GRCh38
rs774783367 Chr.2: 27212505-27212505 on Build GRCh38
rs752042458 Chr.2: 27212384-27212384 on Build GRCh38
rs963223191 Chr.2: 27212776-27212776 on Build GRCh38
rs536732085 Chr.2: 27212717-27212717 on Build GRCh38
rs776906751 Chr.2: 27212375-27212375 on Build GRCh38
rs928548142 Chr.2: 27212711-27212711 on Build GRCh38
rs748413481 Chr.2: 27212467-27212467 on Build GRCh38

View Full Product Details