Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28540283-28540774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLB1
Entrez Gene ID
151056
Gene Name
phospholipase B1
Gene Aliases
PLB, PLB/LIP, hPLB
UniGene
Hs.444933
Gene Chromosome Location
Chr.2: 28488579-28644135 on Build GRCh38
Transcripts
NM_001170585.1 NM_153021.4 XM_011532579.2 XM_011532581.2 XM_011532584.2 XM_011532588.2 XM_011532589.2 XM_011532590.2 XM_011532591.2 XM_011532593.2 XM_011532606.2 XM_011532607.2 XM_011532608.2 XM_017003432.1 XM_017003433.1 XM_017003434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043020 c.732-1G>T p.? Chr.2: 28540365-28540365 on Build GRCh38
5396839 c.772C>T p.Q258* Chr.2: 28540439-28540439 on Build GRCh38
322683 c.734C>A p.T245N Chr.2: 28540401-28540401 on Build GRCh38
1752520 c.730G>T p.E244* Chr.2: 28540397-28540397 on Build GRCh38
3723074 c.745C>T p.P249S Chr.2: 28540379-28540379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994089158 Chr.2: 28540713-28540713 on Build GRCh38
rs781530495 Chr.2: 28540348-28540348 on Build GRCh38
rs762967220 Chr.2: 28540372-28540372 on Build GRCh38
rs957355193 Chr.2: 28540591-28540591 on Build GRCh38
rs202064271 Chr.2: 28540407-28540407 on Build GRCh38
rs780581164 Chr.2: 28540414-28540414 on Build GRCh38
rs533730324 Chr.2: 28540632-28540632 on Build GRCh38
rs558080017 Chr.2: 28540574-28540574 on Build GRCh38
rs776808721 Chr.2: 28540653-28540653 on Build GRCh38
rs925905069 Chr.2: 28540589-28540589 on Build GRCh38
rs748339727 Chr.2: 28540428-28540428 on Build GRCh38
rs539343552 Chr.2: 28540564-28540564 on Build GRCh38
rs79986357 Chr.2: 28540363-28540363 on Build GRCh38
rs768731304 Chr.2: 28540346-28540346 on Build GRCh38
rs1047961735 Chr.2: 28540743-28540743 on Build GRCh38
rs755039581 Chr.2: 28540418-28540418 on Build GRCh38
rs1046209063 Chr.2: 28540674-28540674 on Build GRCh38
rs781165167 Chr.2: 28540422-28540422 on Build GRCh38
rs573756887 Chr.2: 28540663-28540663 on Build GRCh38
rs952360532 Chr.2: 28540751-28540751 on Build GRCh38
rs964962262 Chr.2: 28540560-28540560 on Build GRCh38
rs1013388237 Chr.2: 28540667-28540667 on Build GRCh38
rs372207505 Chr.2: 28540525-28540525 on Build GRCh38
rs762990480 Chr.2: 28540340-28540341 on Build GRCh38
rs766208653 Chr.2: 28540352-28540353 on Build GRCh38
rs765580314 Chr.2: 28540402-28540402 on Build GRCh38
rs759133552 Chr.2: 28540378-28540378 on Build GRCh38
rs746308181 Chr.2: 28540334-28540334 on Build GRCh38
rs974750165 Chr.2: 28540561-28540561 on Build GRCh38
rs527493540 Chr.2: 28540482-28540482 on Build GRCh38
rs767237766 Chr.2: 28540318-28540318 on Build GRCh38
rs945225104 Chr.2: 28540655-28540655 on Build GRCh38
rs1026635228 Chr.2: 28540461-28540461 on Build GRCh38
rs757591840 Chr.2: 28540328-28540328 on Build GRCh38
rs902058650 Chr.2: 28540314-28540314 on Build GRCh38
rs144349565 Chr.2: 28540373-28540373 on Build GRCh38
rs770968377 Chr.2: 28540440-28540440 on Build GRCh38
rs569070620 Chr.2: 28540557-28540557 on Build GRCh38
rs372009757 Chr.2: 28540340-28540340 on Build GRCh38
rs147681804 Chr.2: 28540439-28540439 on Build GRCh38
rs370652072 Chr.2: 28540487-28540487 on Build GRCh38
rs113570136 Chr.2: 28540468-28540468 on Build GRCh38
rs773150263 Chr.2: 28540371-28540371 on Build GRCh38
rs774381269 Chr.2: 28540450-28540450 on Build GRCh38
rs906429233 Chr.2: 28540441-28540441 on Build GRCh38
rs377497977 Chr.2: 28540456-28540456 on Build GRCh38
rs11127162 Chr.2: 28540507-28540507 on Build GRCh38
rs374563225 Chr.2: 28540492-28540492 on Build GRCh38
rs751619246 Chr.2: 28540416-28540416 on Build GRCh38
rs370654197 Chr.2: 28540493-28540493 on Build GRCh38
rs961292944 Chr.2: 28540426-28540426 on Build GRCh38
rs143724270 Chr.2: 28540686-28540686 on Build GRCh38
rs771971955 Chr.2: 28540585-28540586 on Build GRCh38
rs903960288 Chr.2: 28540336-28540336 on Build GRCh38
rs769560661 Chr.2: 28540366-28540366 on Build GRCh38
rs148963569 Chr.2: 28540406-28540406 on Build GRCh38
rs199572724 Chr.2: 28540479-28540479 on Build GRCh38
rs754129794 Chr.2: 28540320-28540320 on Build GRCh38
rs566657750 Chr.2: 28540588-28540588 on Build GRCh38
rs778823358 Chr.2: 28540434-28540434 on Build GRCh38
rs913762886 Chr.2: 28540419-28540419 on Build GRCh38
rs377350641 Chr.2: 28540486-28540486 on Build GRCh38
rs773966153 Chr.2: 28540376-28540376 on Build GRCh38
rs769964892 Chr.2: 28540431-28540431 on Build GRCh38
rs886166193 Chr.2: 28540745-28540745 on Build GRCh38
rs751132197 Chr.2: 28540392-28540393 on Build GRCh38
rs368933162 Chr.2: 28540478-28540478 on Build GRCh38
rs766958205 Chr.2: 28540380-28540380 on Build GRCh38
rs779177007 Chr.2: 28540331-28540331 on Build GRCh38
rs927882152 Chr.2: 28540669-28540669 on Build GRCh38
rs557263657 Chr.2: 28540717-28540717 on Build GRCh38
rs745979467 Chr.2: 28540716-28540716 on Build GRCh38
rs747162104 Chr.2: 28540345-28540345 on Build GRCh38
rs369284045 Chr.2: 28540335-28540335 on Build GRCh38
rs115884739 Chr.2: 28540645-28540645 on Build GRCh38
rs78313192 Chr.2: 28540726-28540726 on Build GRCh38
rs923537644 Chr.2: 28540506-28540506 on Build GRCh38
rs149777042 Chr.2: 28540400-28540400 on Build GRCh38
rs976247139 Chr.2: 28540504-28540504 on Build GRCh38
rs1013334552 Chr.2: 28540401-28540401 on Build GRCh38
rs753506890 Chr.2: 28540530-28540530 on Build GRCh38

View Full Product Details