Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 63381814-63382329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDPCP
Entrez Gene ID
51057
Gene Name
WD repeat containing planar cell polarity effector
Gene Aliases
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
UniGene
Hs.414952
Gene Chromosome Location
Chr.2: 63120755-63840825 on Build GRCh38
Transcripts
NM_001042692.2 NM_015910.5 NR_122106.1 XM_005264348.3 XM_011532881.2 XM_011532884.2 XM_011532886.2 XM_011532887.2 XM_011532890.2 XM_011532891.2 XM_017004251.1 XM_017004252.1 XM_017004253.1 XM_017004254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026368 c.1601A>T p.Q534L Chr.2: 63381929-63381929 on Build GRCh38
5350846 c.1568G>T p.S523I Chr.2: 63381962-63381962 on Build GRCh38
6400394 c.1556A>T p.Q519L Chr.2: 63381974-63381974 on Build GRCh38
3054887 c.1621_1622delGA p.E541fs*4 Chr.2: 63381908-63381909 on Build GRCh38
722102 c.1473C>A p.I491I Chr.2: 63382057-63382057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756632510 Chr.2: 63381970-63381970 on Build GRCh38
rs534102944 Chr.2: 63381954-63381954 on Build GRCh38
rs749341542 Chr.2: 63381864-63381864 on Build GRCh38
rs750822710 Chr.2: 63381965-63381965 on Build GRCh38
rs765160818 Chr.2: 63382120-63382120 on Build GRCh38
rs543073234 Chr.2: 63382205-63382205 on Build GRCh38
rs769679304 Chr.2: 63382070-63382070 on Build GRCh38
rs768036084 Chr.2: 63382083-63382083 on Build GRCh38
rs370961530 Chr.2: 63381900-63381900 on Build GRCh38
rs750245323 Chr.2: 63382079-63382079 on Build GRCh38
rs747270776 Chr.2: 63382144-63382144 on Build GRCh38
rs191316235 Chr.2: 63382277-63382277 on Build GRCh38
rs776475058 Chr.2: 63381889-63381889 on Build GRCh38
rs775015955 Chr.2: 63381909-63381909 on Build GRCh38
rs773663574 Chr.2: 63382098-63382098 on Build GRCh38
rs1025370415 Chr.2: 63382303-63382303 on Build GRCh38
rs781719914 Chr.2: 63381882-63381883 on Build GRCh38
rs769732237 Chr.2: 63381906-63381906 on Build GRCh38
rs768877891 Chr.2: 63381866-63381866 on Build GRCh38
rs955196377 Chr.2: 63381924-63381924 on Build GRCh38
rs780217225 Chr.2: 63381855-63381855 on Build GRCh38
rs959923850 Chr.2: 63382095-63382095 on Build GRCh38
rs10186743 Chr.2: 63382248-63382248 on Build GRCh38
rs1041077978 Chr.2: 63382255-63382255 on Build GRCh38
rs767292479 Chr.2: 63382093-63382093 on Build GRCh38
rs560350504 Chr.2: 63382242-63382242 on Build GRCh38
rs574567870 Chr.2: 63382005-63382005 on Build GRCh38
rs375693256 Chr.2: 63381955-63381955 on Build GRCh38
rs762400349 Chr.2: 63381934-63381934 on Build GRCh38
rs559295906 Chr.2: 63382025-63382025 on Build GRCh38
rs946794485 Chr.2: 63382006-63382006 on Build GRCh38
rs763698807 Chr.2: 63381943-63381943 on Build GRCh38
rs754281079 Chr.2: 63382119-63382119 on Build GRCh38
rs577571642 Chr.2: 63382039-63382039 on Build GRCh38
rs779428187 Chr.2: 63382004-63382004 on Build GRCh38
rs987143451 Chr.2: 63381930-63381930 on Build GRCh38
rs745681296 Chr.2: 63382060-63382060 on Build GRCh38
rs920932515 Chr.2: 63382259-63382261 on Build GRCh38
rs939509600 Chr.2: 63382072-63382072 on Build GRCh38
rs1003913232 Chr.2: 63382074-63382074 on Build GRCh38
rs756447932 Chr.2: 63381936-63381936 on Build GRCh38
rs754985173 Chr.2: 63382011-63382011 on Build GRCh38
rs758621800 Chr.2: 63382038-63382038 on Build GRCh38
rs977183769 Chr.2: 63382155-63382155 on Build GRCh38
rs188045310 Chr.2: 63382107-63382107 on Build GRCh38
rs993922285 Chr.2: 63382264-63382264 on Build GRCh38
rs1027267517 Chr.2: 63381842-63381846 on Build GRCh38
rs542294210 Chr.2: 63382218-63382218 on Build GRCh38
rs544657165 Chr.2: 63382082-63382082 on Build GRCh38
rs779761060 Chr.2: 63382258-63382258 on Build GRCh38
rs370842356 Chr.2: 63382064-63382064 on Build GRCh38
rs766821661 Chr.2: 63381984-63381984 on Build GRCh38
rs924335128 Chr.2: 63382243-63382243 on Build GRCh38
rs765940102 Chr.2: 63381887-63381887 on Build GRCh38
rs867801025 Chr.2: 63382027-63382027 on Build GRCh38
rs773852077 Chr.2: 63381950-63381950 on Build GRCh38
rs748711953 Chr.2: 63381934-63381936 on Build GRCh38
rs201412509 Chr.2: 63382092-63382092 on Build GRCh38
rs753917889 Chr.2: 63381991-63381991 on Build GRCh38
rs755369526 Chr.2: 63381861-63381862 on Build GRCh38
rs966853114 Chr.2: 63382153-63382153 on Build GRCh38
rs758563513 Chr.2: 63382125-63382125 on Build GRCh38
rs752784787 Chr.2: 63382122-63382122 on Build GRCh38
rs1042418470 Chr.2: 63382017-63382017 on Build GRCh38
rs766810747 Chr.2: 63382108-63382108 on Build GRCh38
rs145313329 Chr.2: 63382259-63382259 on Build GRCh38
rs779101316 Chr.2: 63382012-63382012 on Build GRCh38
rs915434456 Chr.2: 63382147-63382148 on Build GRCh38
rs779039231 Chr.2: 63381868-63381868 on Build GRCh38
rs368313345 Chr.2: 63382051-63382051 on Build GRCh38
rs2138796 Chr.2: 63381882-63381882 on Build GRCh38
rs778002102 Chr.2: 63382143-63382143 on Build GRCh38

View Full Product Details