Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157529992-157530491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1C
Entrez Gene ID
130399
Gene Name
activin A receptor type 1C
Gene Aliases
ACVRLK7, ALK7
UniGene
Hs.562901
Gene Chromosome Location
Chr.2: 157526767-157628887 on Build GRCh38
Transcripts
NM_001111031.1 NM_001111032.1 NM_001111033.1 NM_145259.2

SNP Detail

SNP ID Chromosome Location
rs984142589 Chr.2: 157530090-157530090 on Build GRCh38
rs114746894 Chr.2: 157530104-157530104 on Build GRCh38
rs913165966 Chr.2: 157530296-157530296 on Build GRCh38
rs4664229 Chr.2: 157530426-157530426 on Build GRCh38
rs753368533 Chr.2: 157530348-157530348 on Build GRCh38
rs867002749 Chr.2: 157530385-157530385 on Build GRCh38
rs964549196 Chr.2: 157530198-157530198 on Build GRCh38
rs557531354 Chr.2: 157530339-157530339 on Build GRCh38
rs1018794049 Chr.2: 157530174-157530174 on Build GRCh38
rs1029662992 Chr.2: 157530153-157530153 on Build GRCh38
rs766053900 Chr.2: 157530310-157530310 on Build GRCh38
rs939029860 Chr.2: 157530392-157530392 on Build GRCh38
rs760448559 Chr.2: 157530246-157530246 on Build GRCh38
rs978008038 Chr.2: 157530376-157530376 on Build GRCh38
rs897412702 Chr.2: 157530103-157530103 on Build GRCh38
rs989728150 Chr.2: 157530245-157530245 on Build GRCh38
rs773117384 Chr.2: 157530044-157530044 on Build GRCh38
rs952801509 Chr.2: 157530038-157530038 on Build GRCh38
rs189976880 Chr.2: 157530302-157530302 on Build GRCh38
rs377518751 Chr.2: 157530154-157530154 on Build GRCh38
rs1038595891 Chr.2: 157530069-157530069 on Build GRCh38
rs184723892 Chr.2: 157530264-157530264 on Build GRCh38
rs905569388 Chr.2: 157530379-157530379 on Build GRCh38
rs923082051 Chr.2: 157530244-157530244 on Build GRCh38
rs552015251 Chr.2: 157530055-157530055 on Build GRCh38
rs545551792 Chr.2: 157530304-157530304 on Build GRCh38
rs572971520 Chr.2: 157530352-157530352 on Build GRCh38
rs201918969 Chr.2: 157530226-157530226 on Build GRCh38
rs529522471 Chr.2: 157530444-157530444 on Build GRCh38
rs79478972 Chr.2: 157530380-157530380 on Build GRCh38
rs552900085 Chr.2: 157530216-157530216 on Build GRCh38
rs994367127 Chr.2: 157530447-157530447 on Build GRCh38
rs571995271 Chr.2: 157530283-157530283 on Build GRCh38
rs192346685 Chr.2: 157530171-157530171 on Build GRCh38
rs916322608 Chr.2: 157530442-157530442 on Build GRCh38
rs529248580 Chr.2: 157530450-157530450 on Build GRCh38
rs373340434 Chr.2: 157530057-157530057 on Build GRCh38

View Full Product Details