Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171318630-171319177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL8
Entrez Gene ID
79828
Gene Name
methyltransferase like 8
Gene Aliases
TIP
UniGene
Hs.135146
Gene Chromosome Location
Chr.2: 171315746-171434803 on Build GRCh38
Transcripts
NM_001321154.1 NM_001321155.1 NM_001321156.1 NM_001321157.1 NM_001321158.1 NM_001321159.1 NM_001321160.1 NM_001321161.1 NM_001321162.1 NM_024770.4 NR_135568.1 XM_011511875.2 XM_017004984.1 XM_017004985.1 XM_017004986.1 XM_017004987.1 XM_017004988.1 XM_017004989.1

SNP Detail

SNP ID Chromosome Location
rs995342031 Chr.2: 171319061-171319061 on Build GRCh38
rs75646511 Chr.2: 171318948-171318948 on Build GRCh38
rs527245799 Chr.2: 171318701-171318701 on Build GRCh38
rs746477448 Chr.2: 171318671-171318671 on Build GRCh38
rs889347573 Chr.2: 171318664-171318664 on Build GRCh38
rs187042361 Chr.2: 171319147-171319147 on Build GRCh38
rs917177831 Chr.2: 171318892-171318892 on Build GRCh38
rs962644800 Chr.2: 171318738-171318738 on Build GRCh38
rs189796747 Chr.2: 171319114-171319114 on Build GRCh38
rs983463959 Chr.2: 171318848-171318849 on Build GRCh38
rs151307994 Chr.2: 171318756-171318756 on Build GRCh38
rs377111104 Chr.2: 171318841-171318841 on Build GRCh38
rs897953815 Chr.2: 171318728-171318728 on Build GRCh38
rs557741212 Chr.2: 171319026-171319026 on Build GRCh38
rs556460280 Chr.2: 171318818-171318818 on Build GRCh38
rs184942114 Chr.2: 171318988-171318988 on Build GRCh38
rs1027713850 Chr.2: 171318755-171318755 on Build GRCh38
rs1054751983 Chr.2: 171318961-171318961 on Build GRCh38
rs887861556 Chr.2: 171319043-171319043 on Build GRCh38
rs962900859 Chr.2: 171319057-171319057 on Build GRCh38
rs576425893 Chr.2: 171319067-171319067 on Build GRCh38
rs1041004835 Chr.2: 171318998-171318998 on Build GRCh38
rs985328537 Chr.2: 171318734-171318734 on Build GRCh38
rs994060680 Chr.2: 171319001-171319001 on Build GRCh38
rs1050047318 Chr.2: 171318977-171318977 on Build GRCh38
rs1024611093 Chr.2: 171319139-171319139 on Build GRCh38
rs983436189 Chr.2: 171319101-171319101 on Build GRCh38
rs537758818 Chr.2: 171318803-171318803 on Build GRCh38
rs543759471 Chr.2: 171319113-171319113 on Build GRCh38
rs898090139 Chr.2: 171318999-171318999 on Build GRCh38
rs182372242 Chr.2: 171319129-171319129 on Build GRCh38
rs935775346 Chr.2: 171318963-171318963 on Build GRCh38
rs182525950 Chr.2: 171318879-171318879 on Build GRCh38
rs754924164 Chr.2: 171318824-171318824 on Build GRCh38
rs891430175 Chr.2: 171318976-171318976 on Build GRCh38
rs771043372 Chr.2: 171318991-171318991 on Build GRCh38
rs917354895 Chr.2: 171318859-171318859 on Build GRCh38
rs370551373 Chr.2: 171318902-171318902 on Build GRCh38
rs896441313 Chr.2: 171319087-171319087 on Build GRCh38
rs112538401 Chr.2: 171318737-171318737 on Build GRCh38
rs140549866 Chr.2: 171318950-171318950 on Build GRCh38
rs993122234 Chr.2: 171318882-171318882 on Build GRCh38
rs774361245 Chr.2: 171319070-171319070 on Build GRCh38
rs562244334 Chr.2: 171319046-171319046 on Build GRCh38
rs1002188113 Chr.2: 171318821-171318821 on Build GRCh38
rs79136732 Chr.2: 171318679-171318679 on Build GRCh38
rs929309792 Chr.2: 171318839-171318839 on Build GRCh38
rs553831026 Chr.2: 171318927-171318927 on Build GRCh38
rs1033722414 Chr.2: 171318838-171318838 on Build GRCh38

View Full Product Details