Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240559950-240560443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1 NM_001033575.1

Gene Symbol
DUSP28
Entrez Gene ID
285193
Gene Name
dual specificity phosphatase 28
Gene Aliases
DUSP26, VHP
UniGene
Hs.369297
Gene Chromosome Location
Chr.2: 240560054-240564014 on Build GRCh38
Transcripts
NM_001308375.1 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511315.2 XM_011511317.1 XM_011511318.1 XM_011511319.2 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1 NM_001033575.1

SNP Detail

SNP ID Chromosome Location
rs371315015 Chr.2: 240560350-240560350 on Build GRCh38
rs146875907 Chr.2: 240560014-240560014 on Build GRCh38
rs968178687 Chr.2: 240560108-240560108 on Build GRCh38
rs370117932 Chr.2: 240560199-240560199 on Build GRCh38
rs139045162 Chr.2: 240560052-240560052 on Build GRCh38
rs941142221 Chr.2: 240560211-240560211 on Build GRCh38
rs770825745 Chr.2: 240560231-240560231 on Build GRCh38
rs955336656 Chr.2: 240560078-240560078 on Build GRCh38
rs770730932 Chr.2: 240560209-240560209 on Build GRCh38
rs1000992931 Chr.2: 240560310-240560310 on Build GRCh38
rs1033258059 Chr.2: 240560316-240560319 on Build GRCh38
rs886868272 Chr.2: 240559995-240559995 on Build GRCh38
rs985026473 Chr.2: 240560348-240560348 on Build GRCh38
rs577783076 Chr.2: 240560208-240560208 on Build GRCh38
rs917141101 Chr.2: 240560320-240560320 on Build GRCh38
rs1000758029 Chr.2: 240560312-240560312 on Build GRCh38
rs548523931 Chr.2: 240560293-240560293 on Build GRCh38
rs1000260964 Chr.2: 240560054-240560054 on Build GRCh38
rs922908543 Chr.2: 240560385-240560385 on Build GRCh38
rs1040021548 Chr.2: 240560255-240560255 on Build GRCh38
rs774322877 Chr.2: 240560263-240560263 on Build GRCh38
rs903426242 Chr.2: 240560411-240560411 on Build GRCh38
rs767619312 Chr.2: 240560346-240560346 on Build GRCh38
rs888766674 Chr.2: 240559983-240559983 on Build GRCh38
rs573934496 Chr.2: 240560044-240560044 on Build GRCh38
rs1007217748 Chr.2: 240560010-240560010 on Build GRCh38
rs991326613 Chr.2: 240560319-240560319 on Build GRCh38
rs1020417001 Chr.2: 240560033-240560033 on Build GRCh38
rs925839141 Chr.2: 240560121-240560121 on Build GRCh38
rs772867314 Chr.2: 240560028-240560028 on Build GRCh38
rs191977005 Chr.2: 240560249-240560249 on Build GRCh38
rs1054993191 Chr.2: 240560420-240560420 on Build GRCh38
rs1033732985 Chr.2: 240560399-240560399 on Build GRCh38
rs1042776288 Chr.2: 240560389-240560389 on Build GRCh38
rs187581090 Chr.2: 240560421-240560421 on Build GRCh38
rs1033133506 Chr.2: 240560316-240560316 on Build GRCh38
rs1033492619 Chr.2: 240560056-240560056 on Build GRCh38
rs1029608655 Chr.2: 240560065-240560065 on Build GRCh38
rs971303467 Chr.2: 240560335-240560335 on Build GRCh38
rs945004128 Chr.2: 240560386-240560386 on Build GRCh38
rs1048829147 Chr.2: 240560232-240560232 on Build GRCh38
rs1024226757 Chr.2: 240560332-240560332 on Build GRCh38
rs368509546 Chr.2: 240560345-240560345 on Build GRCh38
rs1042000068 Chr.2: 240560401-240560401 on Build GRCh38
rs745629630 Chr.2: 240560390-240560390 on Build GRCh38
rs921022368 Chr.2: 240560349-240560349 on Build GRCh38
rs917204066 Chr.2: 240560156-240560156 on Build GRCh38
rs951945421 Chr.2: 240560347-240560347 on Build GRCh38
rs1009500100 Chr.2: 240560260-240560260 on Build GRCh38
rs112370628 Chr.2: 240560266-240560266 on Build GRCh38
rs937249686 Chr.2: 240560126-240560126 on Build GRCh38
rs562667813 Chr.2: 240560112-240560112 on Build GRCh38
rs1042296897 Chr.2: 240560267-240560267 on Build GRCh38
rs943054476 Chr.2: 240560251-240560251 on Build GRCh38
rs555695453 Chr.2: 240560037-240560037 on Build GRCh38
rs571793153 Chr.2: 240560353-240560353 on Build GRCh38
rs762875792 Chr.2: 240560057-240560057 on Build GRCh38
rs891391532 Chr.2: 240560255-240560255 on Build GRCh38
rs991441994 Chr.2: 240560155-240560155 on Build GRCh38
rs971319549 Chr.2: 240560322-240560322 on Build GRCh38
rs971290026 Chr.2: 240560415-240560415 on Build GRCh38
rs971882422 Chr.2: 240560196-240560196 on Build GRCh38
rs998355191 Chr.2: 240560031-240560031 on Build GRCh38
rs536684132 Chr.2: 240560015-240560015 on Build GRCh38
rs971249463 Chr.2: 240560162-240560162 on Build GRCh38
rs538894732 Chr.2: 240560354-240560354 on Build GRCh38
rs975758395 Chr.2: 240560362-240560362 on Build GRCh38
rs545072412 Chr.2: 240560221-240560221 on Build GRCh38
rs560561975 Chr.2: 240560318-240560318 on Build GRCh38
rs376352125 Chr.2: 240560262-240560262 on Build GRCh38
rs186078597 Chr.2: 240560224-240560224 on Build GRCh38
rs908265761 Chr.2: 240560193-240560193 on Build GRCh38
rs978986409 Chr.2: 240560110-240560110 on Build GRCh38
rs982426198 Chr.2: 240560337-240560337 on Build GRCh38
rs1001277949 Chr.2: 240560398-240560398 on Build GRCh38
rs760823937 Chr.2: 240560363-240560363 on Build GRCh38
rs988109867 Chr.2: 240560088-240560092 on Build GRCh38
rs968526834 Chr.2: 240560294-240560294 on Build GRCh38
rs1046775668 Chr.2: 240560181-240560181 on Build GRCh38
rs1020967017 Chr.2: 240560091-240560091 on Build GRCh38
rs991442559 Chr.2: 240560136-240560136 on Build GRCh38
rs917172277 Chr.2: 240560146-240560146 on Build GRCh38
rs547857857 Chr.2: 240560384-240560384 on Build GRCh38
rs183249869 Chr.2: 240560336-240560336 on Build GRCh38
rs776474180 Chr.2: 240560328-240560328 on Build GRCh38
rs764440802 Chr.2: 240560404-240560404 on Build GRCh38
rs55672855 Chr.2: 240560414-240560414 on Build GRCh38

View Full Product Details