Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241335894-241336390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEPT2
Entrez Gene ID
4735
Gene Name
septin 2
Gene Aliases
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
UniGene
Hs.721234
Gene Chromosome Location
Chr.2: 241315187-241354027 on Build GRCh38
Transcripts
NM_001008491.2 NM_001008492.2 NM_001282972.1 NM_001282973.1 NM_001321029.1 NM_001321030.1 NM_001321031.1 NM_001321032.1 NM_001321033.1 NM_001321034.1 NM_001321035.1 NM_004404.4 NM_006155.2 XM_011511237.1 XM_011511238.1 XM_011511240.1 XM_017004189.1 XM_017004190.1 XM_017004191.1 XM_017004192.1 XM_017004193.1 XM_017004194.1 XM_017004195.1 XM_017004196.1 XM_017004197.1 XM_017004198.1 XM_017004199.1 XM_017004200.1 XM_017004201.1 XM_017004202.1 XM_017004203.1 XM_017004204.1 XM_017004205.1 XM_017004206.1 XM_017004207.1 XM_017004208.1 XM_017004209.1 XM_017004210.1 XM_017004211.1 XM_017004212.1 XM_017004213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839099 c.257T>C p.V86A Chr.2: 241336014-241336014 on Build GRCh38
3962665 c.341+1G>T p.? Chr.2: 241336099-241336099 on Build GRCh38
4447087 c.341+8C>T p.? Chr.2: 241336106-241336106 on Build GRCh38
6567316 c.270G>C p.E90D Chr.2: 241336027-241336027 on Build GRCh38
222765 c.230G>A p.R77K Chr.2: 241335987-241335987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781171356 Chr.2: 241336050-241336050 on Build GRCh38
rs1017865797 Chr.2: 241335982-241335982 on Build GRCh38
rs765520585 Chr.2: 241336091-241336091 on Build GRCh38
rs988934669 Chr.2: 241335919-241335919 on Build GRCh38
rs147156230 Chr.2: 241336011-241336011 on Build GRCh38
rs890889391 Chr.2: 241336252-241336252 on Build GRCh38
rs754701681 Chr.2: 241336206-241336206 on Build GRCh38
rs372533691 Chr.2: 241336332-241336332 on Build GRCh38
rs942556236 Chr.2: 241336206-241336207 on Build GRCh38
rs573093010 Chr.2: 241336311-241336311 on Build GRCh38
rs997506233 Chr.2: 241336232-241336232 on Build GRCh38
rs1039064824 Chr.2: 241336217-241336217 on Build GRCh38
rs539648563 Chr.2: 241336282-241336282 on Build GRCh38
rs771443480 Chr.2: 241336109-241336109 on Build GRCh38
rs914096448 Chr.2: 241336221-241336221 on Build GRCh38
rs766923305 Chr.2: 241336003-241336003 on Build GRCh38
rs760011491 Chr.2: 241336111-241336111 on Build GRCh38
rs767540919 Chr.2: 241336161-241336161 on Build GRCh38
rs775144020 Chr.2: 241335954-241335954 on Build GRCh38
rs979741355 Chr.2: 241336045-241336045 on Build GRCh38
rs970827757 Chr.2: 241336363-241336363 on Build GRCh38
rs1007856236 Chr.2: 241335957-241335957 on Build GRCh38
rs886983388 Chr.2: 241336329-241336329 on Build GRCh38
rs780108424 Chr.2: 241336068-241336068 on Build GRCh38
rs6739848 Chr.2: 241336136-241336136 on Build GRCh38
rs945568644 Chr.2: 241336229-241336229 on Build GRCh38
rs749093456 Chr.2: 241336179-241336179 on Build GRCh38
rs899494355 Chr.2: 241336305-241336305 on Build GRCh38
rs534119991 Chr.2: 241335958-241335958 on Build GRCh38
rs769432062 Chr.2: 241335953-241335953 on Build GRCh38
rs191979337 Chr.2: 241336165-241336165 on Build GRCh38
rs1039367260 Chr.2: 241336285-241336285 on Build GRCh38
rs533422343 Chr.2: 241336010-241336010 on Build GRCh38
rs745865551 Chr.2: 241335956-241335957 on Build GRCh38
rs781216770 Chr.2: 241336084-241336084 on Build GRCh38
rs780982655 Chr.2: 241335938-241335938 on Build GRCh38
rs773934980 Chr.2: 241336099-241336099 on Build GRCh38
rs746889013 Chr.2: 241336069-241336069 on Build GRCh38
rs757213659 Chr.2: 241336078-241336078 on Build GRCh38
rs1025207903 Chr.2: 241336333-241336333 on Build GRCh38
rs540560605 Chr.2: 241336323-241336323 on Build GRCh38
rs763401725 Chr.2: 241336131-241336131 on Build GRCh38
rs139680984 Chr.2: 241336307-241336307 on Build GRCh38
rs773916861 Chr.2: 241335970-241335970 on Build GRCh38
rs761299365 Chr.2: 241335972-241335972 on Build GRCh38
rs748239389 Chr.2: 241336062-241336062 on Build GRCh38
rs989742223 Chr.2: 241336202-241336202 on Build GRCh38
rs749076083 Chr.2: 241336094-241336094 on Build GRCh38
rs778141095 Chr.2: 241336062-241336062 on Build GRCh38
rs770971980 Chr.2: 241335935-241335935 on Build GRCh38
rs551578168 Chr.2: 241336118-241336118 on Build GRCh38
rs918324671 Chr.2: 241336133-241336133 on Build GRCh38
rs768355434 Chr.2: 241336095-241336095 on Build GRCh38
rs769745495 Chr.2: 241336088-241336088 on Build GRCh38
rs1034594304 Chr.2: 241336347-241336347 on Build GRCh38
rs926928192 Chr.2: 241336082-241336082 on Build GRCh38
rs575652720 Chr.2: 241336262-241336262 on Build GRCh38
rs758354393 Chr.2: 241335974-241335975 on Build GRCh38
rs1026898956 Chr.2: 241336324-241336324 on Build GRCh38
rs776650757 Chr.2: 241336155-241336155 on Build GRCh38
rs557188476 Chr.2: 241336230-241336230 on Build GRCh38
rs1027260699 Chr.2: 241336105-241336105 on Build GRCh38
rs375338910 Chr.2: 241335926-241335926 on Build GRCh38
rs751788345 Chr.2: 241336139-241336139 on Build GRCh38
rs376514681 Chr.2: 241335942-241335942 on Build GRCh38
rs879652639 Chr.2: 241336292-241336292 on Build GRCh38
rs1025068225 Chr.2: 241336114-241336114 on Build GRCh38
rs769932259 Chr.2: 241336157-241336160 on Build GRCh38
rs776066202 Chr.2: 241336121-241336121 on Build GRCh38
rs777218934 Chr.2: 241336110-241336110 on Build GRCh38
rs780999149 Chr.2: 241335967-241335967 on Build GRCh38
rs758575347 Chr.2: 241335916-241335916 on Build GRCh38
rs765470606 Chr.2: 241336033-241336033 on Build GRCh38
rs968907441 Chr.2: 241335990-241335990 on Build GRCh38
rs753092078 Chr.2: 241336051-241336051 on Build GRCh38
rs747057410 Chr.2: 241335929-241335929 on Build GRCh38
rs769883194 Chr.2: 241336211-241336211 on Build GRCh38
rs778051298 Chr.2: 241335917-241335917 on Build GRCh38
rs747699317 Chr.2: 241336108-241336108 on Build GRCh38
rs41266959 Chr.2: 241336164-241336164 on Build GRCh38
rs745741245 Chr.2: 241336087-241336087 on Build GRCh38
rs768265462 Chr.2: 241335964-241335964 on Build GRCh38

View Full Product Details