Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 346977-347469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929937
Entrez Gene ID
101929937
Gene Name
uncharacterized LOC101929937
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.20: 346266-353538 on Build GRCh38
Transcripts
XM_017028182.1 NM_001323679.1 NM_001323680.1 NM_001323681.1 NM_001323682.1 NM_001323683.1 NM_001323684.1 NM_024958.2 NR_136649.1 XM_011529360.1 XM_011529362.2 XM_017028073.1 XM_017028075.1

Gene Symbol
NRSN2
Entrez Gene ID
80023
Gene Name
neurensin 2
Gene Aliases
C20orf98, dJ1103G7.6
UniGene
Hs.416024
Gene Chromosome Location
Chr.20: 346726-359705 on Build GRCh38
Transcripts
XM_017028182.1 NM_001323679.1 NM_001323680.1 NM_001323681.1 NM_001323682.1 NM_001323683.1 NM_001323684.1 NM_024958.2 NR_136649.1 XM_011529360.1 XM_011529362.2 XM_017028073.1 XM_017028075.1

SNP Detail

SNP ID Chromosome Location
rs374847332 Chr.20: 347425-347425 on Build GRCh38
rs371737730 Chr.20: 347337-347337 on Build GRCh38
rs1056672926 Chr.20: 347317-347317 on Build GRCh38
rs955812067 Chr.20: 347239-347239 on Build GRCh38
rs976242363 Chr.20: 347024-347024 on Build GRCh38
rs1054028961 Chr.20: 347178-347178 on Build GRCh38
rs960994872 Chr.20: 347077-347077 on Build GRCh38
rs904983848 Chr.20: 347160-347160 on Build GRCh38
rs1024009760 Chr.20: 347380-347380 on Build GRCh38
rs984694538 Chr.20: 347008-347008 on Build GRCh38
rs543355934 Chr.20: 347154-347154 on Build GRCh38
rs1032166468 Chr.20: 347025-347025 on Build GRCh38
rs115011160 Chr.20: 347391-347391 on Build GRCh38
rs930229516 Chr.20: 347092-347092 on Build GRCh38
rs997823648 Chr.20: 347427-347427 on Build GRCh38
rs975976729 Chr.20: 347212-347212 on Build GRCh38
rs73891939 Chr.20: 347385-347385 on Build GRCh38
rs974686876 Chr.20: 347072-347072 on Build GRCh38
rs905376766 Chr.20: 347091-347091 on Build GRCh38
rs570920569 Chr.20: 347033-347033 on Build GRCh38
rs1026348587 Chr.20: 347148-347148 on Build GRCh38
rs986782242 Chr.20: 347055-347055 on Build GRCh38
rs947028413 Chr.20: 347263-347263 on Build GRCh38
rs955337347 Chr.20: 347046-347046 on Build GRCh38
rs1005442234 Chr.20: 347023-347023 on Build GRCh38
rs140567143 Chr.20: 347008-347009 on Build GRCh38
rs946997265 Chr.20: 347076-347076 on Build GRCh38
rs974151789 Chr.20: 347349-347349 on Build GRCh38
rs11422468 Chr.20: 347011-347011 on Build GRCh38
rs557192906 Chr.20: 347175-347175 on Build GRCh38
rs146238823 Chr.20: 347104-347104 on Build GRCh38
rs577381988 Chr.20: 347113-347113 on Build GRCh38
rs964912503 Chr.20: 347182-347182 on Build GRCh38
rs1044769221 Chr.20: 347009-347009 on Build GRCh38
rs559369036 Chr.20: 347313-347313 on Build GRCh38
rs545114140 Chr.20: 347355-347355 on Build GRCh38
rs534771156 Chr.20: 347100-347100 on Build GRCh38
rs760521385 Chr.20: 347010-347010 on Build GRCh38
rs964699394 Chr.20: 347421-347422 on Build GRCh38
rs1008267754 Chr.20: 347321-347321 on Build GRCh38
rs951997441 Chr.20: 347158-347158 on Build GRCh38
rs917600662 Chr.20: 347129-347129 on Build GRCh38
rs972810041 Chr.20: 347197-347197 on Build GRCh38
rs911340089 Chr.20: 347056-347056 on Build GRCh38
rs993596369 Chr.20: 347362-347362 on Build GRCh38
rs964154960 Chr.20: 347068-347068 on Build GRCh38
rs920068669 Chr.20: 347075-347075 on Build GRCh38
rs536509111 Chr.20: 347111-347111 on Build GRCh38
rs1044698041 Chr.20: 347159-347159 on Build GRCh38
rs779333612 Chr.20: 347439-347439 on Build GRCh38
rs1018287310 Chr.20: 347330-347330 on Build GRCh38
rs1020301850 Chr.20: 347034-347034 on Build GRCh38
rs949078997 Chr.20: 347138-347138 on Build GRCh38
rs113561249 Chr.20: 347058-347058 on Build GRCh38
rs1009909274 Chr.20: 347249-347249 on Build GRCh38
rs550226638 Chr.20: 347406-347406 on Build GRCh38
rs923106966 Chr.20: 347236-347236 on Build GRCh38
rs1018016774 Chr.20: 347413-347413 on Build GRCh38
rs988506257 Chr.20: 347242-347242 on Build GRCh38
rs542641894 Chr.20: 347302-347302 on Build GRCh38
rs1016870222 Chr.20: 347071-347071 on Build GRCh38
rs1044372164 Chr.20: 347276-347276 on Build GRCh38
rs892263615 Chr.20: 347241-347241 on Build GRCh38
rs954599756 Chr.20: 347311-347311 on Build GRCh38
rs956873298 Chr.20: 347026-347026 on Build GRCh38
rs918196546 Chr.20: 347399-347399 on Build GRCh38
rs368137699 Chr.20: 347278-347278 on Build GRCh38
rs754110598 Chr.20: 347013-347013 on Build GRCh38
rs1030280309 Chr.20: 347257-347257 on Build GRCh38
rs971541528 Chr.20: 347386-347386 on Build GRCh38

View Full Product Details