Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 417610-418113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBCK1
Entrez Gene ID
10616
Gene Name
RANBP2-type and C3HC4-type zinc finger containing 1
Gene Aliases
C20orf18, HOIL-1, HOIL1, PBMEI, PGBM1, RBCK2, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4
UniGene
Hs.247280
Gene Chromosome Location
Chr.20: 408050-432139 on Build GRCh38
Transcripts
NM_001323956.1 NM_001323958.1 NM_006462.5 NM_031229.3 NR_136659.1 XM_005260645.1 XM_011529137.1 XM_011529138.1 XM_011529139.2 XM_011529140.1 XM_017027594.1 XM_017027595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039998 c.331G>A p.E111K Chr.20: 417927-417927 on Build GRCh38
7281181 c.433C>T p.R145W Chr.20: 417903-417903 on Build GRCh38
4098412 c.315G>A p.Q105Q Chr.20: 417785-417785 on Build GRCh38
3785396 c.358C>T p.R120W Chr.20: 417828-417828 on Build GRCh38
4098413 c.189G>A p.Q63Q Chr.20: 417785-417785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011030569 Chr.20: 417695-417695 on Build GRCh38
rs375598489 Chr.20: 417956-417956 on Build GRCh38
rs372591105 Chr.20: 417808-417808 on Build GRCh38
rs369452684 Chr.20: 417964-417964 on Build GRCh38
rs778995926 Chr.20: 417918-417918 on Build GRCh38
rs369877444 Chr.20: 417778-417778 on Build GRCh38
rs762880682 Chr.20: 417726-417726 on Build GRCh38
rs749518640 Chr.20: 417819-417819 on Build GRCh38
rs73567132 Chr.20: 418035-418035 on Build GRCh38
rs775672608 Chr.20: 417707-417707 on Build GRCh38
rs759717818 Chr.20: 418005-418005 on Build GRCh38
rs750834643 Chr.20: 417905-417905 on Build GRCh38
rs745803854 Chr.20: 417943-417943 on Build GRCh38
rs962744015 Chr.20: 417967-417967 on Build GRCh38
rs146801967 Chr.20: 418011-418011 on Build GRCh38
rs755160706 Chr.20: 417981-417981 on Build GRCh38
rs11698154 Chr.20: 417634-417634 on Build GRCh38
rs778086791 Chr.20: 417687-417687 on Build GRCh38
rs766673147 Chr.20: 417976-417976 on Build GRCh38
rs770038981 Chr.20: 417633-417633 on Build GRCh38
rs773087121 Chr.20: 417729-417729 on Build GRCh38
rs748158080 Chr.20: 417919-417919 on Build GRCh38
rs766154575 Chr.20: 417656-417656 on Build GRCh38
rs770095642 Chr.20: 417705-417705 on Build GRCh38
rs770866712 Chr.20: 417935-417935 on Build GRCh38
rs1008968180 Chr.20: 417968-417968 on Build GRCh38
rs746197240 Chr.20: 417801-417801 on Build GRCh38
rs766442885 Chr.20: 418074-418074 on Build GRCh38
rs747538597 Chr.20: 417688-417688 on Build GRCh38
rs62191448 Chr.20: 417955-417955 on Build GRCh38
rs546230153 Chr.20: 417951-417951 on Build GRCh38
rs1000517448 Chr.20: 417907-417908 on Build GRCh38
rs377298288 Chr.20: 417759-417759 on Build GRCh38
rs769517890 Chr.20: 417945-417945 on Build GRCh38
rs139191851 Chr.20: 417871-417871 on Build GRCh38
rs768765441 Chr.20: 417727-417727 on Build GRCh38
rs746145296 Chr.20: 417700-417700 on Build GRCh38
rs753525234 Chr.20: 417658-417658 on Build GRCh38
rs759216580 Chr.20: 417755-417755 on Build GRCh38
rs774464341 Chr.20: 417828-417828 on Build GRCh38
rs907637205 Chr.20: 417868-417868 on Build GRCh38
rs756500607 Chr.20: 417910-417910 on Build GRCh38
rs540263731 Chr.20: 417649-417649 on Build GRCh38
rs754190543 Chr.20: 417979-417979 on Build GRCh38
rs778506636 Chr.20: 417663-417663 on Build GRCh38
rs752266924 Chr.20: 417770-417770 on Build GRCh38
rs771489690 Chr.20: 417696-417696 on Build GRCh38
rs778915066 Chr.20: 417819-417820 on Build GRCh38
rs749518835 Chr.20: 417911-417911 on Build GRCh38
rs752107228 Chr.20: 417669-417669 on Build GRCh38
rs371556646 Chr.20: 417686-417686 on Build GRCh38
rs764728866 Chr.20: 417760-417760 on Build GRCh38
rs1019084055 Chr.20: 417785-417785 on Build GRCh38
rs763562321 Chr.20: 417892-417892 on Build GRCh38
rs920299034 Chr.20: 417821-417821 on Build GRCh38
rs752153553 Chr.20: 417787-417787 on Build GRCh38
rs762539213 Chr.20: 417952-417952 on Build GRCh38
rs951705150 Chr.20: 417826-417826 on Build GRCh38
rs752086794 Chr.20: 417965-417965 on Build GRCh38
rs770815337 Chr.20: 417830-417830 on Build GRCh38
rs746970150 Chr.20: 417829-417829 on Build GRCh38
rs750244391 Chr.20: 417648-417648 on Build GRCh38
rs113928542 Chr.20: 417702-417702 on Build GRCh38
rs890912186 Chr.20: 417845-417845 on Build GRCh38
rs200546199 Chr.20: 417640-417640 on Build GRCh38
rs532542504 Chr.20: 417942-417942 on Build GRCh38
rs762636240 Chr.20: 417882-417882 on Build GRCh38
rs201072906 Chr.20: 417965-417965 on Build GRCh38
rs574468909 Chr.20: 418056-418056 on Build GRCh38
rs921229089 Chr.20: 418010-418010 on Build GRCh38
rs781667012 Chr.20: 417697-417697 on Build GRCh38
rs753580979 Chr.20: 417752-417752 on Build GRCh38
rs996416956 Chr.20: 418012-418012 on Build GRCh38
rs757874797 Chr.20: 417904-417904 on Build GRCh38
rs146325218 Chr.20: 417870-417870 on Build GRCh38
rs755178275 Chr.20: 417914-417914 on Build GRCh38
rs751068368 Chr.20: 417903-417903 on Build GRCh38
rs368668838 Chr.20: 417954-417954 on Build GRCh38
rs568820415 Chr.20: 417796-417796 on Build GRCh38
rs560528051 Chr.20: 417824-417824 on Build GRCh38
rs758405216 Chr.20: 417803-417803 on Build GRCh38
rs200146141 Chr.20: 417848-417848 on Build GRCh38
rs767320205 Chr.20: 417641-417641 on Build GRCh38
rs374913274 Chr.20: 417659-417659 on Build GRCh38
rs952276423 Chr.20: 418054-418054 on Build GRCh38
rs763766234 Chr.20: 417953-417953 on Build GRCh38
rs983538178 Chr.20: 418064-418064 on Build GRCh38
rs941351965 Chr.20: 417650-417650 on Build GRCh38
rs867202046 Chr.20: 417786-417786 on Build GRCh38
rs548526459 Chr.20: 417991-417991 on Build GRCh38
rs780195821 Chr.20: 417812-417812 on Build GRCh38
rs763655919 Chr.20: 417779-417779 on Build GRCh38
rs143871997 Chr.20: 417741-417741 on Build GRCh38
rs766210330 Chr.20: 417749-417749 on Build GRCh38

View Full Product Details