Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 69837869-69838378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC7
Entrez Gene ID
57554
Gene Name
leucine rich repeat containing 7
Gene Aliases
DENSIN
UniGene
Hs.479658
Gene Chromosome Location
Chr.1: 69567130-70152426 on Build GRCh38
Transcripts
NM_020794.2 XM_017001885.1 XM_017001886.1 XM_017001887.1 XM_017001888.1 XM_017001889.1 XM_017001890.1 XM_017001891.1 XM_017001892.1 XM_017001893.1 XM_017001894.1 XM_017001895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911933 c.605G>A p.R202Q Chr.1: 69838241-69838241 on Build GRCh38
292490 c.612G>A p.L204L Chr.1: 69838248-69838248 on Build GRCh38
911932 c.491G>A p.R164Q Chr.1: 69838241-69838241 on Build GRCh38
3772079 c.491G>T p.R164L Chr.1: 69838241-69838241 on Build GRCh38
2240688 c.604C>A p.R202R Chr.1: 69838240-69838240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017806714 Chr.1: 69837961-69837961 on Build GRCh38
rs898859044 Chr.1: 69837955-69837955 on Build GRCh38
rs982344085 Chr.1: 69838111-69838111 on Build GRCh38
rs764781045 Chr.1: 69838245-69838245 on Build GRCh38
rs769915215 Chr.1: 69838190-69838190 on Build GRCh38
rs550485768 Chr.1: 69838162-69838162 on Build GRCh38
rs930549961 Chr.1: 69837957-69837957 on Build GRCh38
rs900578508 Chr.1: 69838022-69838022 on Build GRCh38
rs202175098 Chr.1: 69838241-69838241 on Build GRCh38
rs569456233 Chr.1: 69837953-69837953 on Build GRCh38
rs760485145 Chr.1: 69838203-69838203 on Build GRCh38
rs578183399 Chr.1: 69838132-69838132 on Build GRCh38
rs751936060 Chr.1: 69838310-69838310 on Build GRCh38
rs750931876 Chr.1: 69838328-69838328 on Build GRCh38
rs993723593 Chr.1: 69837910-69837910 on Build GRCh38
rs756200434 Chr.1: 69838333-69838333 on Build GRCh38
rs560214067 Chr.1: 69838236-69838236 on Build GRCh38
rs1031953461 Chr.1: 69837997-69837998 on Build GRCh38
rs542755409 Chr.1: 69838292-69838292 on Build GRCh38
rs762130752 Chr.1: 69838000-69838000 on Build GRCh38
rs770994640 Chr.1: 69838216-69838216 on Build GRCh38
rs1054717323 Chr.1: 69837933-69837933 on Build GRCh38
rs937765240 Chr.1: 69837919-69837919 on Build GRCh38
rs988823138 Chr.1: 69838160-69838160 on Build GRCh38
rs781617715 Chr.1: 69838318-69838318 on Build GRCh38
rs185518275 Chr.1: 69838075-69838075 on Build GRCh38
rs757553288 Chr.1: 69838312-69838312 on Build GRCh38
rs773160289 Chr.1: 69838202-69838202 on Build GRCh38
rs987993441 Chr.1: 69838006-69838006 on Build GRCh38
rs927696257 Chr.1: 69837898-69837898 on Build GRCh38
rs780155492 Chr.1: 69838334-69838334 on Build GRCh38
rs749508203 Chr.1: 69838194-69838194 on Build GRCh38
rs1009488754 Chr.1: 69837952-69837952 on Build GRCh38
rs180781345 Chr.1: 69838183-69838183 on Build GRCh38
rs769180099 Chr.1: 69838027-69838027 on Build GRCh38
rs780201964 Chr.1: 69838192-69838192 on Build GRCh38
rs1008693484 Chr.1: 69837998-69837998 on Build GRCh38
rs759439516 Chr.1: 69838190-69838191 on Build GRCh38
rs766947501 Chr.1: 69838223-69838223 on Build GRCh38
rs749328723 Chr.1: 69838343-69838343 on Build GRCh38
rs996299357 Chr.1: 69838094-69838094 on Build GRCh38
rs767646452 Chr.1: 69838106-69838106 on Build GRCh38
rs763688928 Chr.1: 69838295-69838295 on Build GRCh38
rs975156496 Chr.1: 69837984-69837984 on Build GRCh38
rs745941631 Chr.1: 69838188-69838188 on Build GRCh38
rs532201586 Chr.1: 69838039-69838039 on Build GRCh38
rs1047610047 Chr.1: 69837962-69837962 on Build GRCh38
rs189638972 Chr.1: 69838139-69838139 on Build GRCh38
rs886376077 Chr.1: 69837974-69837974 on Build GRCh38
rs781053627 Chr.1: 69837910-69837910 on Build GRCh38
rs890590966 Chr.1: 69837942-69837942 on Build GRCh38
rs930289931 Chr.1: 69838336-69838336 on Build GRCh38
rs776624843 Chr.1: 69838217-69838217 on Build GRCh38
rs1020510314 Chr.1: 69838164-69838164 on Build GRCh38
rs375015731 Chr.1: 69838240-69838240 on Build GRCh38
rs371575240 Chr.1: 69838201-69838201 on Build GRCh38
rs756570503 Chr.1: 69838172-69838172 on Build GRCh38
rs752332115 Chr.1: 69838266-69838266 on Build GRCh38

View Full Product Details