Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70237170-70237690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRSF11
Entrez Gene ID
9295
Gene Name
serine and arginine rich splicing factor 11
Gene Aliases
NET2, SFRS11, dJ677H15.2, p54
UniGene
Hs.479693
Gene Chromosome Location
Chr.1: 70205682-70252020 on Build GRCh38
Transcripts
NM_001190987.1 NM_004768.3 XM_005271338.3 XM_005271339.3 XM_011542427.2 XM_011542428.2 XM_011542431.2 XM_017002811.1 XM_017002812.1 XM_017002813.1 XM_017002814.1 XM_017002815.1 XM_017002816.1 XM_017002817.1 XM_017002818.1 XM_017002819.1 XM_017002820.1 XM_017002821.1 XM_017002822.1 XM_017002823.1 XM_017002824.1 XM_017002825.1 XM_017002826.1 XM_017002827.1 XM_017002828.1 XM_017002829.1 XM_017002830.1 XM_017002831.1 XM_017002832.1 XM_017002833.1 XM_017002834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3492139 c.644C>A p.S215Y Chr.1: 70237478-70237478 on Build GRCh38
5827982 c.455C>T p.S152L Chr.1: 70237469-70237469 on Build GRCh38
3667577 c.709A>G p.I237V Chr.1: 70237543-70237543 on Build GRCh38
5992291 c.511T>A p.S171T Chr.1: 70237525-70237525 on Build GRCh38
3670165 c.613C>T p.L205F Chr.1: 70237447-70237447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763296948 Chr.1: 70237501-70237501 on Build GRCh38
rs928097687 Chr.1: 70237416-70237416 on Build GRCh38
rs747873308 Chr.1: 70237398-70237398 on Build GRCh38
rs571799835 Chr.1: 70237434-70237434 on Build GRCh38
rs996733508 Chr.1: 70237303-70237303 on Build GRCh38
rs528239981 Chr.1: 70237256-70237256 on Build GRCh38
rs766227757 Chr.1: 70237566-70237566 on Build GRCh38
rs767633845 Chr.1: 70237301-70237301 on Build GRCh38
rs747385054 Chr.1: 70237414-70237414 on Build GRCh38
rs984663162 Chr.1: 70237260-70237260 on Build GRCh38
rs369526035 Chr.1: 70237470-70237470 on Build GRCh38
rs930656718 Chr.1: 70237203-70237203 on Build GRCh38
rs761848968 Chr.1: 70237249-70237249 on Build GRCh38
rs996703809 Chr.1: 70237635-70237635 on Build GRCh38
rs1050575204 Chr.1: 70237382-70237382 on Build GRCh38
rs966406871 Chr.1: 70237659-70237659 on Build GRCh38
rs1010972479 Chr.1: 70237349-70237349 on Build GRCh38
rs867982073 Chr.1: 70237642-70237642 on Build GRCh38
rs777291579 Chr.1: 70237594-70237594 on Build GRCh38
rs529444737 Chr.1: 70237240-70237240 on Build GRCh38
rs184501239 Chr.1: 70237640-70237640 on Build GRCh38
rs930938153 Chr.1: 70237462-70237462 on Build GRCh38
rs764238943 Chr.1: 70237543-70237543 on Build GRCh38
rs865930524 Chr.1: 70237608-70237608 on Build GRCh38
rs866038465 Chr.1: 70237222-70237223 on Build GRCh38
rs920826740 Chr.1: 70237461-70237461 on Build GRCh38
rs549474152 Chr.1: 70237299-70237299 on Build GRCh38
rs192338704 Chr.1: 70237571-70237571 on Build GRCh38
rs760579267 Chr.1: 70237549-70237549 on Build GRCh38
rs777149983 Chr.1: 70237435-70237435 on Build GRCh38
rs886507267 Chr.1: 70237262-70237262 on Build GRCh38
rs149050962 Chr.1: 70237570-70237570 on Build GRCh38
rs372560128 Chr.1: 70237555-70237555 on Build GRCh38
rs775431877 Chr.1: 70237489-70237489 on Build GRCh38
rs757865637 Chr.1: 70237579-70237579 on Build GRCh38
rs754557549 Chr.1: 70237376-70237376 on Build GRCh38
rs923014195 Chr.1: 70237360-70237360 on Build GRCh38
rs750376809 Chr.1: 70237478-70237478 on Build GRCh38
rs1049614876 Chr.1: 70237637-70237637 on Build GRCh38
rs907826611 Chr.1: 70237287-70237287 on Build GRCh38
rs959544016 Chr.1: 70237425-70237425 on Build GRCh38
rs201981666 Chr.1: 70237500-70237500 on Build GRCh38
rs778257010 Chr.1: 70237412-70237412 on Build GRCh38
rs201108984 Chr.1: 70237517-70237517 on Build GRCh38
rs199837001 Chr.1: 70237401-70237401 on Build GRCh38
rs552059237 Chr.1: 70237373-70237373 on Build GRCh38
rs560532152 Chr.1: 70237196-70237196 on Build GRCh38
rs116621146 Chr.1: 70237315-70237315 on Build GRCh38
rs773680379 Chr.1: 70237201-70237201 on Build GRCh38
rs1009015650 Chr.1: 70237268-70237268 on Build GRCh38
rs540530808 Chr.1: 70237288-70237288 on Build GRCh38
rs746322449 Chr.1: 70237450-70237450 on Build GRCh38
rs951919981 Chr.1: 70237243-70237243 on Build GRCh38
rs778264645 Chr.1: 70237377-70237377 on Build GRCh38
rs1028259123 Chr.1: 70237310-70237310 on Build GRCh38
rs573566267 Chr.1: 70237664-70237664 on Build GRCh38
rs139343064 Chr.1: 70237224-70237225 on Build GRCh38
rs907907115 Chr.1: 70237528-70237528 on Build GRCh38
rs903461033 Chr.1: 70237605-70237605 on Build GRCh38
rs947572087 Chr.1: 70237514-70237514 on Build GRCh38
rs753875941 Chr.1: 70237568-70237568 on Build GRCh38
rs538405751 Chr.1: 70237350-70237350 on Build GRCh38
rs1021038236 Chr.1: 70237647-70237647 on Build GRCh38
rs746621297 Chr.1: 70237602-70237602 on Build GRCh38
rs1003316454 Chr.1: 70237197-70237197 on Build GRCh38

View Full Product Details