Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10042997-10043499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKEF1
Entrez Gene ID
63926
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
UniGene
Hs.70903
Gene Chromosome Location
Chr.20: 10034999-10056764 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.658178
Gene Chromosome Location
Chr.20: 10023812-10219506 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096595 c.426C>T p.V142V Chr.20: 10043201-10043201 on Build GRCh38
6599995 c.411A>G p.E137E Chr.20: 10043186-10043186 on Build GRCh38
5655872 c.463A>G p.R155G Chr.20: 10043238-10043238 on Build GRCh38
5391544 c.446G>A p.G149E Chr.20: 10043221-10043221 on Build GRCh38
3371608 c.372G>A p.P124P Chr.20: 10043147-10043147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056528269 Chr.20: 10043164-10043164 on Build GRCh38
rs896392412 Chr.20: 10043030-10043030 on Build GRCh38
rs769984434 Chr.20: 10043099-10043099 on Build GRCh38
rs201507677 Chr.20: 10043313-10043313 on Build GRCh38
rs756391922 Chr.20: 10043268-10043268 on Build GRCh38
rs547299482 Chr.20: 10043419-10043419 on Build GRCh38
rs756090350 Chr.20: 10043154-10043154 on Build GRCh38
rs781618151 Chr.20: 10043085-10043085 on Build GRCh38
rs962826068 Chr.20: 10043128-10043128 on Build GRCh38
rs142793528 Chr.20: 10043225-10043225 on Build GRCh38
rs369966784 Chr.20: 10043113-10043113 on Build GRCh38
rs753658083 Chr.20: 10043135-10043135 on Build GRCh38
rs760095258 Chr.20: 10043127-10043127 on Build GRCh38
rs73069599 Chr.20: 10043459-10043459 on Build GRCh38
rs899912434 Chr.20: 10043280-10043280 on Build GRCh38
rs765914346 Chr.20: 10043134-10043134 on Build GRCh38
rs6118866 Chr.20: 10043305-10043305 on Build GRCh38
rs749717274 Chr.20: 10043158-10043158 on Build GRCh38
rs376465137 Chr.20: 10043262-10043262 on Build GRCh38
rs147439785 Chr.20: 10043282-10043282 on Build GRCh38
rs903649339 Chr.20: 10043044-10043044 on Build GRCh38
rs750663943 Chr.20: 10043153-10043153 on Build GRCh38
rs377375975 Chr.20: 10043163-10043163 on Build GRCh38
rs148656825 Chr.20: 10043199-10043199 on Build GRCh38
rs373440566 Chr.20: 10043229-10043229 on Build GRCh38
rs4813909 Chr.20: 10043041-10043041 on Build GRCh38
rs200517861 Chr.20: 10043241-10043241 on Build GRCh38
rs6118865 Chr.20: 10043235-10043235 on Build GRCh38
rs772488530 Chr.20: 10043177-10043177 on Build GRCh38
rs149577177 Chr.20: 10043169-10043169 on Build GRCh38
rs1042449358 Chr.20: 10043206-10043206 on Build GRCh38
rs772907325 Chr.20: 10043122-10043122 on Build GRCh38
rs945039233 Chr.20: 10043198-10043198 on Build GRCh38
rs994139913 Chr.20: 10043346-10043346 on Build GRCh38
rs766548540 Chr.20: 10043271-10043271 on Build GRCh38
rs114733618 Chr.20: 10043355-10043355 on Build GRCh38
rs191252051 Chr.20: 10043302-10043302 on Build GRCh38
rs144052434 Chr.20: 10043178-10043178 on Build GRCh38
rs964191109 Chr.20: 10043083-10043083 on Build GRCh38
rs778220220 Chr.20: 10043318-10043318 on Build GRCh38
rs974224380 Chr.20: 10043136-10043136 on Build GRCh38
rs745925388 Chr.20: 10043092-10043092 on Build GRCh38
rs374348341 Chr.20: 10043116-10043116 on Build GRCh38
rs759536729 Chr.20: 10043138-10043138 on Build GRCh38
rs1001517576 Chr.20: 10043453-10043453 on Build GRCh38
rs112322825 Chr.20: 10043473-10043473 on Build GRCh38
rs200586932 Chr.20: 10043257-10043257 on Build GRCh38
rs566901868 Chr.20: 10043425-10043425 on Build GRCh38
rs997234581 Chr.20: 10043086-10043086 on Build GRCh38
rs758330608 Chr.20: 10043150-10043150 on Build GRCh38
rs183162477 Chr.20: 10043384-10043384 on Build GRCh38
rs138217597 Chr.20: 10043159-10043159 on Build GRCh38
rs937816950 Chr.20: 10043155-10043155 on Build GRCh38
rs779445622 Chr.20: 10043170-10043170 on Build GRCh38
rs746599255 Chr.20: 10043322-10043322 on Build GRCh38
rs897067600 Chr.20: 10043334-10043334 on Build GRCh38
rs572488472 Chr.20: 10043156-10043156 on Build GRCh38
rs150401286 Chr.20: 10043146-10043146 on Build GRCh38
rs551107724 Chr.20: 10043324-10043324 on Build GRCh38
rs917560281 Chr.20: 10043179-10043179 on Build GRCh38
rs778860842 Chr.20: 10043076-10043076 on Build GRCh38
rs755107512 Chr.20: 10043289-10043289 on Build GRCh38
rs760386428 Chr.20: 10043207-10043207 on Build GRCh38
rs752521390 Chr.20: 10043149-10043149 on Build GRCh38
rs373223253 Chr.20: 10043079-10043079 on Build GRCh38
rs111502963 Chr.20: 10043096-10043096 on Build GRCh38
rs762203545 Chr.20: 10043114-10043114 on Build GRCh38
rs1000719833 Chr.20: 10043052-10043052 on Build GRCh38
rs771205746 Chr.20: 10043104-10043105 on Build GRCh38
rs141987321 Chr.20: 10043145-10043145 on Build GRCh38
rs1049463892 Chr.20: 10043053-10043053 on Build GRCh38
rs41275626 Chr.20: 10043102-10043102 on Build GRCh38
rs748655025 Chr.20: 10043311-10043311 on Build GRCh38
rs745370956 Chr.20: 10043367-10043367 on Build GRCh38
rs1008000160 Chr.20: 10043089-10043089 on Build GRCh38
rs781564609 Chr.20: 10043151-10043151 on Build GRCh38
rs577018485 Chr.20: 10043040-10043040 on Build GRCh38
rs748497727 Chr.20: 10043171-10043171 on Build GRCh38
rs370726207 Chr.20: 10043214-10043214 on Build GRCh38
rs775339142 Chr.20: 10043339-10043339 on Build GRCh38

View Full Product Details