Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10435223-10435729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLX4IP
Entrez Gene ID
128710
Gene Name
SLX4 interacting protein
Gene Aliases
C20orf94, bA204H22.1, bA254M13.1, dJ1099D15.3
UniGene
Hs.668782
Gene Chromosome Location
Chr.20: 10434453-10628138 on Build GRCh38
Transcripts
NM_001009608.2 XM_017027648.1 XM_017027650.1 XM_017027652.1

SNP Detail

SNP ID Chromosome Location
rs952119018 Chr.20: 10435294-10435294 on Build GRCh38
rs910720563 Chr.20: 10435391-10435391 on Build GRCh38
rs1047775999 Chr.20: 10435524-10435524 on Build GRCh38
rs529862891 Chr.20: 10435699-10435699 on Build GRCh38
rs531340721 Chr.20: 10435661-10435663 on Build GRCh38
rs929261349 Chr.20: 10435522-10435522 on Build GRCh38
rs559744398 Chr.20: 10435580-10435580 on Build GRCh38
rs955969363 Chr.20: 10435582-10435582 on Build GRCh38
rs537812386 Chr.20: 10435692-10435692 on Build GRCh38
rs376214358 Chr.20: 10435463-10435463 on Build GRCh38
rs944127267 Chr.20: 10435378-10435378 on Build GRCh38
rs957926049 Chr.20: 10435636-10435636 on Build GRCh38
rs769207478 Chr.20: 10435322-10435322 on Build GRCh38
rs985406546 Chr.20: 10435666-10435666 on Build GRCh38
rs575099026 Chr.20: 10435364-10435364 on Build GRCh38
rs774146854 Chr.20: 10435296-10435296 on Build GRCh38
rs929944385 Chr.20: 10435467-10435467 on Build GRCh38
rs968407448 Chr.20: 10435601-10435601 on Build GRCh38
rs901877239 Chr.20: 10435247-10435247 on Build GRCh38
rs905729900 Chr.20: 10435573-10435573 on Build GRCh38
rs543869013 Chr.20: 10435410-10435410 on Build GRCh38
rs910720016 Chr.20: 10435606-10435606 on Build GRCh38
rs977757372 Chr.20: 10435357-10435357 on Build GRCh38
rs761390267 Chr.20: 10435325-10435325 on Build GRCh38
rs1049598451 Chr.20: 10435281-10435281 on Build GRCh38
rs901830644 Chr.20: 10435494-10435494 on Build GRCh38
rs532747698 Chr.20: 10435415-10435415 on Build GRCh38
rs993561550 Chr.20: 10435287-10435287 on Build GRCh38
rs149335157 Chr.20: 10435320-10435320 on Build GRCh38
rs1032033356 Chr.20: 10435626-10435626 on Build GRCh38
rs758822947 Chr.20: 10435534-10435534 on Build GRCh38
rs988507873 Chr.20: 10435361-10435361 on Build GRCh38
rs781086911 Chr.20: 10435682-10435682 on Build GRCh38
rs1048394273 Chr.20: 10435507-10435507 on Build GRCh38
rs543418579 Chr.20: 10435527-10435527 on Build GRCh38
rs1002775995 Chr.20: 10435579-10435579 on Build GRCh38
rs577128222 Chr.20: 10435431-10435431 on Build GRCh38
rs766878884 Chr.20: 10435354-10435354 on Build GRCh38
rs1001627993 Chr.20: 10435324-10435324 on Build GRCh38
rs563633302 Chr.20: 10435411-10435411 on Build GRCh38
rs979588423 Chr.20: 10435374-10435374 on Build GRCh38
rs1025108383 Chr.20: 10435585-10435585 on Build GRCh38
rs918681662 Chr.20: 10435438-10435438 on Build GRCh38
rs1040138785 Chr.20: 10435436-10435436 on Build GRCh38
rs959136010 Chr.20: 10435338-10435338 on Build GRCh38
rs41280282 Chr.20: 10435290-10435290 on Build GRCh38
rs992346597 Chr.20: 10435347-10435347 on Build GRCh38
rs966819394 Chr.20: 10435355-10435355 on Build GRCh38
rs936451455 Chr.20: 10435257-10435257 on Build GRCh38
rs138380161 Chr.20: 10435259-10435259 on Build GRCh38
rs547323626 Chr.20: 10435662-10435662 on Build GRCh38
rs183171313 Chr.20: 10435326-10435326 on Build GRCh38
rs372989257 Chr.20: 10435362-10435362 on Build GRCh38
rs538261628 Chr.20: 10435269-10435269 on Build GRCh38
rs144594630 Chr.20: 10435603-10435603 on Build GRCh38
rs1021251578 Chr.20: 10435584-10435584 on Build GRCh38
rs545868002 Chr.20: 10435563-10435563 on Build GRCh38
rs768822953 Chr.20: 10435469-10435491 on Build GRCh38
rs764729736 Chr.20: 10435581-10435581 on Build GRCh38
rs1001766826 Chr.20: 10435537-10435537 on Build GRCh38
rs766100074 Chr.20: 10435450-10435450 on Build GRCh38
rs994427333 Chr.20: 10435318-10435318 on Build GRCh38
rs897179880 Chr.20: 10435303-10435303 on Build GRCh38
rs545597701 Chr.20: 10435386-10435399 on Build GRCh38
rs757374772 Chr.20: 10435642-10435642 on Build GRCh38
rs1041047163 Chr.20: 10435468-10435468 on Build GRCh38
rs988412941 Chr.20: 10435583-10435583 on Build GRCh38
rs1022787752 Chr.20: 10435566-10435566 on Build GRCh38
rs772938576 Chr.20: 10435363-10435363 on Build GRCh38
rs564065492 Chr.20: 10435652-10435652 on Build GRCh38
rs761414562 Chr.20: 10435571-10435571 on Build GRCh38
rs555183524 Chr.20: 10435337-10435337 on Build GRCh38

View Full Product Details