Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20070776-20071282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
UniGene
Hs.729168
Gene Chromosome Location
Chr.20: 20052514-20360714 on Build GRCh38
Transcripts
NM_001167816.1 NM_015585.3 XM_005260688.3 XM_005260689.2 XM_005260690.3 XM_005260691.3 XM_005260692.3 XM_005260693.3 XM_005260697.1 XM_011529211.1 XM_011529212.2 XM_011529213.1 XM_017027786.1 XM_017027787.1 XM_017027788.1 XM_017027789.1 XM_017027790.1 XM_017027795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6093058 c.192C>A p.I64I Chr.20: 20070902-20070902 on Build GRCh38
6093059 c.192C>A p.I64I Chr.20: 20070902-20070902 on Build GRCh38
1483480 c.157C>T p.L53F Chr.20: 20070867-20070867 on Build GRCh38
5641656 c.171C>G p.L57L Chr.20: 20070881-20070881 on Build GRCh38
5425087 c.268G>T p.E90* Chr.20: 20070978-20070978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926714235 Chr.20: 20071178-20071178 on Build GRCh38
rs939186471 Chr.20: 20071078-20071078 on Build GRCh38
rs1022747772 Chr.20: 20071164-20071164 on Build GRCh38
rs763397411 Chr.20: 20070963-20070964 on Build GRCh38
rs1049224660 Chr.20: 20071108-20071108 on Build GRCh38
rs774135652 Chr.20: 20070984-20070984 on Build GRCh38
rs528400114 Chr.20: 20071110-20071110 on Build GRCh38
rs778441785 Chr.20: 20070812-20070812 on Build GRCh38
rs145673992 Chr.20: 20070983-20070983 on Build GRCh38
rs764542801 Chr.20: 20070902-20070902 on Build GRCh38
rs776756606 Chr.20: 20070875-20070875 on Build GRCh38
rs769947760 Chr.20: 20071043-20071043 on Build GRCh38
rs763565732 Chr.20: 20071056-20071056 on Build GRCh38
rs1007755250 Chr.20: 20071111-20071111 on Build GRCh38
rs567126937 Chr.20: 20071050-20071050 on Build GRCh38
rs17852602 Chr.20: 20070931-20070931 on Build GRCh38
rs773980997 Chr.20: 20070860-20070860 on Build GRCh38
rs763329044 Chr.20: 20070895-20070895 on Build GRCh38
rs749124582 Chr.20: 20070850-20070850 on Build GRCh38
rs943050733 Chr.20: 20071239-20071239 on Build GRCh38
rs1029922388 Chr.20: 20070976-20070976 on Build GRCh38
rs899339389 Chr.20: 20071104-20071104 on Build GRCh38
rs746021626 Chr.20: 20071038-20071038 on Build GRCh38
rs768506505 Chr.20: 20070858-20070858 on Build GRCh38
rs939406992 Chr.20: 20071185-20071185 on Build GRCh38
rs926495007 Chr.20: 20071064-20071064 on Build GRCh38
rs114511279 Chr.20: 20071020-20071020 on Build GRCh38
rs775382677 Chr.20: 20071049-20071049 on Build GRCh38
rs968071091 Chr.20: 20071172-20071172 on Build GRCh38
rs565978761 Chr.20: 20071213-20071213 on Build GRCh38
rs901606741 Chr.20: 20071249-20071249 on Build GRCh38
rs61746667 Chr.20: 20070881-20070881 on Build GRCh38
rs929213776 Chr.20: 20071093-20071093 on Build GRCh38
rs756189001 Chr.20: 20070835-20070835 on Build GRCh38
rs771107950 Chr.20: 20070870-20070870 on Build GRCh38
rs761578625 Chr.20: 20071101-20071101 on Build GRCh38
rs535313366 Chr.20: 20070934-20070934 on Build GRCh38
rs993574107 Chr.20: 20070827-20070827 on Build GRCh38
rs529366723 Chr.20: 20071226-20071226 on Build GRCh38
rs765170934 Chr.20: 20070891-20070891 on Build GRCh38
rs1029808452 Chr.20: 20071138-20071138 on Build GRCh38
rs1009133246 Chr.20: 20071136-20071136 on Build GRCh38
rs887540683 Chr.20: 20070841-20070841 on Build GRCh38
rs764034853 Chr.20: 20071230-20071230 on Build GRCh38
rs897536229 Chr.20: 20070799-20070799 on Build GRCh38
rs199790525 Chr.20: 20070979-20070979 on Build GRCh38
rs145567908 Chr.20: 20070861-20070861 on Build GRCh38
rs946814971 Chr.20: 20071001-20071001 on Build GRCh38
rs915222583 Chr.20: 20070999-20070999 on Build GRCh38
rs569455548 Chr.20: 20071111-20071111 on Build GRCh38
rs997243995 Chr.20: 20071253-20071253 on Build GRCh38
rs775026148 Chr.20: 20071212-20071212 on Build GRCh38
rs778804883 Chr.20: 20070992-20070992 on Build GRCh38
rs991969555 Chr.20: 20071209-20071209 on Build GRCh38
rs560888927 Chr.20: 20071081-20071081 on Build GRCh38
rs771493333 Chr.20: 20070886-20070886 on Build GRCh38
rs542819991 Chr.20: 20071060-20071060 on Build GRCh38
rs369677182 Chr.20: 20070823-20070823 on Build GRCh38
rs200184 Chr.20: 20071246-20071246 on Build GRCh38
rs115995777 Chr.20: 20070991-20070991 on Build GRCh38
rs747929927 Chr.20: 20071000-20071000 on Build GRCh38
rs373374235 Chr.20: 20070893-20070893 on Build GRCh38
rs988392777 Chr.20: 20071162-20071162 on Build GRCh38
rs957196780 Chr.20: 20071140-20071140 on Build GRCh38
rs570168091 Chr.20: 20071122-20071122 on Build GRCh38
rs750015159 Chr.20: 20070978-20070978 on Build GRCh38
rs35865562 Chr.20: 20070932-20070933 on Build GRCh38
rs761590068 Chr.20: 20071250-20071250 on Build GRCh38
rs74700794 Chr.20: 20071170-20071170 on Build GRCh38
rs564581189 Chr.20: 20071131-20071131 on Build GRCh38
rs897757242 Chr.20: 20071082-20071082 on Build GRCh38
rs555640095 Chr.20: 20070925-20070925 on Build GRCh38
rs147290599 Chr.20: 20070975-20070975 on Build GRCh38
rs749601387 Chr.20: 20070833-20070833 on Build GRCh38
rs780030013 Chr.20: 20070847-20070847 on Build GRCh38
rs550261745 Chr.20: 20071177-20071177 on Build GRCh38
rs964199067 Chr.20: 20070929-20070929 on Build GRCh38
rs1027291486 Chr.20: 20070831-20070831 on Build GRCh38
rs531979699 Chr.20: 20071134-20071134 on Build GRCh38
rs781404397 Chr.20: 20071028-20071028 on Build GRCh38
rs567964794 Chr.20: 20070930-20070930 on Build GRCh38

View Full Product Details