Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21330536-21331046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRN2
Entrez Gene ID
22803
Gene Name
5'-3' exoribonuclease 2
Gene Aliases
-
UniGene
Hs.255932
Gene Chromosome Location
Chr.20: 21303284-21389825 on Build GRCh38
Transcripts
NM_001317960.1 NM_012255.4 XM_017027722.1 XM_017027723.1 XM_017027724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653409 c.486A>G p.P162P Chr.20: 21330539-21330539 on Build GRCh38
4097060 c.524G>A p.R175H Chr.20: 21330653-21330653 on Build GRCh38
1410715 c.539A>T p.D180V Chr.20: 21330668-21330668 on Build GRCh38
2705693 c.541C>T p.R181C Chr.20: 21330670-21330670 on Build GRCh38
4741762 c.547A>C p.N183H Chr.20: 21330676-21330676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913185803 Chr.20: 21330566-21330566 on Build GRCh38
rs370840647 Chr.20: 21330676-21330676 on Build GRCh38
rs746803930 Chr.20: 21330600-21330600 on Build GRCh38
rs912220552 Chr.20: 21331002-21331002 on Build GRCh38
rs749527365 Chr.20: 21330578-21330579 on Build GRCh38
rs765177263 Chr.20: 21330668-21330668 on Build GRCh38
rs374071813 Chr.20: 21330597-21330597 on Build GRCh38
rs559140358 Chr.20: 21330656-21330656 on Build GRCh38
rs973862273 Chr.20: 21330857-21330857 on Build GRCh38
rs867332455 Chr.20: 21330670-21330670 on Build GRCh38
rs186081042 Chr.20: 21330651-21330651 on Build GRCh38
rs142162270 Chr.20: 21330622-21330622 on Build GRCh38
rs943953757 Chr.20: 21330700-21330700 on Build GRCh38
rs565123287 Chr.20: 21330775-21330775 on Build GRCh38
rs775089193 Chr.20: 21330647-21330647 on Build GRCh38
rs762477878 Chr.20: 21330649-21330649 on Build GRCh38
rs770933289 Chr.20: 21330606-21330606 on Build GRCh38
rs755043756 Chr.20: 21330570-21330570 on Build GRCh38
rs992404087 Chr.20: 21330809-21330809 on Build GRCh38
rs919698082 Chr.20: 21330910-21330910 on Build GRCh38
rs964058306 Chr.20: 21330973-21330973 on Build GRCh38
rs960988086 Chr.20: 21330797-21330797 on Build GRCh38
rs961818861 Chr.20: 21330850-21330850 on Build GRCh38
rs887388485 Chr.20: 21330862-21330862 on Build GRCh38
rs769166293 Chr.20: 21330632-21330632 on Build GRCh38
rs754308794 Chr.20: 21330661-21330661 on Build GRCh38
rs904247329 Chr.20: 21330665-21330665 on Build GRCh38
rs1005479405 Chr.20: 21330895-21330895 on Build GRCh38
rs1029779362 Chr.20: 21331011-21331011 on Build GRCh38
rs748291751 Chr.20: 21330575-21330575 on Build GRCh38
rs373066883 Chr.20: 21330926-21330926 on Build GRCh38
rs757089878 Chr.20: 21330690-21330690 on Build GRCh38
rs745842377 Chr.20: 21330609-21330609 on Build GRCh38
rs190951556 Chr.20: 21330760-21330760 on Build GRCh38
rs771259074 Chr.20: 21330752-21330752 on Build GRCh38
rs781183018 Chr.20: 21330706-21330706 on Build GRCh38
rs769804845 Chr.20: 21330722-21330722 on Build GRCh38
rs550700099 Chr.20: 21330816-21330816 on Build GRCh38
rs780343272 Chr.20: 21330572-21330572 on Build GRCh38
rs112528477 Chr.20: 21331017-21331017 on Build GRCh38
rs752853648 Chr.20: 21330671-21330671 on Build GRCh38
rs779369068 Chr.20: 21330737-21330737 on Build GRCh38
rs777391313 Chr.20: 21330586-21330586 on Build GRCh38
rs987343653 Chr.20: 21331023-21331023 on Build GRCh38
rs530232896 Chr.20: 21331003-21331003 on Build GRCh38
rs372790307 Chr.20: 21330787-21330787 on Build GRCh38
rs6137317 Chr.20: 21330653-21330653 on Build GRCh38
rs375445862 Chr.20: 21330715-21330715 on Build GRCh38
rs758668167 Chr.20: 21330578-21330578 on Build GRCh38
rs748846580 Chr.20: 21330740-21330740 on Build GRCh38
rs985128871 Chr.20: 21330969-21330969 on Build GRCh38
rs769009634 Chr.20: 21330593-21330593 on Build GRCh38
rs776706975 Chr.20: 21330608-21330608 on Build GRCh38
rs751886153 Chr.20: 21330688-21330688 on Build GRCh38
rs199542652 Chr.20: 21330621-21330621 on Build GRCh38
rs572787648 Chr.20: 21330675-21330675 on Build GRCh38
rs747493011 Chr.20: 21330652-21330652 on Build GRCh38
rs1016908334 Chr.20: 21330922-21330922 on Build GRCh38
rs768111860 Chr.20: 21330745-21330745 on Build GRCh38
rs932382972 Chr.20: 21330947-21330947 on Build GRCh38
rs561402161 Chr.20: 21330859-21330859 on Build GRCh38
rs773986077 Chr.20: 21330751-21330751 on Build GRCh38
rs1014459952 Chr.20: 21330793-21330793 on Build GRCh38
rs149456248 Chr.20: 21330811-21330811 on Build GRCh38

View Full Product Details