Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34935498-34935998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSS
Entrez Gene ID
2937
Gene Name
glutathione synthetase
Gene Aliases
GSHS, HEL-S-64p, HEL-S-88n
UniGene
Hs.82327
Gene Chromosome Location
Chr.20: 34928433-34956027 on Build GRCh38
Transcripts
NM_000178.3 NM_001322494.1 NM_001322495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424928 c.800G>A p.R267Q Chr.20: 34935610-34935610 on Build GRCh38
5936598 c.768-4C>T p.? Chr.20: 34935646-34935646 on Build GRCh38
4097839 c.799C>T p.R267W Chr.20: 34935611-34935611 on Build GRCh38
4136844 c.773G>T p.G258V Chr.20: 34935637-34935637 on Build GRCh38
2758404 c.817C>T p.R273C Chr.20: 34935593-34935593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053064174 Chr.20: 34935671-34935671 on Build GRCh38
rs1051776898 Chr.20: 34935795-34935795 on Build GRCh38
rs374630530 Chr.20: 34935961-34935961 on Build GRCh38
rs762102866 Chr.20: 34935695-34935696 on Build GRCh38
rs867601630 Chr.20: 34935620-34935620 on Build GRCh38
rs933853304 Chr.20: 34935736-34935736 on Build GRCh38
rs201032147 Chr.20: 34935532-34935532 on Build GRCh38
rs746499783 Chr.20: 34935607-34935607 on Build GRCh38
rs193080023 Chr.20: 34935530-34935530 on Build GRCh38
rs889247933 Chr.20: 34935807-34935807 on Build GRCh38
rs770335357 Chr.20: 34935610-34935610 on Build GRCh38
rs780105536 Chr.20: 34935546-34935546 on Build GRCh38
rs35051256 Chr.20: 34935594-34935594 on Build GRCh38
rs121909308 Chr.20: 34935611-34935611 on Build GRCh38
rs925828335 Chr.20: 34935691-34935691 on Build GRCh38
rs554473553 Chr.20: 34935658-34935658 on Build GRCh38
rs751409699 Chr.20: 34935946-34935946 on Build GRCh38
rs371625892 Chr.20: 34935649-34935649 on Build GRCh38
rs960534306 Chr.20: 34935894-34935894 on Build GRCh38
rs983499197 Chr.20: 34935811-34935811 on Build GRCh38
rs934598003 Chr.20: 34935699-34935699 on Build GRCh38
rs202063285 Chr.20: 34935675-34935675 on Build GRCh38
rs992997754 Chr.20: 34935918-34935918 on Build GRCh38
rs184506175 Chr.20: 34935645-34935645 on Build GRCh38
rs747688114 Chr.20: 34935592-34935592 on Build GRCh38
rs992590485 Chr.20: 34935674-34935674 on Build GRCh38
rs763921517 Chr.20: 34935735-34935735 on Build GRCh38
rs17092180 Chr.20: 34935718-34935718 on Build GRCh38
rs548744439 Chr.20: 34935816-34935816 on Build GRCh38
rs201359061 Chr.20: 34935572-34935572 on Build GRCh38
rs141290618 Chr.20: 34935923-34935923 on Build GRCh38
rs186477360 Chr.20: 34935680-34935680 on Build GRCh38
rs868130563 Chr.20: 34935580-34935580 on Build GRCh38
rs566568086 Chr.20: 34935812-34935812 on Build GRCh38
rs1027349172 Chr.20: 34935732-34935732 on Build GRCh38
rs761895365 Chr.20: 34935678-34935678 on Build GRCh38
rs749141246 Chr.20: 34935551-34935551 on Build GRCh38
rs1037716620 Chr.20: 34935858-34935858 on Build GRCh38
rs756073588 Chr.20: 34935539-34935539 on Build GRCh38
rs992968304 Chr.20: 34935584-34935584 on Build GRCh38
rs750506249 Chr.20: 34935681-34935681 on Build GRCh38
rs75143849 Chr.20: 34935773-34935773 on Build GRCh38
rs1015861814 Chr.20: 34935836-34935836 on Build GRCh38
rs1008742699 Chr.20: 34935676-34935676 on Build GRCh38
rs900343192 Chr.20: 34935705-34935705 on Build GRCh38
rs978867821 Chr.20: 34935962-34935962 on Build GRCh38
rs950437862 Chr.20: 34935778-34935778 on Build GRCh38
rs959755710 Chr.20: 34935664-34935664 on Build GRCh38
rs916120128 Chr.20: 34935952-34935952 on Build GRCh38
rs7363022 Chr.20: 34935717-34935717 on Build GRCh38
rs144955088 Chr.20: 34935593-34935593 on Build GRCh38

View Full Product Details