Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41179945-41180477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZHX3
Entrez Gene ID
23051
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
TIX1
UniGene
Hs.380133
Gene Chromosome Location
Chr.20: 41178449-41317606 on Build GRCh38
Transcripts
NM_015035.3 XM_005260341.3 XM_011528720.1 XM_017027737.1 XM_017027738.1

SNP Detail

SNP ID Chromosome Location
rs896002792 Chr.20: 41180071-41180071 on Build GRCh38
rs371187746 Chr.20: 41180167-41180167 on Build GRCh38
rs982170878 Chr.20: 41180341-41180341 on Build GRCh38
rs554782689 Chr.20: 41180000-41180000 on Build GRCh38
rs952981509 Chr.20: 41180247-41180247 on Build GRCh38
rs34630169 Chr.20: 41180015-41180015 on Build GRCh38
rs1014462142 Chr.20: 41180385-41180385 on Build GRCh38
rs147985105 Chr.20: 41179991-41179991 on Build GRCh38
rs573606101 Chr.20: 41180187-41180187 on Build GRCh38
rs996065663 Chr.20: 41180106-41180106 on Build GRCh38
rs751461620 Chr.20: 41180205-41180205 on Build GRCh38
rs930565280 Chr.20: 41180289-41180289 on Build GRCh38
rs773819871 Chr.20: 41180121-41180121 on Build GRCh38
rs542871870 Chr.20: 41180112-41180112 on Build GRCh38
rs760554944 Chr.20: 41180012-41180015 on Build GRCh38
rs771187127 Chr.20: 41180414-41180414 on Build GRCh38
rs1049229743 Chr.20: 41180311-41180311 on Build GRCh38
rs2235366 Chr.20: 41180020-41180020 on Build GRCh38
rs1027088384 Chr.20: 41180229-41180229 on Build GRCh38
rs967646198 Chr.20: 41180178-41180178 on Build GRCh38
rs1053597773 Chr.20: 41180058-41180058 on Build GRCh38
rs562731524 Chr.20: 41180315-41180315 on Build GRCh38
rs902250524 Chr.20: 41180043-41180043 on Build GRCh38
rs755744352 Chr.20: 41180412-41180412 on Build GRCh38
rs757150369 Chr.20: 41180347-41180347 on Build GRCh38
rs147702119 Chr.20: 41180027-41180028 on Build GRCh38
rs931547085 Chr.20: 41180023-41180023 on Build GRCh38
rs916221752 Chr.20: 41180026-41180027 on Build GRCh38
rs1008643498 Chr.20: 41180129-41180129 on Build GRCh38
rs923734516 Chr.20: 41180349-41180349 on Build GRCh38
rs556307128 Chr.20: 41180302-41180302 on Build GRCh38
rs1028334601 Chr.20: 41180108-41180108 on Build GRCh38
rs879761324 Chr.20: 41180034-41180034 on Build GRCh38
rs548839561 Chr.20: 41180026-41180027 on Build GRCh38
rs6093451 Chr.20: 41180276-41180276 on Build GRCh38
rs1046109699 Chr.20: 41180037-41180037 on Build GRCh38
rs1056236620 Chr.20: 41180357-41180357 on Build GRCh38
rs797005186 Chr.20: 41180226-41180226 on Build GRCh38
rs369778715 Chr.20: 41180346-41180346 on Build GRCh38
rs919211059 Chr.20: 41180287-41180287 on Build GRCh38
rs940536481 Chr.20: 41180062-41180062 on Build GRCh38
rs937647192 Chr.20: 41180353-41180353 on Build GRCh38
rs543669171 Chr.20: 41180030-41180030 on Build GRCh38
rs184482897 Chr.20: 41180350-41180350 on Build GRCh38
rs1056204139 Chr.20: 41180052-41180052 on Build GRCh38
rs777267377 Chr.20: 41180438-41180438 on Build GRCh38
rs971869481 Chr.20: 41180259-41180259 on Build GRCh38
rs779724099 Chr.20: 41180439-41180439 on Build GRCh38
rs953478197 Chr.20: 41180224-41180224 on Build GRCh38
rs986191175 Chr.20: 41180235-41180235 on Build GRCh38
rs1053631111 Chr.20: 41180352-41180352 on Build GRCh38
rs1014399919 Chr.20: 41180137-41180137 on Build GRCh38
rs949113815 Chr.20: 41180324-41180324 on Build GRCh38
rs533687858 Chr.20: 41180024-41180024 on Build GRCh38
rs911943118 Chr.20: 41180239-41180239 on Build GRCh38
rs896200731 Chr.20: 41180377-41180377 on Build GRCh38
rs930344318 Chr.20: 41179997-41179997 on Build GRCh38
rs1000468986 Chr.20: 41180424-41180424 on Build GRCh38
rs553851657 Chr.20: 41180136-41180136 on Build GRCh38
rs990999932 Chr.20: 41180266-41180268 on Build GRCh38
rs1020586234 Chr.20: 41180175-41180175 on Build GRCh38
rs1000632686 Chr.20: 41180186-41180186 on Build GRCh38
rs903455156 Chr.20: 41180418-41180418 on Build GRCh38
rs41277002 Chr.20: 41180387-41180387 on Build GRCh38
rs763162641 Chr.20: 41180078-41180078 on Build GRCh38
rs542819539 Chr.20: 41180249-41180249 on Build GRCh38

View Full Product Details