Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63215568-63216114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YTHDF1
Entrez Gene ID
54915
Gene Name
YTH N6-methyladenosine RNA binding protein 1
Gene Aliases
C20orf21
UniGene
Hs.744852
Gene Chromosome Location
Chr.20: 63195429-63216186 on Build GRCh38
Transcripts
NM_017798.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1029033 c.47A>C p.N16T Chr.20: 63215582-63215582 on Build GRCh38
6824557 c.29G>T p.R10I Chr.20: 63215600-63215600 on Build GRCh38
6247751 c.28-3C>A p.? Chr.20: 63215604-63215604 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs889795672 Chr.20: 63215722-63215722 on Build GRCh38
rs1025492807 Chr.20: 63215770-63215770 on Build GRCh38
rs779485737 Chr.20: 63215642-63215642 on Build GRCh38
rs942797188 Chr.20: 63215959-63215959 on Build GRCh38
rs1041034986 Chr.20: 63215727-63215727 on Build GRCh38
rs929887613 Chr.20: 63216049-63216049 on Build GRCh38
rs768835249 Chr.20: 63215626-63215626 on Build GRCh38
rs45518532 Chr.20: 63215704-63215704 on Build GRCh38
rs1053869932 Chr.20: 63215941-63215941 on Build GRCh38
rs557862312 Chr.20: 63215743-63215743 on Build GRCh38
rs888295253 Chr.20: 63216062-63216062 on Build GRCh38
rs971394433 Chr.20: 63215921-63215921 on Build GRCh38
rs554284238 Chr.20: 63215937-63215937 on Build GRCh38
rs372451844 Chr.20: 63215706-63215706 on Build GRCh38
rs770431884 Chr.20: 63215619-63215619 on Build GRCh38
rs927286268 Chr.20: 63215781-63215781 on Build GRCh38
rs1042458005 Chr.20: 63215947-63215947 on Build GRCh38
rs763919122 Chr.20: 63215641-63215641 on Build GRCh38
rs1013968676 Chr.20: 63216078-63216078 on Build GRCh38
rs1005411243 Chr.20: 63216064-63216064 on Build GRCh38
rs767500809 Chr.20: 63215627-63215627 on Build GRCh38
rs964567630 Chr.20: 63215695-63215695 on Build GRCh38
rs1046992626 Chr.20: 63216052-63216052 on Build GRCh38
rs535901375 Chr.20: 63215913-63215913 on Build GRCh38
rs577008731 Chr.20: 63216005-63216005 on Build GRCh38
rs769394044 Chr.20: 63215597-63215597 on Build GRCh38
rs1017228473 Chr.20: 63215662-63215662 on Build GRCh38
rs527757945 Chr.20: 63215616-63215616 on Build GRCh38
rs116161732 Chr.20: 63215652-63215652 on Build GRCh38
rs978274403 Chr.20: 63215922-63215922 on Build GRCh38
rs370282205 Chr.20: 63215604-63215604 on Build GRCh38
rs942110424 Chr.20: 63215752-63215752 on Build GRCh38
rs755367938 Chr.20: 63215868-63215868 on Build GRCh38
rs1005326467 Chr.20: 63215848-63215848 on Build GRCh38
rs992492748 Chr.20: 63215615-63215615 on Build GRCh38
rs6090310 Chr.20: 63215716-63215716 on Build GRCh38
rs1020724617 Chr.20: 63215645-63215645 on Build GRCh38
rs758664113 Chr.20: 63215651-63215651 on Build GRCh38
rs781773948 Chr.20: 63215874-63215874 on Build GRCh38
rs756471009 Chr.20: 63215709-63215709 on Build GRCh38
rs894247634 Chr.20: 63216066-63216066 on Build GRCh38
rs1024393380 Chr.20: 63216088-63216088 on Build GRCh38
rs1041477017 Chr.20: 63215965-63215965 on Build GRCh38
rs575629572 Chr.20: 63215995-63215995 on Build GRCh38
rs992940794 Chr.20: 63215888-63215888 on Build GRCh38
rs929179315 Chr.20: 63215724-63215724 on Build GRCh38
rs1044377824 Chr.20: 63215767-63215767 on Build GRCh38
rs765335841 Chr.20: 63215646-63215646 on Build GRCh38
rs1034180953 Chr.20: 63216065-63216065 on Build GRCh38
rs1034128539 Chr.20: 63215861-63215861 on Build GRCh38
rs936812076 Chr.20: 63215938-63215938 on Build GRCh38
rs752611045 Chr.20: 63215638-63215638 on Build GRCh38
rs571480926 Chr.20: 63215708-63215708 on Build GRCh38
rs1042982160 Chr.20: 63215691-63215691 on Build GRCh38
rs547245953 Chr.20: 63215742-63215742 on Build GRCh38
rs540064029 Chr.20: 63216029-63216029 on Build GRCh38
rs891996266 Chr.20: 63215613-63215613 on Build GRCh38
rs774199443 Chr.20: 63215621-63215621 on Build GRCh38
rs77101697 Chr.20: 63215606-63215606 on Build GRCh38
rs748692823 Chr.20: 63215901-63215901 on Build GRCh38
rs748589907 Chr.20: 63215937-63215937 on Build GRCh38
rs376339170 Chr.20: 63215639-63215639 on Build GRCh38
rs550048138 Chr.20: 63215705-63215705 on Build GRCh38
rs952486760 Chr.20: 63215834-63215834 on Build GRCh38
rs911320690 Chr.20: 63215946-63215946 on Build GRCh38
rs549502612 Chr.20: 63215649-63215649 on Build GRCh38
rs556799457 Chr.20: 63216093-63216093 on Build GRCh38
rs926949982 Chr.20: 63215611-63215611 on Build GRCh38
rs191723561 Chr.20: 63216071-63216071 on Build GRCh38
rs79500085 Chr.20: 63215774-63215774 on Build GRCh38
rs1007333316 Chr.20: 63215723-63215723 on Build GRCh38

View Full Product Details