Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17608887-17609385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG3
Entrez Gene ID
10950
Gene Name
BTG anti-proliferation factor 3
Gene Aliases
ANA, APRO4, TOB5, TOB55, TOFA
UniGene
Hs.473420
Gene Chromosome Location
Chr.21: 17593650-17612950 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 XM_011529476.2 XM_011529478.2 XM_011529479.1

Gene Symbol
CXADR
Entrez Gene ID
1525
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
CAR, CAR4/6, HCAR
UniGene
Hs.627078
Gene Chromosome Location
Chr.21: 17512906-17636213 on Build GRCh38
Transcripts
NM_001130914.1 NM_006806.4 XM_011529441.1 XM_017028265.1 XM_017028266.1 XM_011529476.2 XM_011529478.2 XM_011529479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4860563 c.73G>C p.E25Q Chr.21: 17609072-17609072 on Build GRCh38
364560 c.66G>A p.L22L Chr.21: 17609079-17609079 on Build GRCh38
6351300 c.21C>T p.A7A Chr.21: 17609124-17609124 on Build GRCh38
221373 c.101T>A p.L34* Chr.21: 17609044-17609044 on Build GRCh38
4996047 c.145G>A p.E49K Chr.21: 17609000-17609000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776852903 Chr.21: 17609302-17609302 on Build GRCh38
rs61754145 Chr.21: 17609049-17609049 on Build GRCh38
rs768550367 Chr.21: 17609143-17609144 on Build GRCh38
rs984678105 Chr.21: 17609243-17609243 on Build GRCh38
rs781416294 Chr.21: 17609182-17609182 on Build GRCh38
rs532697588 Chr.21: 17608913-17608913 on Build GRCh38
rs151224976 Chr.21: 17608989-17608989 on Build GRCh38
rs765106721 Chr.21: 17609150-17609150 on Build GRCh38
rs748108470 Chr.21: 17609336-17609336 on Build GRCh38
rs780461076 Chr.21: 17608926-17608926 on Build GRCh38
rs150362946 Chr.21: 17609078-17609078 on Build GRCh38
rs1036193482 Chr.21: 17609328-17609328 on Build GRCh38
rs111446804 Chr.21: 17609245-17609245 on Build GRCh38
rs879104147 Chr.21: 17609145-17609145 on Build GRCh38
rs748192406 Chr.21: 17608966-17608966 on Build GRCh38
rs1018641058 Chr.21: 17608913-17608913 on Build GRCh38
rs773659238 Chr.21: 17608990-17608990 on Build GRCh38
rs11538274 Chr.21: 17609002-17609002 on Build GRCh38
rs916531752 Chr.21: 17609163-17609163 on Build GRCh38
rs968157664 Chr.21: 17608915-17608915 on Build GRCh38
rs543304479 Chr.21: 17608956-17608956 on Build GRCh38
rs552877473 Chr.21: 17609327-17609327 on Build GRCh38
rs992011999 Chr.21: 17609291-17609291 on Build GRCh38
rs756177207 Chr.21: 17609193-17609193 on Build GRCh38
rs35890691 Chr.21: 17609104-17609104 on Build GRCh38
rs867821244 Chr.21: 17609080-17609080 on Build GRCh38
rs200953650 Chr.21: 17609117-17609117 on Build GRCh38
rs774040887 Chr.21: 17609039-17609039 on Build GRCh38
rs117552129 Chr.21: 17609124-17609124 on Build GRCh38
rs369375684 Chr.21: 17609169-17609169 on Build GRCh38
rs917927843 Chr.21: 17609295-17609295 on Build GRCh38
rs541004196 Chr.21: 17609189-17609189 on Build GRCh38
rs933030652 Chr.21: 17609352-17609352 on Build GRCh38
rs923077765 Chr.21: 17609342-17609342 on Build GRCh38
rs561585495 Chr.21: 17609121-17609121 on Build GRCh38
rs779851913 Chr.21: 17608930-17608931 on Build GRCh38
rs559421947 Chr.21: 17609222-17609222 on Build GRCh38
rs372473598 Chr.21: 17608940-17608940 on Build GRCh38
rs376642429 Chr.21: 17609307-17609308 on Build GRCh38
rs771256877 Chr.21: 17609017-17609017 on Build GRCh38
rs909117776 Chr.21: 17609326-17609326 on Build GRCh38
rs749683422 Chr.21: 17608992-17608992 on Build GRCh38
rs747208895 Chr.21: 17608934-17608934 on Build GRCh38
rs755159087 Chr.21: 17609180-17609180 on Build GRCh38
rs532874033 Chr.21: 17609319-17609319 on Build GRCh38
rs749234042 Chr.21: 17609085-17609087 on Build GRCh38
rs757165408 Chr.21: 17609140-17609140 on Build GRCh38

View Full Product Details