Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 17796482-17797031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf91
Entrez Gene ID
54149
Gene Name
chromosome 21 open reading frame 91
Gene Aliases
C21orf14, C21orf38, CSSG1, EURL, YG81
UniGene
Hs.293811
Gene Chromosome Location
Chr.21: 17788967-17819386 on Build GRCh38
Transcripts
NM_001100420.1 NM_001100421.1 NM_017447.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612363 c.370A>T p.R124W Chr.21: 17796876-17796876 on Build GRCh38
265397 c.597_598insA p.E200fs*6 Chr.21: 17796648-17796649 on Build GRCh38
6992672 c.493A>T p.R165W Chr.21: 17796753-17796753 on Build GRCh38
69935 c.400C>G p.Q134E Chr.21: 17796846-17796846 on Build GRCh38
3549884 c.346C>T p.P116S Chr.21: 17796900-17796900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776870138 Chr.21: 17796883-17796883 on Build GRCh38
rs761370327 Chr.21: 17796754-17796754 on Build GRCh38
rs747048120 Chr.21: 17797004-17797004 on Build GRCh38
rs753405217 Chr.21: 17796644-17796644 on Build GRCh38
rs971282718 Chr.21: 17796873-17796873 on Build GRCh38
rs866847371 Chr.21: 17796749-17796750 on Build GRCh38
rs761751614 Chr.21: 17796997-17796997 on Build GRCh38
rs773263510 Chr.21: 17796569-17796569 on Build GRCh38
rs200019621 Chr.21: 17796577-17796577 on Build GRCh38
rs200192230 Chr.21: 17796695-17796695 on Build GRCh38
rs984979354 Chr.21: 17796650-17796650 on Build GRCh38
rs745977322 Chr.21: 17796837-17796837 on Build GRCh38
rs550131947 Chr.21: 17796599-17796599 on Build GRCh38
rs779914171 Chr.21: 17796673-17796673 on Build GRCh38
rs771201183 Chr.21: 17796700-17796700 on Build GRCh38
rs967665441 Chr.21: 17796922-17796922 on Build GRCh38
rs766700071 Chr.21: 17796797-17796803 on Build GRCh38
rs756842780 Chr.21: 17796648-17796648 on Build GRCh38
rs779318693 Chr.21: 17796967-17796967 on Build GRCh38
rs752456963 Chr.21: 17796958-17796958 on Build GRCh38
rs766341007 Chr.21: 17796778-17796778 on Build GRCh38
rs764345412 Chr.21: 17796738-17796738 on Build GRCh38
rs370559602 Chr.21: 17796726-17796726 on Build GRCh38
rs537781367 Chr.21: 17796988-17796988 on Build GRCh38
rs962158312 Chr.21: 17796812-17796812 on Build GRCh38
rs891717972 Chr.21: 17796833-17796833 on Build GRCh38
rs373695866 Chr.21: 17796909-17796909 on Build GRCh38
rs770072668 Chr.21: 17796554-17796554 on Build GRCh38
rs774577595 Chr.21: 17796701-17796701 on Build GRCh38
rs746069099 Chr.21: 17796982-17796982 on Build GRCh38
rs200770394 Chr.21: 17796601-17796601 on Build GRCh38
rs76202022 Chr.21: 17796639-17796639 on Build GRCh38
rs1047978 Chr.21: 17796838-17796838 on Build GRCh38
rs2824495 Chr.21: 17796901-17796901 on Build GRCh38
rs1047976 Chr.21: 17796896-17796896 on Build GRCh38
rs1002006418 Chr.21: 17796899-17796899 on Build GRCh38
rs764899347 Chr.21: 17796902-17796902 on Build GRCh38
rs77632878 Chr.21: 17796511-17796511 on Build GRCh38
rs908142147 Chr.21: 17796659-17796659 on Build GRCh38
rs1024550360 Chr.21: 17796851-17796851 on Build GRCh38
rs35992524 Chr.21: 17796642-17796642 on Build GRCh38
rs778867079 Chr.21: 17796538-17796538 on Build GRCh38
rs935087463 Chr.21: 17796949-17796949 on Build GRCh38
rs767432023 Chr.21: 17796943-17796943 on Build GRCh38
rs569768776 Chr.21: 17796725-17796725 on Build GRCh38
rs2824494 Chr.21: 17796597-17796597 on Build GRCh38
rs776407247 Chr.21: 17796752-17796752 on Build GRCh38
rs566233057 Chr.21: 17796799-17796799 on Build GRCh38
rs779230611 Chr.21: 17796830-17796830 on Build GRCh38
rs760899581 Chr.21: 17796637-17796637 on Build GRCh38
rs868225553 Chr.21: 17796748-17796749 on Build GRCh38
rs779826361 Chr.21: 17796550-17796550 on Build GRCh38
rs770891578 Chr.21: 17796581-17796581 on Build GRCh38
rs952014448 Chr.21: 17796913-17796913 on Build GRCh38
rs747461880 Chr.21: 17796876-17796876 on Build GRCh38
rs751427763 Chr.21: 17796784-17796784 on Build GRCh38
rs760324934 Chr.21: 17796780-17796781 on Build GRCh38
rs757861893 Chr.21: 17796770-17796770 on Build GRCh38
rs887766056 Chr.21: 17796715-17796715 on Build GRCh38
rs757568024 Chr.21: 17796818-17796818 on Build GRCh38
rs777018700 Chr.21: 17796995-17796997 on Build GRCh38
rs764807830 Chr.21: 17796755-17796755 on Build GRCh38
rs8128004 Chr.21: 17796661-17796661 on Build GRCh38
rs36095555 Chr.21: 17796816-17796816 on Build GRCh38
rs769135295 Chr.21: 17796882-17796882 on Build GRCh38
rs201245548 Chr.21: 17796696-17796696 on Build GRCh38
rs772769196 Chr.21: 17796743-17796744 on Build GRCh38
rs749953082 Chr.21: 17796760-17796760 on Build GRCh38
rs914837838 Chr.21: 17796912-17796912 on Build GRCh38
rs1024352120 Chr.21: 17796563-17796563 on Build GRCh38
rs142700004 Chr.21: 17796643-17796643 on Build GRCh38
rs536630928 Chr.21: 17796526-17796526 on Build GRCh38
rs368816028 Chr.21: 17796961-17796961 on Build GRCh38
rs953696365 Chr.21: 17796852-17796852 on Build GRCh38
rs745785658 Chr.21: 17796546-17796546 on Build GRCh38
rs758342581 Chr.21: 17796667-17796667 on Build GRCh38
rs200420591 Chr.21: 17797007-17797007 on Build GRCh38
rs1032949436 Chr.21: 17796521-17796521 on Build GRCh38
rs560313792 Chr.21: 17796689-17796689 on Build GRCh38
rs768071986 Chr.21: 17796606-17796606 on Build GRCh38
rs753126442 Chr.21: 17796611-17796611 on Build GRCh38
rs778384745 Chr.21: 17796660-17796660 on Build GRCh38
rs754875633 Chr.21: 17796939-17796939 on Build GRCh38
rs547937971 Chr.21: 17796786-17796786 on Build GRCh38
rs776116159 Chr.21: 17796863-17796863 on Build GRCh38
rs746133121 Chr.21: 17796868-17796868 on Build GRCh38
rs371714407 Chr.21: 17796549-17796549 on Build GRCh38
rs754332060 Chr.21: 17796699-17796700 on Build GRCh38
rs759632033 Chr.21: 17796710-17796710 on Build GRCh38
rs771502455 Chr.21: 17796644-17796646 on Build GRCh38
rs761144724 Chr.21: 17796729-17796729 on Build GRCh38
rs999874077 Chr.21: 17796780-17796780 on Build GRCh38
rs202099162 Chr.21: 17796930-17796930 on Build GRCh38
rs377672791 Chr.21: 17796698-17796698 on Build GRCh38

View Full Product Details