Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42071126-42071637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
UniGene
Hs.242520
Gene Chromosome Location
Chr.21: 42062959-42142996 on Build GRCh38
Transcripts
NM_001004416.2 NM_001199527.1 NM_001199528.2 NM_173568.3 XM_017028506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3733771 c.52C>A p.P18T Chr.21: 42071368-42071368 on Build GRCh38
6498533 c.67G>A p.G23S Chr.21: 42071383-42071383 on Build GRCh38
6161251 c.3G>A p.M1I Chr.21: 42071319-42071319 on Build GRCh38
4739187 c.44C>A p.A15D Chr.21: 42071360-42071360 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141113049 Chr.21: 42071529-42071529 on Build GRCh38
rs533235264 Chr.21: 42071454-42071454 on Build GRCh38
rs754921915 Chr.21: 42071295-42071295 on Build GRCh38
rs569078093 Chr.21: 42071587-42071587 on Build GRCh38
rs187477180 Chr.21: 42071494-42071494 on Build GRCh38
rs766174925 Chr.21: 42071383-42071383 on Build GRCh38
rs559880690 Chr.21: 42071299-42071299 on Build GRCh38
rs761424257 Chr.21: 42071370-42071370 on Build GRCh38
rs757516015 Chr.21: 42071410-42071410 on Build GRCh38
rs748339708 Chr.21: 42071284-42071284 on Build GRCh38
rs951596887 Chr.21: 42071201-42071201 on Build GRCh38
rs1025967234 Chr.21: 42071466-42071466 on Build GRCh38
rs183139030 Chr.21: 42071468-42071468 on Build GRCh38
rs753777828 Chr.21: 42071288-42071288 on Build GRCh38
rs965888776 Chr.21: 42071495-42071495 on Build GRCh38
rs773033791 Chr.21: 42071381-42071381 on Build GRCh38
rs924609256 Chr.21: 42071554-42071554 on Build GRCh38
rs766380846 Chr.21: 42071286-42071286 on Build GRCh38
rs758494396 Chr.21: 42071404-42071404 on Build GRCh38
rs918793592 Chr.21: 42071446-42071446 on Build GRCh38
rs573512096 Chr.21: 42071311-42071311 on Build GRCh38
rs575476112 Chr.21: 42071157-42071157 on Build GRCh38
rs968758725 Chr.21: 42071542-42071542 on Build GRCh38
rs200548782 Chr.21: 42071278-42071278 on Build GRCh38
rs1027212346 Chr.21: 42071177-42071177 on Build GRCh38
rs903587420 Chr.21: 42071590-42071590 on Build GRCh38
rs752761983 Chr.21: 42071400-42071400 on Build GRCh38
rs185789208 Chr.21: 42071160-42071160 on Build GRCh38
rs762962060 Chr.21: 42071208-42071208 on Build GRCh38
rs753636502 Chr.21: 42071384-42071384 on Build GRCh38
rs749466443 Chr.21: 42071254-42071254 on Build GRCh38
rs534989906 Chr.21: 42071541-42071541 on Build GRCh38
rs766292154 Chr.21: 42071374-42071374 on Build GRCh38
rs747625743 Chr.21: 42071443-42071443 on Build GRCh38
rs200823234 Chr.21: 42071393-42071393 on Build GRCh38
rs368977312 Chr.21: 42071331-42071331 on Build GRCh38
rs114868134 Chr.21: 42071414-42071414 on Build GRCh38
rs550757197 Chr.21: 42071409-42071409 on Build GRCh38
rs764259799 Chr.21: 42071406-42071406 on Build GRCh38
rs79846567 Chr.21: 42071211-42071211 on Build GRCh38
rs965524194 Chr.21: 42071242-42071242 on Build GRCh38
rs771312275 Chr.21: 42071316-42071316 on Build GRCh38
rs756589973 Chr.21: 42071425-42071425 on Build GRCh38
rs557977546 Chr.21: 42071606-42071606 on Build GRCh38
rs771806147 Chr.21: 42071375-42071375 on Build GRCh38
rs963001147 Chr.21: 42071156-42071156 on Build GRCh38
rs375703715 Chr.21: 42071250-42071250 on Build GRCh38
rs956609575 Chr.21: 42071312-42071312 on Build GRCh38
rs968259804 Chr.21: 42071248-42071248 on Build GRCh38
rs777983951 Chr.21: 42071314-42071314 on Build GRCh38
rs975540490 Chr.21: 42071513-42071513 on Build GRCh38
rs1004863318 Chr.21: 42071480-42071480 on Build GRCh38
rs994412368 Chr.21: 42071204-42071205 on Build GRCh38
rs994534636 Chr.21: 42071445-42071445 on Build GRCh38
rs143381815 Chr.21: 42071588-42071588 on Build GRCh38
rs781604366 Chr.21: 42071411-42071411 on Build GRCh38
rs374098400 Chr.21: 42071333-42071333 on Build GRCh38
rs978753680 Chr.21: 42071549-42071549 on Build GRCh38
rs535758685 Chr.21: 42071516-42071516 on Build GRCh38
rs773848599 Chr.21: 42071362-42071362 on Build GRCh38
rs752785707 Chr.21: 42071305-42071305 on Build GRCh38
rs376886035 Chr.21: 42071330-42071330 on Build GRCh38
rs921276225 Chr.21: 42071246-42071246 on Build GRCh38
rs562313753 Chr.21: 42071382-42071382 on Build GRCh38
rs1019787268 Chr.21: 42071489-42071489 on Build GRCh38
rs192353206 Chr.21: 42071244-42071244 on Build GRCh38
rs747325576 Chr.21: 42071315-42071315 on Build GRCh38
rs780449566 Chr.21: 42071436-42071436 on Build GRCh38
rs577810640 Chr.21: 42071253-42071253 on Build GRCh38

View Full Product Details